Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113733450-113733993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZW10
Entrez Gene ID
9183
Gene Name
zw10 kinetochore protein
Gene Aliases
HZW10, KNTC1AP
UniGene
Hs.503886
Gene Chromosome Location
Chr.11: 113733183-113773763 on Build GRCh38
Transcripts
NM_004724.3 XM_017018558.1 XM_017018559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4145323 c.2266G>T p.E756* Chr.11: 113733768-113733768 on Build GRCh38
6448701 c.2223G>A p.W741* Chr.11: 113733811-113733811 on Build GRCh38
923222 c.2309G>T p.R770I Chr.11: 113733725-113733725 on Build GRCh38
7180305 c.2258C>T p.S753F Chr.11: 113733776-113733776 on Build GRCh38
2103421 c.2284C>T p.R762C Chr.11: 113733750-113733750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540501275 Chr.11: 113733870-113733870 on Build GRCh38
rs144914370 Chr.11: 113733727-113733727 on Build GRCh38
rs767964225 Chr.11: 113733653-113733653 on Build GRCh38
rs751014398 Chr.11: 113733851-113733851 on Build GRCh38
rs781093655 Chr.11: 113733732-113733732 on Build GRCh38
rs143944107 Chr.11: 113733622-113733622 on Build GRCh38
rs866920624 Chr.11: 113733708-113733708 on Build GRCh38
rs750012307 Chr.11: 113733691-113733691 on Build GRCh38
rs749505110 Chr.11: 113733749-113733749 on Build GRCh38
rs145667481 Chr.11: 113733746-113733746 on Build GRCh38
rs911148673 Chr.11: 113733556-113733556 on Build GRCh38
rs564257569 Chr.11: 113733862-113733862 on Build GRCh38
rs553332580 Chr.11: 113733555-113733555 on Build GRCh38
rs1052909148 Chr.11: 113733554-113733554 on Build GRCh38
rs763021444 Chr.11: 113733791-113733791 on Build GRCh38
rs41468448 Chr.11: 113733610-113733610 on Build GRCh38
rs368204880 Chr.11: 113733838-113733838 on Build GRCh38
rs182971822 Chr.11: 113733483-113733483 on Build GRCh38
rs752981812 Chr.11: 113733672-113733672 on Build GRCh38
rs776023969 Chr.11: 113733769-113733769 on Build GRCh38
rs771125718 Chr.11: 113733791-113733792 on Build GRCh38
rs1042212601 Chr.11: 113733641-113733641 on Build GRCh38
rs907353427 Chr.11: 113733661-113733661 on Build GRCh38
rs933829544 Chr.11: 113733511-113733511 on Build GRCh38
rs754464712 Chr.11: 113733861-113733861 on Build GRCh38
rs764386508 Chr.11: 113733782-113733782 on Build GRCh38
rs186989077 Chr.11: 113733649-113733649 on Build GRCh38
rs202014087 Chr.11: 113733675-113733675 on Build GRCh38
rs564542696 Chr.11: 113733750-113733750 on Build GRCh38
rs539759994 Chr.11: 113733535-113733535 on Build GRCh38
rs200095366 Chr.11: 113733748-113733748 on Build GRCh38
rs942637700 Chr.11: 113733606-113733606 on Build GRCh38
rs762590711 Chr.11: 113733836-113733836 on Build GRCh38
rs749563926 Chr.11: 113733797-113733797 on Build GRCh38
rs891630068 Chr.11: 113733566-113733566 on Build GRCh38
rs191581906 Chr.11: 113733910-113733910 on Build GRCh38
rs955489062 Chr.11: 113733765-113733765 on Build GRCh38
rs962756076 Chr.11: 113733853-113733853 on Build GRCh38
rs368619933 Chr.11: 113733526-113733529 on Build GRCh38
rs772478529 Chr.11: 113733766-113733766 on Build GRCh38
rs757919131 Chr.11: 113733696-113733696 on Build GRCh38
rs779527346 Chr.11: 113733713-113733713 on Build GRCh38
rs774634686 Chr.11: 113733801-113733802 on Build GRCh38
rs764899413 Chr.11: 113733683-113733683 on Build GRCh38
rs943044549 Chr.11: 113733863-113733863 on Build GRCh38
rs751581068 Chr.11: 113733725-113733725 on Build GRCh38
rs61749021 Chr.11: 113733781-113733781 on Build GRCh38
rs149006252 Chr.11: 113733733-113733733 on Build GRCh38
rs752997071 Chr.11: 113733865-113733866 on Build GRCh38
rs563624895 Chr.11: 113733515-113733515 on Build GRCh38
rs746434838 Chr.11: 113733751-113733751 on Build GRCh38
rs141123769 Chr.11: 113733523-113733526 on Build GRCh38
rs566067923 Chr.11: 113733480-113733480 on Build GRCh38
rs1017335205 Chr.11: 113733674-113733674 on Build GRCh38
rs1001514606 Chr.11: 113733624-113733624 on Build GRCh38
rs113125167 Chr.11: 113733939-113733939 on Build GRCh38
rs775051835 Chr.11: 113733647-113733647 on Build GRCh38
rs746663586 Chr.11: 113733565-113733565 on Build GRCh38
rs572873112 Chr.11: 113733561-113733561 on Build GRCh38
rs993987367 Chr.11: 113733505-113733518 on Build GRCh38
rs574937454 Chr.11: 113733631-113733631 on Build GRCh38
rs767571699 Chr.11: 113733850-113733850 on Build GRCh38
rs972963129 Chr.11: 113733476-113733476 on Build GRCh38
rs772908829 Chr.11: 113733787-113733787 on Build GRCh38
rs974465916 Chr.11: 113733916-113733916 on Build GRCh38
rs1052797376 Chr.11: 113733956-113733956 on Build GRCh38
rs759679512 Chr.11: 113733830-113733831 on Build GRCh38
rs777584288 Chr.11: 113733532-113733532 on Build GRCh38
rs1016630642 Chr.11: 113733830-113733830 on Build GRCh38
rs555191491 Chr.11: 113733572-113733572 on Build GRCh38
rs773258662 Chr.11: 113733689-113733689 on Build GRCh38
rs557190229 Chr.11: 113733920-113733920 on Build GRCh38
rs946176542 Chr.11: 113733623-113733623 on Build GRCh38

View Full Product Details