Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126273214-126273660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXRED1
Entrez Gene ID
55572
Gene Name
FAD dependent oxidoreductase domain containing 1
Gene Aliases
FP634, H17
UniGene
Hs.317190
Gene Chromosome Location
Chr.11: 126269040-126278132 on Build GRCh38
Transcripts
NM_017547.3 NR_037647.1 NR_037648.1 XM_006718879.3 XM_006718881.3 XM_011542895.2 XM_011542896.2 XM_017018000.1 XM_017018001.1 XM_017018002.1 XM_017018003.1 XM_017018004.1 XM_017018005.1 XM_017018006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4685496 c.465A>G p.A155A Chr.11: 126273425-126273425 on Build GRCh38
3397529 c.429C>T p.A143A Chr.11: 126273347-126273347 on Build GRCh38
5487648 c.388G>A p.V130I Chr.11: 126273348-126273348 on Build GRCh38
3868770 c.478C>T p.L160F Chr.11: 126273396-126273396 on Build GRCh38
5578597 c.424C>T p.P142S Chr.11: 126273384-126273384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004494741 Chr.11: 126273340-126273340 on Build GRCh38
rs759917961 Chr.11: 126273363-126273363 on Build GRCh38
rs772260644 Chr.11: 126273457-126273457 on Build GRCh38
rs766610401 Chr.11: 126273293-126273293 on Build GRCh38
rs192191971 Chr.11: 126273629-126273629 on Build GRCh38
rs142867355 Chr.11: 126273389-126273389 on Build GRCh38
rs748813627 Chr.11: 126273324-126273324 on Build GRCh38
rs112855709 Chr.11: 126273289-126273289 on Build GRCh38
rs746956683 Chr.11: 126273347-126273347 on Build GRCh38
rs755681660 Chr.11: 126273320-126273320 on Build GRCh38
rs762004291 Chr.11: 126273277-126273277 on Build GRCh38
rs761101658 Chr.11: 126273464-126273464 on Build GRCh38
rs774019820 Chr.11: 126273527-126273527 on Build GRCh38
rs375352144 Chr.11: 126273467-126273467 on Build GRCh38
rs367942589 Chr.11: 126273469-126273469 on Build GRCh38
rs34542988 Chr.11: 126273351-126273351 on Build GRCh38
rs770950491 Chr.11: 126273416-126273416 on Build GRCh38
rs886944589 Chr.11: 126273333-126273333 on Build GRCh38
rs1051244681 Chr.11: 126273599-126273599 on Build GRCh38
rs777189187 Chr.11: 126273321-126273321 on Build GRCh38
rs754006850 Chr.11: 126273337-126273337 on Build GRCh38
rs147235743 Chr.11: 126273353-126273353 on Build GRCh38
rs927493267 Chr.11: 126273390-126273390 on Build GRCh38
rs370312170 Chr.11: 126273463-126273463 on Build GRCh38
rs549910800 Chr.11: 126273510-126273510 on Build GRCh38
rs768542986 Chr.11: 126273404-126273404 on Build GRCh38
rs763082363 Chr.11: 126273354-126273354 on Build GRCh38
rs139461794 Chr.11: 126273612-126273612 on Build GRCh38
rs552088922 Chr.11: 126273382-126273382 on Build GRCh38
rs776563561 Chr.11: 126273407-126273407 on Build GRCh38
rs200864616 Chr.11: 126273441-126273441 on Build GRCh38
rs764877662 Chr.11: 126273488-126273488 on Build GRCh38
rs765187255 Chr.11: 126273287-126273287 on Build GRCh38
rs754743956 Chr.11: 126273506-126273506 on Build GRCh38
rs780939514 Chr.11: 126273368-126273368 on Build GRCh38
rs2298454 Chr.11: 126273236-126273236 on Build GRCh38
rs75386903 Chr.11: 126273508-126273508 on Build GRCh38
rs778727697 Chr.11: 126273388-126273388 on Build GRCh38
rs759347514 Chr.11: 126273460-126273460 on Build GRCh38
rs144650025 Chr.11: 126273440-126273440 on Build GRCh38
rs1048170615 Chr.11: 126273529-126273529 on Build GRCh38
rs751985297 Chr.11: 126273295-126273295 on Build GRCh38
rs756921599 Chr.11: 126273329-126273329 on Build GRCh38
rs375413966 Chr.11: 126273309-126273309 on Build GRCh38
rs774500202 Chr.11: 126273359-126273359 on Build GRCh38
rs754319183 Chr.11: 126273476-126273476 on Build GRCh38
rs150510019 Chr.11: 126273601-126273601 on Build GRCh38
rs771941278 Chr.11: 126273335-126273335 on Build GRCh38
rs779847213 Chr.11: 126273501-126273501 on Build GRCh38
rs756631739 Chr.11: 126273367-126273367 on Build GRCh38
rs369712457 Chr.11: 126273494-126273494 on Build GRCh38
rs775507828 Chr.11: 126273276-126273276 on Build GRCh38
rs886893949 Chr.11: 126273548-126273548 on Build GRCh38
rs747688755 Chr.11: 126273434-126273434 on Build GRCh38
rs778707231 Chr.11: 126273330-126273330 on Build GRCh38
rs761436384 Chr.11: 126273479-126273479 on Build GRCh38
rs878971256 Chr.11: 126273308-126273308 on Build GRCh38
rs531412126 Chr.11: 126273366-126273366 on Build GRCh38
rs751317023 Chr.11: 126273504-126273504 on Build GRCh38
rs770896312 Chr.11: 126273526-126273526 on Build GRCh38
rs745617116 Chr.11: 126273551-126273551 on Build GRCh38
rs780345055 Chr.11: 126273331-126273331 on Build GRCh38
rs775595762 Chr.11: 126273341-126273341 on Build GRCh38
rs755266959 Chr.11: 126273306-126273306 on Build GRCh38
rs746806442 Chr.11: 126273402-126273402 on Build GRCh38
rs749927495 Chr.11: 126273489-126273489 on Build GRCh38
rs750482528 Chr.11: 126273288-126273288 on Build GRCh38
rs1029510972 Chr.11: 126273260-126273260 on Build GRCh38
rs201873350 Chr.11: 126273355-126273355 on Build GRCh38
rs953976894 Chr.11: 126273266-126273266 on Build GRCh38
rs756723001 Chr.11: 126273384-126273384 on Build GRCh38
rs863224018 Chr.11: 126273352-126273352 on Build GRCh38
rs749340203 Chr.11: 126273412-126273412 on Build GRCh38
rs61898853 Chr.11: 126273635-126273635 on Build GRCh38
rs1016844031 Chr.11: 126273296-126273296 on Build GRCh38
rs140745629 Chr.11: 126273375-126273375 on Build GRCh38
rs753156614 Chr.11: 126273313-126273313 on Build GRCh38
rs759795078 Chr.11: 126273454-126273454 on Build GRCh38
rs376227631 Chr.11: 126273365-126273365 on Build GRCh38
rs549577651 Chr.11: 126273497-126273497 on Build GRCh38
rs375703314 Chr.11: 126273273-126273273 on Build GRCh38
rs997950693 Chr.11: 126273563-126273563 on Build GRCh38
rs1042072744 Chr.11: 126273512-126273512 on Build GRCh38

View Full Product Details