Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4538323-4538826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf4
Entrez Gene ID
57102
Gene Name
chromosome 12 open reading frame 4
Gene Aliases
-
UniGene
Hs.302977
Gene Chromosome Location
Chr.12: 4487730-4538508 on Build GRCh38
Transcripts
NM_001304811.1 NM_020374.3 XM_005253716.1 XM_006718992.3 NM_001130862.1 NM_006479.4

Gene Symbol
RAD51AP1
Entrez Gene ID
10635
Gene Name
RAD51 associated protein 1
Gene Aliases
PIR51
UniGene
Hs.730696
Gene Chromosome Location
Chr.12: 4538784-4560047 on Build GRCh38
Transcripts
NM_001304811.1 NM_020374.3 XM_005253716.1 XM_006718992.3 NM_001130862.1 NM_006479.4

SNP Detail

SNP ID Chromosome Location
rs556107267 Chr.12: 4538614-4538614 on Build GRCh38
rs905991110 Chr.12: 4538585-4538585 on Build GRCh38
rs568776638 Chr.12: 4538775-4538775 on Build GRCh38
rs1024800195 Chr.12: 4538582-4538582 on Build GRCh38
rs911620310 Chr.12: 4538683-4538683 on Build GRCh38
rs867189793 Chr.12: 4538774-4538774 on Build GRCh38
rs576052843 Chr.12: 4538628-4538628 on Build GRCh38
rs866436504 Chr.12: 4538546-4538546 on Build GRCh38
rs368092310 Chr.12: 4538430-4538430 on Build GRCh38
rs1039283246 Chr.12: 4538414-4538414 on Build GRCh38
rs569422549 Chr.12: 4538435-4538435 on Build GRCh38
rs184310784 Chr.12: 4538556-4538556 on Build GRCh38
rs553027846 Chr.12: 4538384-4538384 on Build GRCh38
rs1014727396 Chr.12: 4538436-4538436 on Build GRCh38
rs889663419 Chr.12: 4538608-4538608 on Build GRCh38
rs536751660 Chr.12: 4538589-4538589 on Build GRCh38
rs146383515 Chr.12: 4538464-4538464 on Build GRCh38
rs919050448 Chr.12: 4538482-4538482 on Build GRCh38
rs1029527270 Chr.12: 4538403-4538403 on Build GRCh38
rs1016852189 Chr.12: 4538596-4538596 on Build GRCh38
rs557616611 Chr.12: 4538753-4538753 on Build GRCh38
rs953932483 Chr.12: 4538408-4538408 on Build GRCh38
rs552125504 Chr.12: 4538538-4538538 on Build GRCh38
rs562944762 Chr.12: 4538366-4538366 on Build GRCh38
rs987602194 Chr.12: 4538509-4538509 on Build GRCh38
rs752701335 Chr.12: 4538598-4538598 on Build GRCh38
rs377375044 Chr.12: 4538344-4538344 on Build GRCh38
rs546809881 Chr.12: 4538459-4538459 on Build GRCh38
rs181074727 Chr.12: 4538539-4538539 on Build GRCh38
rs548655826 Chr.12: 4538488-4538488 on Build GRCh38
rs943000858 Chr.12: 4538519-4538519 on Build GRCh38
rs974491461 Chr.12: 4538723-4538723 on Build GRCh38
rs1015723476 Chr.12: 4538367-4538367 on Build GRCh38
rs994684185 Chr.12: 4538627-4538627 on Build GRCh38
rs891937684 Chr.12: 4538560-4538560 on Build GRCh38
rs568880362 Chr.12: 4538494-4538494 on Build GRCh38
rs1006731691 Chr.12: 4538624-4538624 on Build GRCh38
rs370696783 Chr.12: 4538745-4538745 on Build GRCh38
rs544117591 Chr.12: 4538420-4538420 on Build GRCh38
rs745727178 Chr.12: 4538447-4538447 on Build GRCh38
rs554383039 Chr.12: 4538557-4538557 on Build GRCh38
rs527721103 Chr.12: 4538719-4538719 on Build GRCh38
rs1026248369 Chr.12: 4538640-4538640 on Build GRCh38
rs532941351 Chr.12: 4538457-4538457 on Build GRCh38
rs773606250 Chr.12: 4538426-4538426 on Build GRCh38
rs950587587 Chr.12: 4538645-4538645 on Build GRCh38
rs546115530 Chr.12: 4538457-4538458 on Build GRCh38
rs879476923 Chr.12: 4538443-4538443 on Build GRCh38
rs958241179 Chr.12: 4538451-4538451 on Build GRCh38
rs548850366 Chr.12: 4538740-4538740 on Build GRCh38
rs574043673 Chr.12: 4538576-4538576 on Build GRCh38
rs577909880 Chr.12: 4538798-4538798 on Build GRCh38
rs529032761 Chr.12: 4538469-4538469 on Build GRCh38
rs970447183 Chr.12: 4538450-4538450 on Build GRCh38
rs1014450252 Chr.12: 4538561-4538561 on Build GRCh38
rs76125004 Chr.12: 4538510-4538510 on Build GRCh38
rs963305694 Chr.12: 4538467-4538467 on Build GRCh38
rs552419548 Chr.12: 4538417-4538417 on Build GRCh38
rs987348604 Chr.12: 4538651-4538651 on Build GRCh38
rs565818256 Chr.12: 4538701-4538701 on Build GRCh38
rs997997538 Chr.12: 4538390-4538390 on Build GRCh38

View Full Product Details