Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 15899803-15900225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRAP
Entrez Gene ID
11171
Gene Name
serine/threonine kinase receptor associated protein
Gene Aliases
MAWD, PT-WD, UNRIP
UniGene
Hs.743971
Gene Chromosome Location
Chr.12: 15882354-15903478 on Build GRCh38
Transcripts
NM_007178.3 XM_005253294.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
692993 c.897G>C p.T299T Chr.12: 15900025-15900025 on Build GRCh38
6358689 c.925+5G>C p.? Chr.12: 15900058-15900058 on Build GRCh38
937640 c.896C>T p.T299M Chr.12: 15900024-15900024 on Build GRCh38
2003355 c.837C>T p.L279L Chr.12: 15899965-15899965 on Build GRCh38
3458680 c.786G>A p.K262K Chr.12: 15899914-15899914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916705884 Chr.12: 15900201-15900201 on Build GRCh38
rs764894800 Chr.12: 15899948-15899948 on Build GRCh38
rs920658609 Chr.12: 15900188-15900188 on Build GRCh38
rs757306741 Chr.12: 15900031-15900031 on Build GRCh38
rs749478371 Chr.12: 15900096-15900096 on Build GRCh38
rs116355896 Chr.12: 15900182-15900182 on Build GRCh38
rs750712885 Chr.12: 15900085-15900085 on Build GRCh38
rs368728573 Chr.12: 15899965-15899965 on Build GRCh38
rs192619066 Chr.12: 15899864-15899864 on Build GRCh38
rs148366456 Chr.12: 15899905-15899905 on Build GRCh38
rs769503512 Chr.12: 15900072-15900072 on Build GRCh38
rs374482822 Chr.12: 15899967-15899967 on Build GRCh38
rs140559082 Chr.12: 15900025-15900025 on Build GRCh38
rs964549489 Chr.12: 15900134-15900134 on Build GRCh38
rs192157321 Chr.12: 15900162-15900162 on Build GRCh38
rs777491937 Chr.12: 15900020-15900020 on Build GRCh38
rs930628010 Chr.12: 15899831-15899831 on Build GRCh38
rs377025940 Chr.12: 15899895-15899895 on Build GRCh38
rs958092280 Chr.12: 15900137-15900137 on Build GRCh38
rs536091666 Chr.12: 15899849-15899849 on Build GRCh38
rs553115575 Chr.12: 15899944-15899944 on Build GRCh38
rs577936344 Chr.12: 15899947-15899947 on Build GRCh38
rs377695797 Chr.12: 15900001-15900001 on Build GRCh38
rs758070802 Chr.12: 15899917-15899917 on Build GRCh38
rs865946903 Chr.12: 15900048-15900048 on Build GRCh38
rs745431738 Chr.12: 15899978-15899979 on Build GRCh38
rs374693662 Chr.12: 15899954-15899954 on Build GRCh38
rs3830193 Chr.12: 15899845-15899845 on Build GRCh38
rs776623485 Chr.12: 15899860-15899860 on Build GRCh38
rs750835585 Chr.12: 15899935-15899935 on Build GRCh38
rs571997083 Chr.12: 15899834-15899834 on Build GRCh38
rs545723988 Chr.12: 15899950-15899950 on Build GRCh38
rs775609672 Chr.12: 15899874-15899874 on Build GRCh38
rs775727791 Chr.12: 15900094-15900094 on Build GRCh38
rs1054796992 Chr.12: 15899962-15899962 on Build GRCh38
rs867656774 Chr.12: 15900050-15900050 on Build GRCh38
rs756639412 Chr.12: 15900102-15900102 on Build GRCh38
rs1018750318 Chr.12: 15900115-15900115 on Build GRCh38
rs907796276 Chr.12: 15899839-15899839 on Build GRCh38
rs1008835668 Chr.12: 15900114-15900114 on Build GRCh38
rs113264192 Chr.12: 15899852-15899852 on Build GRCh38
rs781098945 Chr.12: 15900052-15900052 on Build GRCh38
rs753008816 Chr.12: 15900097-15900097 on Build GRCh38
rs34382225 Chr.12: 15900019-15900019 on Build GRCh38
rs188476524 Chr.12: 15900143-15900143 on Build GRCh38
rs1032775941 Chr.12: 15900131-15900131 on Build GRCh38
rs763043813 Chr.12: 15899876-15899876 on Build GRCh38
rs61756202 Chr.12: 15900058-15900058 on Build GRCh38
rs543283895 Chr.12: 15900105-15900105 on Build GRCh38
rs79857037 Chr.12: 15899834-15899834 on Build GRCh38
rs1046343527 Chr.12: 15899837-15899837 on Build GRCh38
rs368464244 Chr.12: 15899885-15899885 on Build GRCh38
rs764097415 Chr.12: 15899881-15899881 on Build GRCh38
rs201320904 Chr.12: 15899845-15899846 on Build GRCh38
rs35710467 Chr.12: 15899865-15899866 on Build GRCh38
rs1019786152 Chr.12: 15900087-15900087 on Build GRCh38

View Full Product Details