Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27719706-27720112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS35
Entrez Gene ID
60488
Gene Name
mitochondrial ribosomal protein S35
Gene Aliases
HDCMD11P, MDS023, MRP-S28, MRPS28
UniGene
Hs.714076
Gene Chromosome Location
Chr.12: 27710773-27756304 on Build GRCh38
Transcripts
NM_001190864.1 NM_021821.3 XM_017019780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5643678 c.346C>G p.P116A Chr.12: 27719832-27719832 on Build GRCh38
5681572 c.373G>T p.A125S Chr.12: 27719859-27719859 on Build GRCh38
3704051 c.342G>A p.L114L Chr.12: 27719828-27719828 on Build GRCh38
6520071 c.340T>A p.L114M Chr.12: 27719826-27719826 on Build GRCh38
7325952 c.330delT p.L112fs*3 Chr.12: 27719816-27719816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766671036 Chr.12: 27719849-27719849 on Build GRCh38
rs768205498 Chr.12: 27719809-27719809 on Build GRCh38
rs1043851795 Chr.12: 27719869-27719869 on Build GRCh38
rs897683923 Chr.12: 27719998-27719998 on Build GRCh38
rs980890343 Chr.12: 27720068-27720068 on Build GRCh38
rs866199998 Chr.12: 27719842-27719842 on Build GRCh38
rs1051107130 Chr.12: 27719943-27719943 on Build GRCh38
rs376571863 Chr.12: 27719887-27719887 on Build GRCh38
rs139780198 Chr.12: 27719890-27719890 on Build GRCh38
rs541378849 Chr.12: 27720060-27720060 on Build GRCh38
rs373978933 Chr.12: 27719891-27719891 on Build GRCh38
rs779779050 Chr.12: 27719910-27719910 on Build GRCh38
rs200802117 Chr.12: 27719851-27719851 on Build GRCh38
rs370001683 Chr.12: 27719815-27719815 on Build GRCh38
rs772682785 Chr.12: 27720054-27720054 on Build GRCh38
rs558566065 Chr.12: 27719924-27719924 on Build GRCh38
rs778865907 Chr.12: 27719770-27719770 on Build GRCh38
rs980474312 Chr.12: 27719751-27719751 on Build GRCh38
rs568090319 Chr.12: 27719888-27719888 on Build GRCh38
rs754918951 Chr.12: 27719919-27719919 on Build GRCh38
rs987460180 Chr.12: 27719959-27719959 on Build GRCh38
rs755870712 Chr.12: 27719902-27719902 on Build GRCh38
rs373440176 Chr.12: 27719833-27719833 on Build GRCh38
rs771753758 Chr.12: 27719828-27719828 on Build GRCh38
rs994688589 Chr.12: 27720008-27720008 on Build GRCh38
rs7342317 Chr.12: 27719971-27719971 on Build GRCh38
rs927584431 Chr.12: 27719776-27719776 on Build GRCh38
rs760980483 Chr.12: 27719837-27719837 on Build GRCh38
rs1008854877 Chr.12: 27719974-27719974 on Build GRCh38
rs537091927 Chr.12: 27719898-27719898 on Build GRCh38
rs761759979 Chr.12: 27719894-27719894 on Build GRCh38
rs554715045 Chr.12: 27719854-27719854 on Build GRCh38
rs758740832 Chr.12: 27719777-27719777 on Build GRCh38
rs565752880 Chr.12: 27719802-27719802 on Build GRCh38
rs749186870 Chr.12: 27719807-27719807 on Build GRCh38
rs920450173 Chr.12: 27720014-27720014 on Build GRCh38
rs969518034 Chr.12: 27719731-27719731 on Build GRCh38
rs745615277 Chr.12: 27719801-27719801 on Build GRCh38
rs749233370 Chr.12: 27719913-27719913 on Build GRCh38
rs1027870955 Chr.12: 27720013-27720013 on Build GRCh38
rs1029452597 Chr.12: 27719894-27719894 on Build GRCh38
rs375862118 Chr.12: 27719771-27719771 on Build GRCh38
rs1014835218 Chr.12: 27719976-27719976 on Build GRCh38
rs553965522 Chr.12: 27719779-27719779 on Build GRCh38

View Full Product Details