Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70521120-70521619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRB
Entrez Gene ID
5787
Gene Name
protein tyrosine phosphatase, receptor type B
Gene Aliases
HPTP-BETA, HPTPB, PTPB, R-PTP-BETA, VEPTP
UniGene
Hs.434375
Gene Chromosome Location
Chr.12: 70516850-70637440 on Build GRCh38
Transcripts
NM_001109754.2 NM_001206971.1 NM_001206972.1 NM_002837.4 XM_017019723.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4890231 c.5975C>A p.P1992Q Chr.12: 70521508-70521508 on Build GRCh38
2100519 c.5974C>T p.P1992S Chr.12: 70521509-70521509 on Build GRCh38
4890229 c.6629C>A p.P2210Q Chr.12: 70521508-70521508 on Build GRCh38
2100517 c.6628C>T p.P2210S Chr.12: 70521509-70521509 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779723023 Chr.12: 70521534-70521534 on Build GRCh38
rs933497006 Chr.12: 70521310-70521310 on Build GRCh38
rs749247128 Chr.12: 70521248-70521248 on Build GRCh38
rs778895437 Chr.12: 70521321-70521321 on Build GRCh38
rs200444338 Chr.12: 70521522-70521522 on Build GRCh38
rs772544125 Chr.12: 70521496-70521496 on Build GRCh38
rs183594297 Chr.12: 70521353-70521353 on Build GRCh38
rs745488202 Chr.12: 70521379-70521379 on Build GRCh38
rs17108283 Chr.12: 70521483-70521483 on Build GRCh38
rs749824154 Chr.12: 70521440-70521440 on Build GRCh38
rs904407600 Chr.12: 70521467-70521467 on Build GRCh38
rs768482966 Chr.12: 70521485-70521485 on Build GRCh38
rs777782529 Chr.12: 70521163-70521163 on Build GRCh38
rs762978426 Chr.12: 70521508-70521508 on Build GRCh38
rs761266073 Chr.12: 70521490-70521490 on Build GRCh38
rs532833023 Chr.12: 70521495-70521495 on Build GRCh38
rs972705851 Chr.12: 70521373-70521373 on Build GRCh38
rs970169551 Chr.12: 70521238-70521238 on Build GRCh38
rs868500092 Chr.12: 70521315-70521315 on Build GRCh38
rs1031432225 Chr.12: 70521187-70521187 on Build GRCh38
rs547897515 Chr.12: 70521569-70521569 on Build GRCh38
rs866820470 Chr.12: 70521305-70521305 on Build GRCh38
rs771306577 Chr.12: 70521443-70521443 on Build GRCh38
rs1023855340 Chr.12: 70521231-70521231 on Build GRCh38
rs772750333 Chr.12: 70521501-70521502 on Build GRCh38
rs957925432 Chr.12: 70521214-70521214 on Build GRCh38
rs369876 Chr.12: 70521567-70521567 on Build GRCh38
rs770849666 Chr.12: 70521290-70521290 on Build GRCh38
rs980587661 Chr.12: 70521240-70521240 on Build GRCh38
rs866883478 Chr.12: 70521494-70521494 on Build GRCh38
rs960011019 Chr.12: 70521362-70521362 on Build GRCh38
rs942045601 Chr.12: 70521330-70521330 on Build GRCh38
rs918858557 Chr.12: 70521403-70521403 on Build GRCh38
rs968410277 Chr.12: 70521147-70521147 on Build GRCh38
rs753797798 Chr.12: 70521555-70521555 on Build GRCh38
rs771709685 Chr.12: 70521380-70521380 on Build GRCh38
rs566535360 Chr.12: 70521588-70521588 on Build GRCh38
rs369488990 Chr.12: 70521544-70521544 on Build GRCh38
rs776416924 Chr.12: 70521488-70521488 on Build GRCh38
rs986281291 Chr.12: 70521568-70521568 on Build GRCh38
rs528692377 Chr.12: 70521421-70521421 on Build GRCh38
rs762412091 Chr.12: 70521528-70521529 on Build GRCh38
rs897934956 Chr.12: 70521395-70521396 on Build GRCh38
rs756457225 Chr.12: 70521441-70521442 on Build GRCh38
rs766236924 Chr.12: 70521518-70521518 on Build GRCh38
rs769414991 Chr.12: 70521485-70521486 on Build GRCh38
rs756243168 Chr.12: 70521548-70521548 on Build GRCh38
rs942497815 Chr.12: 70521471-70521471 on Build GRCh38
rs752943656 Chr.12: 70521545-70521545 on Build GRCh38
rs543119016 Chr.12: 70521227-70521227 on Build GRCh38
rs544744718 Chr.12: 70521438-70521440 on Build GRCh38
rs900868778 Chr.12: 70521507-70521507 on Build GRCh38
rs1030856132 Chr.12: 70521212-70521212 on Build GRCh38
rs931525359 Chr.12: 70521418-70521418 on Build GRCh38
rs564617699 Chr.12: 70521448-70521448 on Build GRCh38
rs759310997 Chr.12: 70521540-70521540 on Build GRCh38
rs73330183 Chr.12: 70521553-70521553 on Build GRCh38
rs140263819 Chr.12: 70521449-70521449 on Build GRCh38
rs770880696 Chr.12: 70521560-70521561 on Build GRCh38

View Full Product Details