Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 117210302-117210810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOS1
Entrez Gene ID
4842
Gene Name
nitric oxide synthase 1
Gene Aliases
IHPS1, N-NOS, NC-NOS, NOS, bNOS, nNOS
UniGene
Hs.654410
Gene Chromosome Location
Chr.12: 117208142-117441843 on Build GRCh38
Transcripts
NM_000620.4 NM_001204213.1 NM_001204214.1 NM_001204218.1 XM_017019347.1

SNP Detail

SNP ID Chromosome Location
rs912505820 Chr.12: 117210561-117210561 on Build GRCh38
rs751885036 Chr.12: 117210494-117210499 on Build GRCh38
rs930572905 Chr.12: 117210675-117210675 on Build GRCh38
rs975239447 Chr.12: 117210452-117210452 on Build GRCh38
rs1016841979 Chr.12: 117210394-117210394 on Build GRCh38
rs533476741 Chr.12: 117210445-117210445 on Build GRCh38
rs886729369 Chr.12: 117210652-117210652 on Build GRCh38
rs1030959578 Chr.12: 117210787-117210787 on Build GRCh38
rs536388333 Chr.12: 117210591-117210591 on Build GRCh38
rs893369925 Chr.12: 117210762-117210762 on Build GRCh38
rs771211871 Chr.12: 117210592-117210592 on Build GRCh38
rs957638347 Chr.12: 117210580-117210580 on Build GRCh38
rs987413001 Chr.12: 117210477-117210477 on Build GRCh38
rs536893456 Chr.12: 117210604-117210604 on Build GRCh38
rs987465352 Chr.12: 117210756-117210756 on Build GRCh38
rs1002512185 Chr.12: 117210342-117210342 on Build GRCh38
rs775811817 Chr.12: 117210736-117210736 on Build GRCh38
rs548222302 Chr.12: 117210582-117210582 on Build GRCh38
rs192063777 Chr.12: 117210412-117210412 on Build GRCh38
rs546549497 Chr.12: 117210647-117210647 on Build GRCh38
rs184716708 Chr.12: 117210579-117210579 on Build GRCh38
rs566527404 Chr.12: 117210689-117210689 on Build GRCh38
rs778662466 Chr.12: 117210461-117210461 on Build GRCh38
rs1040859030 Chr.12: 117210722-117210722 on Build GRCh38
rs1020429419 Chr.12: 117210780-117210780 on Build GRCh38
rs772516637 Chr.12: 117210718-117210718 on Build GRCh38
rs938351356 Chr.12: 117210637-117210637 on Build GRCh38
rs927768453 Chr.12: 117210613-117210613 on Build GRCh38
rs36016891 Chr.12: 117210577-117210577 on Build GRCh38
rs1011836770 Chr.12: 117210774-117210774 on Build GRCh38
rs34018958 Chr.12: 117210526-117210527 on Build GRCh38
rs1029048369 Chr.12: 117210456-117210456 on Build GRCh38
rs78784523 Chr.12: 117210687-117210687 on Build GRCh38
rs567892807 Chr.12: 117210584-117210584 on Build GRCh38
rs756141646 Chr.12: 117210341-117210341 on Build GRCh38
rs761313431 Chr.12: 117210744-117210744 on Build GRCh38
rs901834910 Chr.12: 117210545-117210545 on Build GRCh38
rs1004927679 Chr.12: 117210433-117210433 on Build GRCh38
rs886673452 Chr.12: 117210359-117210359 on Build GRCh38
rs375987172 Chr.12: 117210457-117210457 on Build GRCh38
rs546928600 Chr.12: 117210459-117210459 on Build GRCh38
rs566777423 Chr.12: 117210499-117210499 on Build GRCh38
rs1016481583 Chr.12: 117210706-117210706 on Build GRCh38
rs369901276 Chr.12: 117210697-117210697 on Build GRCh38
rs1040342798 Chr.12: 117210491-117210491 on Build GRCh38
rs1035965092 Chr.12: 117210343-117210343 on Build GRCh38
rs960649538 Chr.12: 117210614-117210614 on Build GRCh38
rs901950484 Chr.12: 117210729-117210729 on Build GRCh38
rs930458276 Chr.12: 117210381-117210381 on Build GRCh38
rs977842701 Chr.12: 117210597-117210597 on Build GRCh38
rs905535525 Chr.12: 117210334-117210334 on Build GRCh38
rs996233626 Chr.12: 117210559-117210559 on Build GRCh38
rs560995789 Chr.12: 117210739-117210739 on Build GRCh38
rs779325120 Chr.12: 117210601-117210601 on Build GRCh38
rs551070627 Chr.12: 117210405-117210405 on Build GRCh38
rs916468447 Chr.12: 117210667-117210667 on Build GRCh38
rs905589662 Chr.12: 117210608-117210608 on Build GRCh38
rs916539077 Chr.12: 117210373-117210373 on Build GRCh38
rs777879198 Chr.12: 117210390-117210390 on Build GRCh38

View Full Product Details