Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20002716-20003257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMYM2
Entrez Gene ID
7750
Gene Name
zinc finger MYM-type containing 2
Gene Aliases
FIM, MYM, RAMP, SCLL, ZNF198
UniGene
Hs.507433
Gene Chromosome Location
Chr.13: 19957412-20091845 on Build GRCh38
Transcripts
NM_001190964.2 NM_001190965.2 NM_003453.4 NM_197968.3 XM_005266520.3 XM_011535216.2 XM_011535222.1 XM_017020728.1 XM_017020729.1 XM_017020731.1 XM_017020732.1 XM_017020733.1 XM_017020734.1 XM_017020735.1 XM_017020736.1 XM_017020737.1 XM_017020738.1 XM_017020739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6827476 c.1113A>T p.K371N Chr.13: 20003115-20003115 on Build GRCh38
1493127 c.1046G>A p.G349E Chr.13: 20003048-20003048 on Build GRCh38
1365719 c.1002_1003insA p.P337fs*41 Chr.13: 20003004-20003005 on Build GRCh38
7288032 c.1022G>A p.G341D Chr.13: 20003024-20003024 on Build GRCh38
7288031 c.1022G>A p.G341D Chr.13: 20003024-20003024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570925827 Chr.13: 20002800-20002800 on Build GRCh38
rs893877001 Chr.13: 20002866-20002866 on Build GRCh38
rs376739207 Chr.13: 20002822-20002824 on Build GRCh38
rs765326720 Chr.13: 20003097-20003097 on Build GRCh38
rs758563671 Chr.13: 20003099-20003099 on Build GRCh38
rs965706987 Chr.13: 20003186-20003186 on Build GRCh38
rs769320844 Chr.13: 20002817-20002817 on Build GRCh38
rs755058792 Chr.13: 20003088-20003088 on Build GRCh38
rs762679425 Chr.13: 20002836-20002836 on Build GRCh38
rs1041214933 Chr.13: 20003184-20003184 on Build GRCh38
rs751218581 Chr.13: 20002873-20002873 on Build GRCh38
rs769559641 Chr.13: 20002990-20002990 on Build GRCh38
rs775068884 Chr.13: 20003004-20003004 on Build GRCh38
rs761272661 Chr.13: 20003071-20003071 on Build GRCh38
rs972670814 Chr.13: 20003189-20003189 on Build GRCh38
rs748814883 Chr.13: 20003011-20003011 on Build GRCh38
rs982528359 Chr.13: 20003156-20003156 on Build GRCh38
rs372928628 Chr.13: 20002897-20002897 on Build GRCh38
rs750999463 Chr.13: 20002910-20002910 on Build GRCh38
rs1035569095 Chr.13: 20003056-20003056 on Build GRCh38
rs773657609 Chr.13: 20003069-20003069 on Build GRCh38
rs1001001416 Chr.13: 20003112-20003112 on Build GRCh38
rs369904406 Chr.13: 20002949-20002949 on Build GRCh38
rs766864570 Chr.13: 20003072-20003072 on Build GRCh38
rs755931391 Chr.13: 20003147-20003147 on Build GRCh38
rs761517805 Chr.13: 20003054-20003054 on Build GRCh38
rs953754467 Chr.13: 20003141-20003141 on Build GRCh38
rs745826727 Chr.13: 20002986-20002986 on Build GRCh38
rs779911890 Chr.13: 20003150-20003150 on Build GRCh38
rs887827743 Chr.13: 20002893-20002893 on Build GRCh38
rs577616940 Chr.13: 20003166-20003166 on Build GRCh38
rs373306390 Chr.13: 20003052-20003052 on Build GRCh38
rs780977775 Chr.13: 20003118-20003118 on Build GRCh38
rs555116535 Chr.13: 20002952-20002952 on Build GRCh38
rs866900182 Chr.13: 20003019-20003019 on Build GRCh38
rs938078096 Chr.13: 20003225-20003225 on Build GRCh38
rs929418473 Chr.13: 20002796-20002796 on Build GRCh38
rs182244678 Chr.13: 20003155-20003155 on Build GRCh38
rs755833688 Chr.13: 20003165-20003165 on Build GRCh38
rs754360031 Chr.13: 20003074-20003074 on Build GRCh38
rs74463120 Chr.13: 20003195-20003195 on Build GRCh38
rs747817307 Chr.13: 20003164-20003164 on Build GRCh38
rs1037303588 Chr.13: 20002764-20002764 on Build GRCh38
rs763585859 Chr.13: 20002845-20002845 on Build GRCh38
rs1048185973 Chr.13: 20003207-20003207 on Build GRCh38
rs368064457 Chr.13: 20003163-20003163 on Build GRCh38
rs537709724 Chr.13: 20003153-20003153 on Build GRCh38
rs17853601 Chr.13: 20002920-20002920 on Build GRCh38
rs552347037 Chr.13: 20002784-20002784 on Build GRCh38
rs567726850 Chr.13: 20002787-20002787 on Build GRCh38
rs61749503 Chr.13: 20002921-20002921 on Build GRCh38
rs762319164 Chr.13: 20002896-20002896 on Build GRCh38
rs943953868 Chr.13: 20002755-20002755 on Build GRCh38
rs370963592 Chr.13: 20002811-20002811 on Build GRCh38
rs768191182 Chr.13: 20003025-20003025 on Build GRCh38
rs370107310 Chr.13: 20003126-20003126 on Build GRCh38
rs544659997 Chr.13: 20003176-20003176 on Build GRCh38
rs909318357 Chr.13: 20003216-20003216 on Build GRCh38
rs748344244 Chr.13: 20002953-20002953 on Build GRCh38
rs377500017 Chr.13: 20003061-20003061 on Build GRCh38
rs566026404 Chr.13: 20002761-20002761 on Build GRCh38
rs752944141 Chr.13: 20003098-20003098 on Build GRCh38
rs756640822 Chr.13: 20002912-20002912 on Build GRCh38
rs780920594 Chr.13: 20002982-20002982 on Build GRCh38
rs774813972 Chr.13: 20002822-20002822 on Build GRCh38
rs185992670 Chr.13: 20003194-20003194 on Build GRCh38

View Full Product Details