Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 13616202-13616741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDPN
Entrez Gene ID
10630
Gene Name
podoplanin
Gene Aliases
AGGRUS, GP36, GP40, Gp38, HT1A-1, OTS8, PA2.26, T1A, T1A-2, T1A2, TI1A
UniGene
Hs.468675
Gene Chromosome Location
Chr.1: 13583757-13617957 on Build GRCh38
Transcripts
NM_001006624.1 NM_001006625.1 NM_006474.4 NM_198389.2 XM_006710295.1

SNP Detail

SNP ID Chromosome Location
rs954272216 Chr.1: 13616527-13616527 on Build GRCh38
rs192057267 Chr.1: 13616666-13616666 on Build GRCh38
rs966221812 Chr.1: 13616543-13616543 on Build GRCh38
rs568242177 Chr.1: 13616452-13616452 on Build GRCh38
rs758706064 Chr.1: 13616686-13616686 on Build GRCh38
rs951923176 Chr.1: 13616567-13616567 on Build GRCh38
rs565602967 Chr.1: 13616323-13616323 on Build GRCh38
rs895732493 Chr.1: 13616413-13616413 on Build GRCh38
rs1007203579 Chr.1: 13616538-13616538 on Build GRCh38
rs148788852 Chr.1: 13616488-13616488 on Build GRCh38
rs775835993 Chr.1: 13616386-13616386 on Build GRCh38
rs1001382614 Chr.1: 13616228-13616228 on Build GRCh38
rs540907602 Chr.1: 13616316-13616316 on Build GRCh38
rs1061610 Chr.1: 13616690-13616690 on Build GRCh38
rs915846760 Chr.1: 13616589-13616589 on Build GRCh38
rs928610404 Chr.1: 13616665-13616665 on Build GRCh38
rs537186651 Chr.1: 13616470-13616470 on Build GRCh38
rs750479090 Chr.1: 13616431-13616431 on Build GRCh38
rs759519035 Chr.1: 13616307-13616312 on Build GRCh38
rs571059382 Chr.1: 13616287-13616287 on Build GRCh38
rs1011822704 Chr.1: 13616237-13616237 on Build GRCh38
rs539741653 Chr.1: 13616664-13616664 on Build GRCh38
rs1033641955 Chr.1: 13616232-13616232 on Build GRCh38
rs559158771 Chr.1: 13616337-13616337 on Build GRCh38
rs1055532557 Chr.1: 13616681-13616683 on Build GRCh38
rs984596614 Chr.1: 13616587-13616587 on Build GRCh38
rs551788560 Chr.1: 13616706-13616706 on Build GRCh38
rs531994846 Chr.1: 13616699-13616699 on Build GRCh38
rs978680731 Chr.1: 13616269-13616269 on Build GRCh38
rs1037377406 Chr.1: 13616660-13616660 on Build GRCh38
rs958227355 Chr.1: 13616479-13616479 on Build GRCh38
rs765435315 Chr.1: 13616415-13616415 on Build GRCh38
rs542490261 Chr.1: 13616619-13616619 on Build GRCh38
rs954969706 Chr.1: 13616574-13616574 on Build GRCh38
rs35443786 Chr.1: 13616396-13616396 on Build GRCh38
rs985725351 Chr.1: 13616276-13616276 on Build GRCh38
rs937212403 Chr.1: 13616677-13616677 on Build GRCh38
rs890065159 Chr.1: 13616693-13616693 on Build GRCh38
rs953051512 Chr.1: 13616272-13616272 on Build GRCh38
rs199702401 Chr.1: 13616394-13616394 on Build GRCh38
rs552057661 Chr.1: 13616322-13616322 on Build GRCh38
rs558901575 Chr.1: 13616708-13616708 on Build GRCh38
rs998935535 Chr.1: 13616412-13616412 on Build GRCh38
rs370991254 Chr.1: 13616339-13616339 on Build GRCh38
rs1018553866 Chr.1: 13616541-13616541 on Build GRCh38
rs142416988 Chr.1: 13616627-13616627 on Build GRCh38
rs375099050 Chr.1: 13616619-13616621 on Build GRCh38
rs12136663 Chr.1: 13616371-13616371 on Build GRCh38
rs11880 Chr.1: 13616236-13616236 on Build GRCh38
rs548618312 Chr.1: 13616338-13616338 on Build GRCh38
rs1053210335 Chr.1: 13616685-13616685 on Build GRCh38
rs116910237 Chr.1: 13616605-13616605 on Build GRCh38
rs539816512 Chr.1: 13616546-13616546 on Build GRCh38
rs965805375 Chr.1: 13616292-13616292 on Build GRCh38
rs1045736293 Chr.1: 13616392-13616392 on Build GRCh38
rs907510805 Chr.1: 13616610-13616610 on Build GRCh38
rs528422791 Chr.1: 13616715-13616715 on Build GRCh38
rs1049866590 Chr.1: 13616692-13616692 on Build GRCh38
rs1028539435 Chr.1: 13616461-13616461 on Build GRCh38
rs893301406 Chr.1: 13616428-13616428 on Build GRCh38
rs1008427547 Chr.1: 13616711-13616711 on Build GRCh38
rs1037956828 Chr.1: 13616351-13616351 on Build GRCh38
rs761016256 Chr.1: 13616315-13616315 on Build GRCh38
rs114455795 Chr.1: 13616258-13616258 on Build GRCh38
rs577312176 Chr.1: 13616544-13616544 on Build GRCh38
rs553719738 Chr.1: 13616547-13616547 on Build GRCh38
rs377549467 Chr.1: 13616257-13616257 on Build GRCh38
rs398089132 Chr.1: 13616397-13616397 on Build GRCh38
rs902480473 Chr.1: 13616687-13616689 on Build GRCh38
rs12131411 Chr.1: 13616407-13616407 on Build GRCh38
rs1032521456 Chr.1: 13616471-13616471 on Build GRCh38
rs948715234 Chr.1: 13616385-13616385 on Build GRCh38

View Full Product Details