Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 39037842-39038337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHLRC3
Entrez Gene ID
387921
Gene Name
NHL repeat containing 3
Gene Aliases
-
UniGene
Hs.507783
Gene Chromosome Location
Chr.13: 39038311-39050109 on Build GRCh38
Transcripts
NM_001012754.3 NM_001017370.2 NR_073109.1 NM_025138.4 XM_005266545.3 XM_011535239.2

Gene Symbol
PROSER1
Entrez Gene ID
80209
Gene Name
proline and serine rich 1
Gene Aliases
C13orf23
UniGene
Hs.318526
Gene Chromosome Location
Chr.13: 39009865-39038115 on Build GRCh38
Transcripts
NM_001012754.3 NM_001017370.2 NR_073109.1 NM_025138.4 XM_005266545.3 XM_011535239.2

SNP Detail

SNP ID Chromosome Location
rs765076141 Chr.13: 39038044-39038044 on Build GRCh38
rs557900995 Chr.13: 39038109-39038109 on Build GRCh38
rs534997287 Chr.13: 39038091-39038091 on Build GRCh38
rs928405540 Chr.13: 39037986-39037986 on Build GRCh38
rs1052471489 Chr.13: 39038274-39038274 on Build GRCh38
rs938394022 Chr.13: 39038002-39038002 on Build GRCh38
rs1039970678 Chr.13: 39038260-39038260 on Build GRCh38
rs1026189118 Chr.13: 39037866-39037866 on Build GRCh38
rs940334527 Chr.13: 39037944-39037944 on Build GRCh38
rs971706043 Chr.13: 39038136-39038136 on Build GRCh38
rs908899709 Chr.13: 39037938-39037938 on Build GRCh38
rs570233740 Chr.13: 39037996-39037996 on Build GRCh38
rs966096158 Chr.13: 39037900-39037900 on Build GRCh38
rs1007137505 Chr.13: 39038022-39038022 on Build GRCh38
rs1025820169 Chr.13: 39038089-39038089 on Build GRCh38
rs970676417 Chr.13: 39038142-39038142 on Build GRCh38
rs951521934 Chr.13: 39038097-39038097 on Build GRCh38
rs935100376 Chr.13: 39037985-39037985 on Build GRCh38
rs763318182 Chr.13: 39038254-39038254 on Build GRCh38
rs889175802 Chr.13: 39038307-39038307 on Build GRCh38
rs373717255 Chr.13: 39037906-39037906 on Build GRCh38
rs1030968971 Chr.13: 39038046-39038046 on Build GRCh38
rs770049877 Chr.13: 39038146-39038146 on Build GRCh38
rs549918507 Chr.13: 39037923-39037923 on Build GRCh38
rs901938503 Chr.13: 39038066-39038066 on Build GRCh38
rs1006499331 Chr.13: 39038121-39038121 on Build GRCh38
rs962297147 Chr.13: 39038135-39038135 on Build GRCh38
rs768514844 Chr.13: 39037984-39037984 on Build GRCh38
rs947349963 Chr.13: 39038012-39038012 on Build GRCh38
rs928267858 Chr.13: 39038166-39038166 on Build GRCh38
rs748059003 Chr.13: 39038062-39038062 on Build GRCh38
rs1005647048 Chr.13: 39038098-39038098 on Build GRCh38
rs764388768 Chr.13: 39038278-39038278 on Build GRCh38
rs998795487 Chr.13: 39038076-39038076 on Build GRCh38
rs997592346 Chr.13: 39038088-39038088 on Build GRCh38
rs959821802 Chr.13: 39038179-39038179 on Build GRCh38
rs903834828 Chr.13: 39037884-39037884 on Build GRCh38
rs532916725 Chr.13: 39037916-39037916 on Build GRCh38
rs1031107563 Chr.13: 39037892-39037892 on Build GRCh38
rs894298280 Chr.13: 39038008-39038008 on Build GRCh38
rs991181685 Chr.13: 39038182-39038182 on Build GRCh38
rs997623296 Chr.13: 39038285-39038285 on Build GRCh38
rs76942057 Chr.13: 39038223-39038223 on Build GRCh38
rs187485155 Chr.13: 39038034-39038034 on Build GRCh38
rs943233 Chr.13: 39038127-39038127 on Build GRCh38
rs985441897 Chr.13: 39037932-39037932 on Build GRCh38
rs953686969 Chr.13: 39037922-39037922 on Build GRCh38
rs915775576 Chr.13: 39038201-39038201 on Build GRCh38
rs771174706 Chr.13: 39038103-39038103 on Build GRCh38
rs924528110 Chr.13: 39038249-39038249 on Build GRCh38
rs982316462 Chr.13: 39038149-39038149 on Build GRCh38
rs1043353058 Chr.13: 39038024-39038024 on Build GRCh38
rs1042874490 Chr.13: 39038243-39038243 on Build GRCh38
rs1053564765 Chr.13: 39038207-39038207 on Build GRCh38
rs536057832 Chr.13: 39038030-39038030 on Build GRCh38
rs1022335869 Chr.13: 39037865-39037865 on Build GRCh38
rs761012751 Chr.13: 39037888-39037888 on Build GRCh38
rs192402194 Chr.13: 39038045-39038045 on Build GRCh38
rs557040192 Chr.13: 39038306-39038306 on Build GRCh38
rs1050925495 Chr.13: 39038291-39038291 on Build GRCh38
rs1006819690 Chr.13: 39038312-39038312 on Build GRCh38
rs916395135 Chr.13: 39037913-39037913 on Build GRCh38
rs1055560169 Chr.13: 39038006-39038006 on Build GRCh38
rs999645147 Chr.13: 39037891-39037891 on Build GRCh38

View Full Product Details