Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45339363-45339867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPT1
Entrez Gene ID
7178
Gene Name
tumor protein, translationally-controlled 1
Gene Aliases
HRF, TCTP, p02, p23
UniGene
Hs.374596
Gene Chromosome Location
Chr.13: 45333471-45341284 on Build GRCh38
Transcripts
NM_001286272.1 NM_001286273.1 NM_003295.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3469037 c.341T>G p.F114C Chr.13: 45339555-45339555 on Build GRCh38
6807063 c.323C>T p.P108L Chr.13: 45339573-45339573 on Build GRCh38
7285058 c.395A>G p.Y132C Chr.13: 45339501-45339501 on Build GRCh38
3711088 c.325G>C p.E109Q Chr.13: 45339571-45339571 on Build GRCh38
4735650 c.360A>G p.E120E Chr.13: 45339536-45339536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941365428 Chr.13: 45339808-45339808 on Build GRCh38
rs930014513 Chr.13: 45339473-45339473 on Build GRCh38
rs559179357 Chr.13: 45339429-45339429 on Build GRCh38
rs11552486 Chr.13: 45339548-45339548 on Build GRCh38
rs1009901881 Chr.13: 45339672-45339672 on Build GRCh38
rs754407523 Chr.13: 45339653-45339653 on Build GRCh38
rs1032287019 Chr.13: 45339444-45339444 on Build GRCh38
rs964312869 Chr.13: 45339739-45339739 on Build GRCh38
rs1007545632 Chr.13: 45339430-45339430 on Build GRCh38
rs768020738 Chr.13: 45339532-45339532 on Build GRCh38
rs564663837 Chr.13: 45339632-45339632 on Build GRCh38
rs1018964202 Chr.13: 45339738-45339738 on Build GRCh38
rs904230467 Chr.13: 45339714-45339714 on Build GRCh38
rs779614359 Chr.13: 45339541-45339541 on Build GRCh38
rs746853682 Chr.13: 45339612-45339619 on Build GRCh38
rs927216558 Chr.13: 45339830-45339830 on Build GRCh38
rs376813272 Chr.13: 45339814-45339814 on Build GRCh38
rs765999550 Chr.13: 45339647-45339647 on Build GRCh38
rs774955456 Chr.13: 45339627-45339627 on Build GRCh38
rs895573990 Chr.13: 45339662-45339662 on Build GRCh38
rs976926130 Chr.13: 45339439-45339439 on Build GRCh38
rs775435717 Chr.13: 45339500-45339500 on Build GRCh38
rs779666671 Chr.13: 45339489-45339490 on Build GRCh38
rs377202639 Chr.13: 45339547-45339547 on Build GRCh38
rs574799894 Chr.13: 45339807-45339807 on Build GRCh38
rs951451946 Chr.13: 45339788-45339788 on Build GRCh38
rs933523093 Chr.13: 45339745-45339748 on Build GRCh38
rs750041917 Chr.13: 45339533-45339533 on Build GRCh38
rs187228058 Chr.13: 45339411-45339411 on Build GRCh38
rs977336687 Chr.13: 45339762-45339762 on Build GRCh38
rs909968144 Chr.13: 45339795-45339795 on Build GRCh38
rs11540940 Chr.13: 45339517-45339517 on Build GRCh38
rs1018519445 Chr.13: 45339415-45339415 on Build GRCh38
rs982785666 Chr.13: 45339489-45339489 on Build GRCh38
rs200488327 Chr.13: 45339626-45339626 on Build GRCh38
rs748115285 Chr.13: 45339557-45339557 on Build GRCh38
rs201079605 Chr.13: 45339516-45339516 on Build GRCh38
rs879852343 Chr.13: 45339835-45339835 on Build GRCh38
rs553437356 Chr.13: 45339750-45339750 on Build GRCh38
rs1009533634 Chr.13: 45339812-45339813 on Build GRCh38
rs552031146 Chr.13: 45339637-45339637 on Build GRCh38
rs751926495 Chr.13: 45339524-45339524 on Build GRCh38
rs769975408 Chr.13: 45339605-45339605 on Build GRCh38
rs182569557 Chr.13: 45339394-45339394 on Build GRCh38
rs755836071 Chr.13: 45339536-45339536 on Build GRCh38
rs922705594 Chr.13: 45339461-45339461 on Build GRCh38
rs41289549 Chr.13: 45339827-45339827 on Build GRCh38
rs759775022 Chr.13: 45339452-45339452 on Build GRCh38
rs761984720 Chr.13: 45339422-45339422 on Build GRCh38
rs937252383 Chr.13: 45339836-45339836 on Build GRCh38
rs1029792691 Chr.13: 45339777-45339777 on Build GRCh38
rs2234222 Chr.13: 45339402-45339402 on Build GRCh38
rs553357605 Chr.13: 45339418-45339418 on Build GRCh38
rs377491124 Chr.13: 45339405-45339405 on Build GRCh38
rs889390377 Chr.13: 45339769-45339769 on Build GRCh38
rs982900387 Chr.13: 45339794-45339794 on Build GRCh38
rs534506552 Chr.13: 45339734-45339734 on Build GRCh38
rs564699598 Chr.13: 45339408-45339408 on Build GRCh38
rs762304747 Chr.13: 45339528-45339528 on Build GRCh38
rs1058093 Chr.13: 45339558-45339558 on Build GRCh38
rs765209605 Chr.13: 45339483-45339483 on Build GRCh38
rs750696960 Chr.13: 45339549-45339549 on Build GRCh38
rs375302427 Chr.13: 45339635-45339635 on Build GRCh38
rs529694182 Chr.13: 45339487-45339487 on Build GRCh38
rs746021858 Chr.13: 45339599-45339599 on Build GRCh38
rs879039847 Chr.13: 45339496-45339496 on Build GRCh38
rs770505611 Chr.13: 45339640-45339640 on Build GRCh38
rs766785362 Chr.13: 45339838-45339838 on Build GRCh38
rs897533821 Chr.13: 45339742-45339742 on Build GRCh38
rs1021499114 Chr.13: 45339826-45339826 on Build GRCh38
rs775623590 Chr.13: 45339615-45339615 on Build GRCh38
rs753349796 Chr.13: 45339545-45339545 on Build GRCh38
rs895617279 Chr.13: 45339843-45339843 on Build GRCh38
rs958501864 Chr.13: 45339841-45339841 on Build GRCh38
rs753358784 Chr.13: 45339652-45339652 on Build GRCh38
rs1019915533 Chr.13: 45339713-45339713 on Build GRCh38
rs928578518 Chr.13: 45339629-45339629 on Build GRCh38
rs1036085941 Chr.13: 45339389-45339389 on Build GRCh38
rs11552479 Chr.13: 45339520-45339520 on Build GRCh38
rs535860505 Chr.13: 45339811-45339811 on Build GRCh38
rs777557955 Chr.13: 45339597-45339597 on Build GRCh38
rs762989372 Chr.13: 45339625-45339625 on Build GRCh38
rs778757353 Chr.13: 45339551-45339551 on Build GRCh38
rs191262950 Chr.13: 45339723-45339723 on Build GRCh38
rs941646289 Chr.13: 45339737-45339737 on Build GRCh38
rs80348587 Chr.13: 45339828-45339828 on Build GRCh38
rs771804936 Chr.13: 45339593-45339593 on Build GRCh38
rs879105692 Chr.13: 45339494-45339494 on Build GRCh38

View Full Product Details