Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35870990-35871493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRMS1L
Entrez Gene ID
84312
Gene Name
breast cancer metastasis-suppressor 1-like
Gene Aliases
BRMS1
UniGene
Hs.525299
Gene Chromosome Location
Chr.14: 35825596-35871963 on Build GRCh38
Transcripts
NM_032352.3 XM_005268128.1 XM_006720274.2 XM_006720275.2 XM_017021703.1 XM_017021704.1 XM_017021705.1

SNP Detail

SNP ID Chromosome Location
rs1030142550 Chr.14: 35871366-35871366 on Build GRCh38
rs1027669274 Chr.14: 35871379-35871379 on Build GRCh38
rs963117564 Chr.14: 35871397-35871397 on Build GRCh38
rs963445703 Chr.14: 35871195-35871195 on Build GRCh38
rs950450098 Chr.14: 35871412-35871412 on Build GRCh38
rs1022663298 Chr.14: 35871297-35871297 on Build GRCh38
rs1017670680 Chr.14: 35871386-35871386 on Build GRCh38
rs1030173544 Chr.14: 35871072-35871072 on Build GRCh38
rs1035423370 Chr.14: 35871230-35871230 on Build GRCh38
rs982924824 Chr.14: 35871426-35871426 on Build GRCh38
rs1056169634 Chr.14: 35871272-35871272 on Build GRCh38
rs770551650 Chr.14: 35871084-35871084 on Build GRCh38
rs567242730 Chr.14: 35871199-35871199 on Build GRCh38
rs1047113929 Chr.14: 35871062-35871062 on Build GRCh38
rs944030477 Chr.14: 35871126-35871126 on Build GRCh38
rs1043366163 Chr.14: 35871198-35871198 on Build GRCh38
rs954654784 Chr.14: 35871107-35871107 on Build GRCh38
rs982943571 Chr.14: 35871394-35871394 on Build GRCh38
rs960554152 Chr.14: 35871464-35871464 on Build GRCh38
rs998700237 Chr.14: 35871216-35871216 on Build GRCh38
rs894503859 Chr.14: 35871277-35871278 on Build GRCh38
rs998722423 Chr.14: 35871066-35871066 on Build GRCh38
rs944168056 Chr.14: 35871430-35871430 on Build GRCh38
rs975814279 Chr.14: 35871457-35871457 on Build GRCh38
rs951521422 Chr.14: 35871389-35871389 on Build GRCh38
rs890341093 Chr.14: 35871371-35871371 on Build GRCh38
rs187296989 Chr.14: 35871383-35871383 on Build GRCh38
rs116904698 Chr.14: 35871367-35871367 on Build GRCh38
rs77834824 Chr.14: 35871018-35871018 on Build GRCh38
rs899747626 Chr.14: 35871150-35871150 on Build GRCh38
rs531313182 Chr.14: 35871281-35871281 on Build GRCh38
rs912630050 Chr.14: 35871401-35871401 on Build GRCh38
rs974313545 Chr.14: 35871369-35871369 on Build GRCh38
rs947455759 Chr.14: 35871279-35871279 on Build GRCh38
rs969816166 Chr.14: 35871205-35871205 on Build GRCh38
rs982955831 Chr.14: 35871229-35871229 on Build GRCh38
rs147643927 Chr.14: 35871117-35871117 on Build GRCh38
rs963383089 Chr.14: 35871368-35871368 on Build GRCh38
rs1059562 Chr.14: 35871411-35871411 on Build GRCh38
rs879837355 Chr.14: 35871022-35871022 on Build GRCh38
rs1039733072 Chr.14: 35871135-35871135 on Build GRCh38
rs959799585 Chr.14: 35871244-35871244 on Build GRCh38
rs903286015 Chr.14: 35871320-35871320 on Build GRCh38
rs527492292 Chr.14: 35871083-35871083 on Build GRCh38
rs542652631 Chr.14: 35871181-35871181 on Build GRCh38
rs1059561 Chr.14: 35871180-35871180 on Build GRCh38
rs998608098 Chr.14: 35871356-35871356 on Build GRCh38
rs748847336 Chr.14: 35871031-35871031 on Build GRCh38
rs562928394 Chr.14: 35871258-35871258 on Build GRCh38
rs930161569 Chr.14: 35871217-35871217 on Build GRCh38

View Full Product Details