Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52018954-52019463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NID2
Entrez Gene ID
22795
Gene Name
nidogen 2
Gene Aliases
NID-2
UniGene
Hs.369840
Gene Chromosome Location
Chr.14: 52004802-52069231 on Build GRCh38
Transcripts
NM_007361.3 XM_005267405.4 XM_005267406.4 XM_005267407.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
433122 c.2950G>A p.V984M Chr.14: 52019139-52019139 on Build GRCh38
332647 c.2999C>A p.S1000Y Chr.14: 52019090-52019090 on Build GRCh38
3496327 c.2874C>T p.F958F Chr.14: 52019215-52019215 on Build GRCh38
6344386 c.3004C>A p.P1002T Chr.14: 52019085-52019085 on Build GRCh38
956179 c.2968G>T p.E990* Chr.14: 52019121-52019121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042826974 Chr.14: 52019007-52019007 on Build GRCh38
rs772923728 Chr.14: 52019139-52019139 on Build GRCh38
rs774075467 Chr.14: 52019255-52019255 on Build GRCh38
rs59811471 Chr.14: 52018979-52018979 on Build GRCh38
rs755224484 Chr.14: 52019059-52019059 on Build GRCh38
rs17124896 Chr.14: 52019405-52019405 on Build GRCh38
rs775228294 Chr.14: 52019122-52019122 on Build GRCh38
rs745828412 Chr.14: 52019362-52019362 on Build GRCh38
rs371402626 Chr.14: 52019014-52019014 on Build GRCh38
rs746616149 Chr.14: 52019123-52019123 on Build GRCh38
rs372306558 Chr.14: 52019161-52019161 on Build GRCh38
rs369045026 Chr.14: 52019084-52019084 on Build GRCh38
rs751971781 Chr.14: 52019278-52019278 on Build GRCh38
rs761771353 Chr.14: 52019017-52019017 on Build GRCh38
rs778783252 Chr.14: 52019333-52019333 on Build GRCh38
rs759302816 Chr.14: 52019323-52019323 on Build GRCh38
rs768413460 Chr.14: 52019203-52019203 on Build GRCh38
rs1045505270 Chr.14: 52019430-52019430 on Build GRCh38
rs758770416 Chr.14: 52019274-52019274 on Build GRCh38
rs770125015 Chr.14: 52019077-52019078 on Build GRCh38
rs528753623 Chr.14: 52019146-52019146 on Build GRCh38
rs532046798 Chr.14: 52019287-52019287 on Build GRCh38
rs763513584 Chr.14: 52019276-52019276 on Build GRCh38
rs749261995 Chr.14: 52019298-52019298 on Build GRCh38
rs373278535 Chr.14: 52019257-52019257 on Build GRCh38
rs770850270 Chr.14: 52019304-52019304 on Build GRCh38
rs758819165 Chr.14: 52019191-52019191 on Build GRCh38
rs765220272 Chr.14: 52019137-52019137 on Build GRCh38
rs765187493 Chr.14: 52019268-52019268 on Build GRCh38
rs142113804 Chr.14: 52019281-52019281 on Build GRCh38
rs148838155 Chr.14: 52019332-52019332 on Build GRCh38
rs757747671 Chr.14: 52019097-52019097 on Build GRCh38
rs750451791 Chr.14: 52019345-52019345 on Build GRCh38
rs369346124 Chr.14: 52019228-52019228 on Build GRCh38
rs749800161 Chr.14: 52019085-52019085 on Build GRCh38
rs978859049 Chr.14: 52019306-52019306 on Build GRCh38
rs763588365 Chr.14: 52019346-52019346 on Build GRCh38
rs781446920 Chr.14: 52019066-52019066 on Build GRCh38
rs764959685 Chr.14: 52019036-52019036 on Build GRCh38
rs761871061 Chr.14: 52019339-52019339 on Build GRCh38
rs184330202 Chr.14: 52019254-52019254 on Build GRCh38
rs771668505 Chr.14: 52019113-52019113 on Build GRCh38
rs760954125 Chr.14: 52019256-52019256 on Build GRCh38
rs776227850 Chr.14: 52019134-52019134 on Build GRCh38
rs374690784 Chr.14: 52019280-52019280 on Build GRCh38
rs910878494 Chr.14: 52019316-52019316 on Build GRCh38
rs765369534 Chr.14: 52019342-52019342 on Build GRCh38
rs752994827 Chr.14: 52019323-52019327 on Build GRCh38
rs1131484 Chr.14: 52019083-52019083 on Build GRCh38
rs887022948 Chr.14: 52019439-52019439 on Build GRCh38
rs762789547 Chr.14: 52019140-52019140 on Build GRCh38
rs746211590 Chr.14: 52019104-52019104 on Build GRCh38
rs201188347 Chr.14: 52019159-52019159 on Build GRCh38
rs761904056 Chr.14: 52019135-52019135 on Build GRCh38
rs189227708 Chr.14: 52019406-52019406 on Build GRCh38
rs750940299 Chr.14: 52019269-52019269 on Build GRCh38
rs998998042 Chr.14: 52019063-52019063 on Build GRCh38
rs764216800 Chr.14: 52019155-52019155 on Build GRCh38
rs1046927407 Chr.14: 52019158-52019158 on Build GRCh38
rs766041106 Chr.14: 52019145-52019145 on Build GRCh38
rs375701173 Chr.14: 52019329-52019329 on Build GRCh38
rs777080977 Chr.14: 52019297-52019297 on Build GRCh38
rs559980053 Chr.14: 52019373-52019373 on Build GRCh38
rs903456325 Chr.14: 52019038-52019038 on Build GRCh38
rs538887638 Chr.14: 52018986-52018986 on Build GRCh38
rs762761394 Chr.14: 52019235-52019235 on Build GRCh38
rs922445041 Chr.14: 52019372-52019372 on Build GRCh38
rs752825593 Chr.14: 52019077-52019077 on Build GRCh38
rs377171157 Chr.14: 52019162-52019162 on Build GRCh38
rs910939779 Chr.14: 52019018-52019018 on Build GRCh38
rs747708399 Chr.14: 52019283-52019283 on Build GRCh38
rs189773237 Chr.14: 52018978-52018978 on Build GRCh38
rs746308960 Chr.14: 52019163-52019163 on Build GRCh38
rs377588266 Chr.14: 52019196-52019196 on Build GRCh38
rs139021732 Chr.14: 52019220-52019220 on Build GRCh38
rs146139281 Chr.14: 52019219-52019219 on Build GRCh38
rs750222305 Chr.14: 52019053-52019053 on Build GRCh38
rs756262289 Chr.14: 52019149-52019149 on Build GRCh38
rs780626824 Chr.14: 52019277-52019277 on Build GRCh38
rs566678238 Chr.14: 52019388-52019388 on Build GRCh38
rs768282300 Chr.14: 52019131-52019131 on Build GRCh38
rs746566269 Chr.14: 52019437-52019437 on Build GRCh38
rs1046983476 Chr.14: 52019398-52019398 on Build GRCh38
rs774171455 Chr.14: 52019314-52019314 on Build GRCh38
rs1035224600 Chr.14: 52019227-52019227 on Build GRCh38
rs559912470 Chr.14: 52019103-52019103 on Build GRCh38
rs753966543 Chr.14: 52019262-52019262 on Build GRCh38
rs142814187 Chr.14: 52019197-52019197 on Build GRCh38
rs757378542 Chr.14: 52019263-52019263 on Build GRCh38
rs530969304 Chr.14: 52019231-52019231 on Build GRCh38
rs1040887191 Chr.14: 52019087-52019087 on Build GRCh38

View Full Product Details