Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64504765-64505256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB1
Entrez Gene ID
22890
Gene Name
zinc finger and BTB domain containing 1
Gene Aliases
ZNF909
UniGene
Hs.605143
Gene Chromosome Location
Chr.14: 64504574-64533690 on Build GRCh38
Transcripts
NM_001123329.1 NM_014950.2 XM_005267410.1 XM_011536566.2 XM_011536568.2 XM_017021095.1 NM_001304507.1 NM_001304508.1

Gene Symbol
ZBTB25
Entrez Gene ID
7597
Gene Name
zinc finger and BTB domain containing 25
Gene Aliases
C14orf51, KUP, ZNF46
UniGene
Hs.654571
Gene Chromosome Location
Chr.14: 64447172-64505213 on Build GRCh38
Transcripts
NM_001123329.1 NM_014950.2 XM_005267410.1 XM_011536566.2 XM_011536568.2 XM_017021095.1 NM_001304507.1 NM_001304508.1

SNP Detail

SNP ID Chromosome Location
rs1001248968 Chr.14: 64505212-64505215 on Build GRCh38
rs867941104 Chr.14: 64504946-64504946 on Build GRCh38
rs928374918 Chr.14: 64505003-64505003 on Build GRCh38
rs903061496 Chr.14: 64505177-64505177 on Build GRCh38
rs1025860939 Chr.14: 64505037-64505037 on Build GRCh38
rs957272175 Chr.14: 64504829-64504829 on Build GRCh38
rs929999699 Chr.14: 64505175-64505175 on Build GRCh38
rs1029323889 Chr.14: 64504828-64504828 on Build GRCh38
rs1006924254 Chr.14: 64504865-64504865 on Build GRCh38
rs1040427375 Chr.14: 64504942-64504942 on Build GRCh38
rs952876438 Chr.14: 64504898-64504898 on Build GRCh38
rs546397414 Chr.14: 64504960-64504960 on Build GRCh38
rs181685058 Chr.14: 64505000-64505000 on Build GRCh38
rs988693019 Chr.14: 64505097-64505097 on Build GRCh38
rs1043114044 Chr.14: 64504844-64504845 on Build GRCh38
rs913042776 Chr.14: 64504832-64504832 on Build GRCh38
rs991573552 Chr.14: 64505020-64505020 on Build GRCh38
rs761546 Chr.14: 64505025-64505025 on Build GRCh38
rs1035498080 Chr.14: 64505180-64505180 on Build GRCh38
rs1035334691 Chr.14: 64505084-64505084 on Build GRCh38
rs200099822 Chr.14: 64505204-64505205 on Build GRCh38
rs991234596 Chr.14: 64505200-64505200 on Build GRCh38
rs538016482 Chr.14: 64505073-64505073 on Build GRCh38
rs958446795 Chr.14: 64505013-64505013 on Build GRCh38
rs781408981 Chr.14: 64505159-64505159 on Build GRCh38
rs1018924425 Chr.14: 64504819-64504819 on Build GRCh38
rs1041115713 Chr.14: 64505157-64505157 on Build GRCh38
rs988661867 Chr.14: 64504988-64504988 on Build GRCh38
rs893121352 Chr.14: 64505113-64505113 on Build GRCh38
rs922600870 Chr.14: 64505164-64505164 on Build GRCh38
rs901446268 Chr.14: 64504881-64504881 on Build GRCh38
rs936998907 Chr.14: 64504886-64504886 on Build GRCh38
rs1052907668 Chr.14: 64504825-64504825 on Build GRCh38
rs1011530265 Chr.14: 64505127-64505127 on Build GRCh38
rs1024211624 Chr.14: 64505072-64505072 on Build GRCh38
rs908083298 Chr.14: 64505050-64505050 on Build GRCh38
rs957532409 Chr.14: 64504980-64504980 on Build GRCh38
rs996143572 Chr.14: 64504827-64504827 on Build GRCh38
rs938619200 Chr.14: 64504983-64504983 on Build GRCh38
rs886590898 Chr.14: 64504940-64504940 on Build GRCh38
rs1019612159 Chr.14: 64504875-64504875 on Build GRCh38
rs1005753630 Chr.14: 64505056-64505056 on Build GRCh38
rs777081131 Chr.14: 64504935-64504935 on Build GRCh38
rs886626911 Chr.14: 64504861-64504861 on Build GRCh38
rs1004950676 Chr.14: 64504941-64504941 on Build GRCh38
rs938588076 Chr.14: 64504818-64504819 on Build GRCh38
rs959667547 Chr.14: 64505089-64505089 on Build GRCh38
rs930879293 Chr.14: 64504923-64504923 on Build GRCh38
rs967140586 Chr.14: 64504835-64504835 on Build GRCh38
rs142434580 Chr.14: 64504887-64504887 on Build GRCh38
rs886538628 Chr.14: 64504800-64504800 on Build GRCh38
rs901932906 Chr.14: 64505092-64505092 on Build GRCh38
rs911020176 Chr.14: 64505150-64505150 on Build GRCh38
rs940861119 Chr.14: 64505052-64505052 on Build GRCh38
rs1002713608 Chr.14: 64505178-64505178 on Build GRCh38
rs942773518 Chr.14: 64505156-64505156 on Build GRCh38
rs994428513 Chr.14: 64505034-64505034 on Build GRCh38
rs989990438 Chr.14: 64504830-64504830 on Build GRCh38
rs1049287519 Chr.14: 64505036-64505036 on Build GRCh38
rs905330952 Chr.14: 64505210-64505210 on Build GRCh38
rs967192977 Chr.14: 64504995-64504995 on Build GRCh38
rs1025829870 Chr.14: 64504890-64504890 on Build GRCh38
rs932056699 Chr.14: 64505107-64505107 on Build GRCh38
rs1020206898 Chr.14: 64505161-64505161 on Build GRCh38
rs981172455 Chr.14: 64504846-64504846 on Build GRCh38
rs755329440 Chr.14: 64504964-64504966 on Build GRCh38
rs958716522 Chr.14: 64505181-64505181 on Build GRCh38
rs773211196 Chr.14: 64505173-64505173 on Build GRCh38
rs1044452980 Chr.14: 64504804-64504804 on Build GRCh38
rs1011479351 Chr.14: 64504993-64504993 on Build GRCh38
rs533007749 Chr.14: 64504959-64504959 on Build GRCh38
rs979798794 Chr.14: 64505076-64505076 on Build GRCh38
rs111475126 Chr.14: 64505087-64505087 on Build GRCh38
rs750894006 Chr.14: 64504903-64504904 on Build GRCh38
rs1028960315 Chr.14: 64504978-64504978 on Build GRCh38
rs916823118 Chr.14: 64504901-64504901 on Build GRCh38
rs1006200539 Chr.14: 64505199-64505199 on Build GRCh38
rs928316148 Chr.14: 64504877-64504877 on Build GRCh38
rs889789598 Chr.14: 64504848-64504848 on Build GRCh38
rs966427630 Chr.14: 64504823-64504823 on Build GRCh38
rs1027095448 Chr.14: 64505219-64505219 on Build GRCh38
rs1004899938 Chr.14: 64504817-64504817 on Build GRCh38
rs888692120 Chr.14: 64505043-64505043 on Build GRCh38
rs760759740 Chr.14: 64505218-64505218 on Build GRCh38
rs550792234 Chr.14: 64504785-64504785 on Build GRCh38
rs9323451 Chr.14: 64505031-64505031 on Build GRCh38
rs566414287 Chr.14: 64504994-64504994 on Build GRCh38
rs1050877631 Chr.14: 64505111-64505111 on Build GRCh38

View Full Product Details