Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64942815-64943313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHURC1-FNTB
Entrez Gene ID
100529261
Gene Name
CHURC1-FNTB readthrough
Gene Aliases
-
UniGene
Hs.325531
Gene Chromosome Location
Chr.14: 64914361-65062655 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002083.3 NR_046320.1 NR_046321.1

Gene Symbol
GPX2
Entrez Gene ID
2877
Gene Name
glutathione peroxidase 2
Gene Aliases
GI-GPx, GPRP, GPRP-2, GPx-2, GPx-GI, GSHPX-GI, GSHPx-2
UniGene
Hs.2704
Gene Chromosome Location
Chr.14: 64939152-64942905 on Build GRCh38
Transcripts
NM_001202558.1 NM_001202559.1 NM_002083.3 NR_046320.1 NR_046321.1

SNP Detail

SNP ID Chromosome Location
rs905041550 Chr.14: 64943261-64943261 on Build GRCh38
rs996554536 Chr.14: 64942993-64942993 on Build GRCh38
rs898272573 Chr.14: 64943272-64943272 on Build GRCh38
rs1032421286 Chr.14: 64943100-64943100 on Build GRCh38
rs918213050 Chr.14: 64942936-64942936 on Build GRCh38
rs1055294487 Chr.14: 64942927-64942927 on Build GRCh38
rs988234657 Chr.14: 64943056-64943056 on Build GRCh38
rs955346042 Chr.14: 64943044-64943044 on Build GRCh38
rs574886503 Chr.14: 64943229-64943229 on Build GRCh38
rs36206674 Chr.14: 64943119-64943119 on Build GRCh38
rs898156456 Chr.14: 64942960-64942960 on Build GRCh38
rs17886872 Chr.14: 64942941-64942941 on Build GRCh38
rs767235901 Chr.14: 64942849-64942849 on Build GRCh38
rs562859729 Chr.14: 64943254-64943254 on Build GRCh38
rs568421670 Chr.14: 64942913-64942913 on Build GRCh38
rs149233931 Chr.14: 64943064-64943064 on Build GRCh38
rs17093568 Chr.14: 64943271-64943271 on Build GRCh38
rs984002331 Chr.14: 64943237-64943237 on Build GRCh38
rs1028640064 Chr.14: 64943022-64943022 on Build GRCh38
rs887751964 Chr.14: 64943208-64943208 on Build GRCh38
rs966834862 Chr.14: 64942840-64942840 on Build GRCh38
rs993875939 Chr.14: 64942912-64942912 on Build GRCh38
rs1009110972 Chr.14: 64942880-64942880 on Build GRCh38
rs11628603 Chr.14: 64942968-64942968 on Build GRCh38
rs1023033734 Chr.14: 64943258-64943258 on Build GRCh38
rs1027637575 Chr.14: 64942921-64942921 on Build GRCh38
rs886829446 Chr.14: 64942949-64942949 on Build GRCh38
rs1040563129 Chr.14: 64942881-64942881 on Build GRCh38
rs17881154 Chr.14: 64943136-64943136 on Build GRCh38
rs946491132 Chr.14: 64943065-64943065 on Build GRCh38
rs979496058 Chr.14: 64943097-64943097 on Build GRCh38
rs17880939 Chr.14: 64943283-64943283 on Build GRCh38
rs925291017 Chr.14: 64942854-64942854 on Build GRCh38
rs548603440 Chr.14: 64942872-64942872 on Build GRCh38
rs761303730 Chr.14: 64943061-64943064 on Build GRCh38
rs571054892 Chr.14: 64943041-64943041 on Build GRCh38
rs939716450 Chr.14: 64943255-64943255 on Build GRCh38
rs1049563638 Chr.14: 64943205-64943205 on Build GRCh38
rs766764553 Chr.14: 64943045-64943045 on Build GRCh38
rs1003982236 Chr.14: 64942953-64942953 on Build GRCh38
rs572628151 Chr.14: 64943153-64943153 on Build GRCh38
rs755526733 Chr.14: 64943073-64943073 on Build GRCh38
rs897844532 Chr.14: 64942901-64942901 on Build GRCh38
rs17880421 Chr.14: 64943187-64943187 on Build GRCh38
rs533218349 Chr.14: 64943280-64943280 on Build GRCh38
rs528567581 Chr.14: 64942867-64942867 on Build GRCh38
rs2737843 Chr.14: 64942945-64942945 on Build GRCh38
rs370322949 Chr.14: 64942974-64942974 on Build GRCh38
rs36206353 Chr.14: 64942958-64942958 on Build GRCh38
rs750761678 Chr.14: 64942866-64942866 on Build GRCh38
rs944852126 Chr.14: 64943147-64943147 on Build GRCh38
rs36206677 Chr.14: 64943201-64943201 on Build GRCh38
rs17883620 Chr.14: 64942925-64942925 on Build GRCh38
rs754280040 Chr.14: 64943056-64943056 on Build GRCh38

View Full Product Details