Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94101685-94102178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI27L1
Entrez Gene ID
122509
Gene Name
interferon alpha inducible protein 27 like 1
Gene Aliases
FAM14B, ISG12C
UniGene
Hs.19414
Gene Chromosome Location
Chr.14: 94081293-94102714 on Build GRCh38
Transcripts
NM_145249.2 NM_206949.2 XM_005267314.4 XM_011536405.2 XM_011536406.1 XM_011536407.1 XM_017020961.1 XM_017020962.1 XM_017020963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5906556 c.62-7C>T p.? Chr.14: 94101807-94101807 on Build GRCh38
4578078 c.96T>C p.S32S Chr.14: 94101848-94101848 on Build GRCh38
7296937 c.162A>C p.A54A Chr.14: 94101914-94101914 on Build GRCh38
7258099 c.130T>C p.S44P Chr.14: 94101882-94101882 on Build GRCh38
4053204 c.180C>T p.G60G Chr.14: 94101932-94101932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185915352 Chr.14: 94101816-94101816 on Build GRCh38
rs765508462 Chr.14: 94101923-94101923 on Build GRCh38
rs1008194677 Chr.14: 94101848-94101848 on Build GRCh38
rs971708937 Chr.14: 94102083-94102083 on Build GRCh38
rs755247584 Chr.14: 94101828-94101828 on Build GRCh38
rs545646095 Chr.14: 94101758-94101758 on Build GRCh38
rs937742428 Chr.14: 94101901-94101901 on Build GRCh38
rs57677258 Chr.14: 94101855-94101855 on Build GRCh38
rs369456554 Chr.14: 94101791-94101791 on Build GRCh38
rs749033754 Chr.14: 94102059-94102059 on Build GRCh38
rs748261895 Chr.14: 94101834-94101834 on Build GRCh38
rs147197904 Chr.14: 94101849-94101849 on Build GRCh38
rs148533447 Chr.14: 94101879-94101879 on Build GRCh38
rs765982082 Chr.14: 94101857-94101858 on Build GRCh38
rs375179484 Chr.14: 94101786-94101786 on Build GRCh38
rs371073384 Chr.14: 94101959-94101959 on Build GRCh38
rs564140104 Chr.14: 94101775-94101775 on Build GRCh38
rs958574000 Chr.14: 94101995-94101995 on Build GRCh38
rs150961547 Chr.14: 94102016-94102016 on Build GRCh38
rs139698926 Chr.14: 94101720-94101720 on Build GRCh38
rs201602028 Chr.14: 94102023-94102023 on Build GRCh38
rs762161398 Chr.14: 94101808-94101808 on Build GRCh38
rs761443811 Chr.14: 94101799-94101799 on Build GRCh38
rs897569412 Chr.14: 94101821-94101821 on Build GRCh38
rs1019047031 Chr.14: 94102145-94102145 on Build GRCh38
rs138791669 Chr.14: 94101853-94101853 on Build GRCh38
rs774117771 Chr.14: 94101952-94101952 on Build GRCh38
rs566001096 Chr.14: 94101875-94101875 on Build GRCh38
rs778405904 Chr.14: 94102064-94102064 on Build GRCh38
rs1007641467 Chr.14: 94102109-94102109 on Build GRCh38
rs780026517 Chr.14: 94102055-94102055 on Build GRCh38
rs751643663 Chr.14: 94102034-94102034 on Build GRCh38
rs769435068 Chr.14: 94101776-94101776 on Build GRCh38
rs764122953 Chr.14: 94102033-94102033 on Build GRCh38
rs1048814666 Chr.14: 94102098-94102098 on Build GRCh38
rs751082734 Chr.14: 94102038-94102039 on Build GRCh38
rs747578211 Chr.14: 94101957-94101957 on Build GRCh38
rs545021441 Chr.14: 94101876-94101876 on Build GRCh38
rs200925095 Chr.14: 94101930-94101930 on Build GRCh38
rs762500563 Chr.14: 94101791-94101791 on Build GRCh38
rs758574455 Chr.14: 94102027-94102027 on Build GRCh38
rs138047821 Chr.14: 94101821-94101822 on Build GRCh38
rs749123532 Chr.14: 94101949-94101949 on Build GRCh38
rs889306828 Chr.14: 94101845-94101845 on Build GRCh38
rs758548682 Chr.14: 94101927-94101927 on Build GRCh38
rs370886948 Chr.14: 94101837-94101837 on Build GRCh38
rs569783163 Chr.14: 94101933-94101933 on Build GRCh38
rs1026569515 Chr.14: 94101870-94101870 on Build GRCh38
rs930407133 Chr.14: 94101826-94101826 on Build GRCh38
rs755085298 Chr.14: 94101914-94101914 on Build GRCh38
rs145128231 Chr.14: 94101823-94101823 on Build GRCh38
rs537159389 Chr.14: 94101970-94101970 on Build GRCh38
rs754281769 Chr.14: 94101910-94101910 on Build GRCh38
rs201917237 Chr.14: 94101810-94101810 on Build GRCh38
rs369883184 Chr.14: 94101842-94101842 on Build GRCh38
rs775289852 Chr.14: 94101847-94101847 on Build GRCh38
rs755988331 Chr.14: 94102036-94102036 on Build GRCh38
rs759880216 Chr.14: 94102086-94102086 on Build GRCh38
rs756612002 Chr.14: 94101814-94101814 on Build GRCh38
rs761219558 Chr.14: 94101968-94101968 on Build GRCh38
rs754840090 Chr.14: 94102060-94102060 on Build GRCh38
rs903783708 Chr.14: 94101976-94101976 on Build GRCh38
rs538099802 Chr.14: 94101789-94101789 on Build GRCh38
rs925832664 Chr.14: 94101740-94101740 on Build GRCh38
rs142548662 Chr.14: 94101809-94101809 on Build GRCh38
rs968139785 Chr.14: 94101725-94101725 on Build GRCh38
rs751112213 Chr.14: 94101805-94101805 on Build GRCh38
rs541456241 Chr.14: 94102024-94102024 on Build GRCh38
rs772869850 Chr.14: 94101965-94101965 on Build GRCh38
rs879223884 Chr.14: 94101982-94101982 on Build GRCh38
rs745759290 Chr.14: 94101931-94101931 on Build GRCh38
rs930363758 Chr.14: 94102094-94102094 on Build GRCh38
rs752718253 Chr.14: 94101926-94101926 on Build GRCh38
rs150611249 Chr.14: 94101932-94101932 on Build GRCh38
rs910674957 Chr.14: 94102103-94102103 on Build GRCh38
rs778814517 Chr.14: 94101830-94101830 on Build GRCh38
rs770713618 Chr.14: 94101836-94101836 on Build GRCh38
rs528926458 Chr.14: 94101841-94101841 on Build GRCh38
rs937233219 Chr.14: 94101743-94101743 on Build GRCh38
rs12881245 Chr.14: 94101737-94101737 on Build GRCh38
rs943549647 Chr.14: 94102126-94102126 on Build GRCh38
rs140017775 Chr.14: 94101953-94101953 on Build GRCh38
rs117094493 Chr.14: 94101878-94101878 on Build GRCh38
rs553367692 Chr.14: 94102087-94102087 on Build GRCh38

View Full Product Details