Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 47760839-47761345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA6D
Entrez Gene ID
80031
Gene Name
semaphorin 6D
Gene Aliases
-
UniGene
Hs.511265
Gene Chromosome Location
Chr.15: 47184101-47774223 on Build GRCh38
Transcripts
NM_001198999.1 NM_020858.1 NM_024966.2 NM_153616.1 NM_153617.1 NM_153618.1 NM_153619.1 XM_005254685.3 XM_005254686.2 XM_005254687.2 XM_005254689.3 XM_011522075.2 XM_011522076.2 XM_011522077.2 XM_011522078.2 XM_011522079.2 XM_011522080.2 XM_011522081.2 XM_017022617.1 XM_017022618.1 XM_017022619.1 XM_017022620.1 XM_017022621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046098 c.359A>T p.N120I Chr.15: 47761343-47761343 on Build GRCh38
6142427 c.336C>A p.G112G Chr.15: 47761211-47761211 on Build GRCh38
6217018 c.285A>G p.K95K Chr.15: 47761160-47761160 on Build GRCh38
4410013 c.282+1G>A p.? Chr.15: 47761039-47761039 on Build GRCh38
6032830 c.222-1G>T p.? Chr.15: 47760977-47760977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746895205 Chr.15: 47761083-47761083 on Build GRCh38
rs534494384 Chr.15: 47760908-47760908 on Build GRCh38
rs371683860 Chr.15: 47761295-47761295 on Build GRCh38
rs758178406 Chr.15: 47761076-47761076 on Build GRCh38
rs599111 Chr.15: 47761137-47761137 on Build GRCh38
rs540805949 Chr.15: 47761105-47761105 on Build GRCh38
rs142054743 Chr.15: 47761254-47761254 on Build GRCh38
rs757331801 Chr.15: 47761249-47761249 on Build GRCh38
rs562385125 Chr.15: 47761051-47761051 on Build GRCh38
rs774631553 Chr.15: 47761109-47761109 on Build GRCh38
rs777244903 Chr.15: 47761302-47761302 on Build GRCh38
rs773332083 Chr.15: 47761279-47761280 on Build GRCh38
rs990180902 Chr.15: 47761099-47761099 on Build GRCh38
rs769840309 Chr.15: 47760991-47760991 on Build GRCh38
rs777467700 Chr.15: 47760931-47760931 on Build GRCh38
rs374736689 Chr.15: 47761031-47761031 on Build GRCh38
rs780035682 Chr.15: 47760971-47760971 on Build GRCh38
rs368693350 Chr.15: 47761070-47761070 on Build GRCh38
rs763151924 Chr.15: 47761320-47761323 on Build GRCh38
rs778504140 Chr.15: 47760947-47760947 on Build GRCh38
rs745688559 Chr.15: 47760964-47760964 on Build GRCh38
rs778910646 Chr.15: 47761251-47761251 on Build GRCh38
rs150296354 Chr.15: 47761019-47761019 on Build GRCh38
rs377217662 Chr.15: 47761119-47761119 on Build GRCh38
rs747364196 Chr.15: 47761288-47761288 on Build GRCh38
rs764927722 Chr.15: 47761054-47761054 on Build GRCh38
rs535377803 Chr.15: 47760978-47760978 on Build GRCh38
rs370119933 Chr.15: 47761166-47761166 on Build GRCh38
rs780507902 Chr.15: 47761262-47761262 on Build GRCh38
rs78293687 Chr.15: 47761171-47761171 on Build GRCh38
rs147097507 Chr.15: 47761255-47761255 on Build GRCh38
rs749468791 Chr.15: 47761004-47761004 on Build GRCh38
rs960829788 Chr.15: 47761207-47761207 on Build GRCh38
rs763650475 Chr.15: 47761049-47761049 on Build GRCh38
rs147211657 Chr.15: 47761177-47761177 on Build GRCh38
rs781098399 Chr.15: 47761189-47761189 on Build GRCh38
rs1041338687 Chr.15: 47760895-47760895 on Build GRCh38
rs773298756 Chr.15: 47761003-47761003 on Build GRCh38
rs762273367 Chr.15: 47761305-47761305 on Build GRCh38
rs566893145 Chr.15: 47760879-47760879 on Build GRCh38
rs114660817 Chr.15: 47761278-47761278 on Build GRCh38
rs575200196 Chr.15: 47761047-47761047 on Build GRCh38
rs373149085 Chr.15: 47760937-47760937 on Build GRCh38
rs993790174 Chr.15: 47761113-47761113 on Build GRCh38
rs945996459 Chr.15: 47760893-47760893 on Build GRCh38
rs567986890 Chr.15: 47760925-47760925 on Build GRCh38
rs755682710 Chr.15: 47760927-47760927 on Build GRCh38
rs772580647 Chr.15: 47761215-47761215 on Build GRCh38
rs756992590 Chr.15: 47760946-47760946 on Build GRCh38
rs376339849 Chr.15: 47761084-47761084 on Build GRCh38
rs772461994 Chr.15: 47761122-47761122 on Build GRCh38
rs752701003 Chr.15: 47761214-47761214 on Build GRCh38
rs373094795 Chr.15: 47761085-47761085 on Build GRCh38
rs758183824 Chr.15: 47760966-47760966 on Build GRCh38
rs777334923 Chr.15: 47761143-47761143 on Build GRCh38
rs754855116 Chr.15: 47761179-47761179 on Build GRCh38
rs373474008 Chr.15: 47761052-47761052 on Build GRCh38
rs761401878 Chr.15: 47761151-47761151 on Build GRCh38
rs761225388 Chr.15: 47761131-47761131 on Build GRCh38
rs771150512 Chr.15: 47761013-47761013 on Build GRCh38
rs779760382 Chr.15: 47761078-47761078 on Build GRCh38
rs977335758 Chr.15: 47761285-47761285 on Build GRCh38
rs539110815 Chr.15: 47761051-47761051 on Build GRCh38
rs501916 Chr.15: 47761032-47761032 on Build GRCh38
rs138062607 Chr.15: 47760922-47760922 on Build GRCh38
rs573006671 Chr.15: 47761088-47761088 on Build GRCh38
rs200866177 Chr.15: 47761170-47761170 on Build GRCh38
rs764692263 Chr.15: 47761163-47761163 on Build GRCh38

View Full Product Details