Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 47762924-47763434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA6D
Entrez Gene ID
80031
Gene Name
semaphorin 6D
Gene Aliases
-
UniGene
Hs.511265
Gene Chromosome Location
Chr.15: 47184101-47774223 on Build GRCh38
Transcripts
NM_001198999.1 NM_020858.1 NM_024966.2 NM_153616.1 NM_153617.1 NM_153618.1 NM_153619.1 XM_005254685.3 XM_005254686.2 XM_005254687.2 XM_005254689.3 XM_011522075.2 XM_011522076.2 XM_011522077.2 XM_011522078.2 XM_011522079.2 XM_011522080.2 XM_011522081.2 XM_017022617.1 XM_017022618.1 XM_017022619.1 XM_017022620.1 XM_017022621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5960765 c.726C>T p.V242V Chr.15: 47763083-47763083 on Build GRCh38
1587893 c.699C>A p.V233V Chr.15: 47763056-47763056 on Build GRCh38
3887070 c.720C>T p.I240I Chr.15: 47763077-47763077 on Build GRCh38
6142425 c.712C>A p.R238R Chr.15: 47763069-47763069 on Build GRCh38
6390798 c.666C>A p.H222Q Chr.15: 47763023-47763023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754501779 Chr.15: 47763154-47763154 on Build GRCh38
rs1041557383 Chr.15: 47763152-47763152 on Build GRCh38
rs797009125 Chr.15: 47763309-47763309 on Build GRCh38
rs186497912 Chr.15: 47763350-47763350 on Build GRCh38
rs763035961 Chr.15: 47763062-47763062 on Build GRCh38
rs886333500 Chr.15: 47763201-47763201 on Build GRCh38
rs549608816 Chr.15: 47763181-47763181 on Build GRCh38
rs923125197 Chr.15: 47763177-47763177 on Build GRCh38
rs1047763095 Chr.15: 47763313-47763313 on Build GRCh38
rs1037357970 Chr.15: 47763228-47763228 on Build GRCh38
rs895283993 Chr.15: 47763360-47763360 on Build GRCh38
rs547616038 Chr.15: 47763291-47763291 on Build GRCh38
rs762162720 Chr.15: 47763113-47763113 on Build GRCh38
rs775674496 Chr.15: 47763070-47763070 on Build GRCh38
rs1013066771 Chr.15: 47763159-47763159 on Build GRCh38
rs750967980 Chr.15: 47762977-47762977 on Build GRCh38
rs780788604 Chr.15: 47763029-47763029 on Build GRCh38
rs879221775 Chr.15: 47763336-47763336 on Build GRCh38
rs374802346 Chr.15: 47762985-47762985 on Build GRCh38
rs779638752 Chr.15: 47763002-47763002 on Build GRCh38
rs974581443 Chr.15: 47763396-47763396 on Build GRCh38
rs28493558 Chr.15: 47763379-47763379 on Build GRCh38
rs141276774 Chr.15: 47763083-47763083 on Build GRCh38
rs943446801 Chr.15: 47762955-47762955 on Build GRCh38
rs978413424 Chr.15: 47763191-47763191 on Build GRCh38
rs1022642382 Chr.15: 47763369-47763369 on Build GRCh38
rs571364176 Chr.15: 47763185-47763185 on Build GRCh38
rs758974199 Chr.15: 47763129-47763129 on Build GRCh38
rs375353104 Chr.15: 47763127-47763127 on Build GRCh38
rs769775616 Chr.15: 47763035-47763035 on Build GRCh38
rs999781181 Chr.15: 47763374-47763374 on Build GRCh38
rs773256402 Chr.15: 47763054-47763054 on Build GRCh38
rs772001338 Chr.15: 47763069-47763069 on Build GRCh38
rs768015372 Chr.15: 47763066-47763066 on Build GRCh38
rs894838317 Chr.15: 47763231-47763231 on Build GRCh38
rs577303054 Chr.15: 47763383-47763383 on Build GRCh38
rs1039120845 Chr.15: 47763285-47763285 on Build GRCh38
rs146298995 Chr.15: 47763032-47763032 on Build GRCh38
rs755516668 Chr.15: 47762954-47762954 on Build GRCh38
rs531391042 Chr.15: 47763168-47763168 on Build GRCh38
rs886481599 Chr.15: 47763328-47763329 on Build GRCh38
rs139642041 Chr.15: 47763361-47763361 on Build GRCh38
rs754120357 Chr.15: 47763100-47763100 on Build GRCh38
rs10519143 Chr.15: 47763370-47763370 on Build GRCh38
rs545196028 Chr.15: 47763384-47763384 on Build GRCh38
rs377544357 Chr.15: 47763005-47763005 on Build GRCh38
rs924499525 Chr.15: 47763200-47763200 on Build GRCh38
rs989717109 Chr.15: 47763364-47763364 on Build GRCh38
rs538834259 Chr.15: 47763190-47763190 on Build GRCh38
rs983265529 Chr.15: 47763406-47763406 on Build GRCh38
rs376819874 Chr.15: 47762991-47762991 on Build GRCh38
rs560732106 Chr.15: 47763136-47763136 on Build GRCh38
rs964430418 Chr.15: 47763378-47763378 on Build GRCh38
rs376460824 Chr.15: 47763146-47763146 on Build GRCh38
rs560404797 Chr.15: 47763385-47763385 on Build GRCh38
rs958294659 Chr.15: 47763004-47763004 on Build GRCh38
rs1025108223 Chr.15: 47763328-47763328 on Build GRCh38
rs1011997824 Chr.15: 47763288-47763288 on Build GRCh38
rs943246847 Chr.15: 47763267-47763267 on Build GRCh38
rs536566706 Chr.15: 47763341-47763341 on Build GRCh38
rs768356382 Chr.15: 47763009-47763009 on Build GRCh38
rs1039171784 Chr.15: 47762978-47762978 on Build GRCh38
rs930712905 Chr.15: 47763409-47763409 on Build GRCh38
rs1006167008 Chr.15: 47763204-47763204 on Build GRCh38
rs371089902 Chr.15: 47763086-47763086 on Build GRCh38
rs920638886 Chr.15: 47763407-47763407 on Build GRCh38
rs1035593344 Chr.15: 47763138-47763138 on Build GRCh38
rs987445207 Chr.15: 47763211-47763211 on Build GRCh38
rs765637196 Chr.15: 47763123-47763123 on Build GRCh38
rs572327358 Chr.15: 47763393-47763393 on Build GRCh38
rs752024378 Chr.15: 47762951-47762951 on Build GRCh38
rs566002016 Chr.15: 47763340-47763340 on Build GRCh38
rs751089825 Chr.15: 47763139-47763139 on Build GRCh38
rs956156405 Chr.15: 47763206-47763206 on Build GRCh38

View Full Product Details