Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48202411-48202955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTXN2
Entrez Gene ID
399697
Gene Name
cortexin 2
Gene Aliases
-
UniGene
Hs.721408
Gene Chromosome Location
Chr.15: 48178564-48203754 on Build GRCh38
Transcripts
NM_001145668.1

SNP Detail

SNP ID Chromosome Location
rs1008419803 Chr.15: 48202705-48202705 on Build GRCh38
rs939197654 Chr.15: 48202525-48202525 on Build GRCh38
rs888187465 Chr.15: 48202691-48202691 on Build GRCh38
rs907694295 Chr.15: 48202515-48202515 on Build GRCh38
rs1024492826 Chr.15: 48202791-48202791 on Build GRCh38
rs903318310 Chr.15: 48202795-48202795 on Build GRCh38
rs763515177 Chr.15: 48202592-48202592 on Build GRCh38
rs1017531448 Chr.15: 48202881-48202881 on Build GRCh38
rs1036200436 Chr.15: 48202850-48202850 on Build GRCh38
rs1043377841 Chr.15: 48202632-48202632 on Build GRCh38
rs986124166 Chr.15: 48202833-48202833 on Build GRCh38
rs574802583 Chr.15: 48202895-48202895 on Build GRCh38
rs991938282 Chr.15: 48202896-48202896 on Build GRCh38
rs535366895 Chr.15: 48202809-48202809 on Build GRCh38
rs983312976 Chr.15: 48202490-48202490 on Build GRCh38
rs1047217622 Chr.15: 48202685-48202685 on Build GRCh38
rs995784309 Chr.15: 48202794-48202794 on Build GRCh38
rs757160687 Chr.15: 48202626-48202626 on Build GRCh38
rs929743793 Chr.15: 48202637-48202637 on Build GRCh38
rs941042956 Chr.15: 48202712-48202712 on Build GRCh38
rs571249821 Chr.15: 48202920-48202920 on Build GRCh38
rs977035456 Chr.15: 48202894-48202894 on Build GRCh38
rs1052620469 Chr.15: 48202666-48202666 on Build GRCh38
rs922404745 Chr.15: 48202625-48202625 on Build GRCh38
rs916509261 Chr.15: 48202589-48202589 on Build GRCh38
rs551216904 Chr.15: 48202783-48202783 on Build GRCh38
rs963949088 Chr.15: 48202891-48202891 on Build GRCh38
rs922468829 Chr.15: 48202897-48202897 on Build GRCh38
rs753264622 Chr.15: 48202801-48202801 on Build GRCh38
rs558679675 Chr.15: 48202746-48202746 on Build GRCh38
rs891256199 Chr.15: 48202679-48202679 on Build GRCh38
rs1033098011 Chr.15: 48202819-48202819 on Build GRCh38
rs753767298 Chr.15: 48202618-48202618 on Build GRCh38
rs935087320 Chr.15: 48202638-48202638 on Build GRCh38
rs765098497 Chr.15: 48202734-48202734 on Build GRCh38
rs947999213 Chr.15: 48202630-48202630 on Build GRCh38
rs553373504 Chr.15: 48202871-48202871 on Build GRCh38
rs1039950792 Chr.15: 48202767-48202767 on Build GRCh38
rs957435788 Chr.15: 48202829-48202829 on Build GRCh38
rs992013395 Chr.15: 48202565-48202565 on Build GRCh38

View Full Product Details