Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52312309-52312825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO5A
Entrez Gene ID
4644
Gene Name
myosin VA
Gene Aliases
GS1, MYH12, MYO5, MYR12
UniGene
Hs.21213
Gene Chromosome Location
Chr.15: 52307283-52529050 on Build GRCh38
Transcripts
NM_000259.3 NM_001142495.1 XM_005254397.3 XM_011521606.2 XM_011521607.2 XM_011521608.2 XM_011521609.2 XM_011521610.2 XM_011521611.2 XM_011521612.2 XM_017022227.1

SNP Detail

SNP ID Chromosome Location
rs142981340 Chr.15: 52312580-52312580 on Build GRCh38
rs1014958249 Chr.15: 52312370-52312370 on Build GRCh38
rs980994886 Chr.15: 52312414-52312414 on Build GRCh38
rs552518770 Chr.15: 52312501-52312501 on Build GRCh38
rs747478222 Chr.15: 52312416-52312416 on Build GRCh38
rs1048264588 Chr.15: 52312540-52312540 on Build GRCh38
rs1032684566 Chr.15: 52312430-52312430 on Build GRCh38
rs999948736 Chr.15: 52312717-52312717 on Build GRCh38
rs969537190 Chr.15: 52312630-52312630 on Build GRCh38
rs962110611 Chr.15: 52312378-52312378 on Build GRCh38
rs773588214 Chr.15: 52312550-52312550 on Build GRCh38
rs932470473 Chr.15: 52312517-52312517 on Build GRCh38
rs574071153 Chr.15: 52312471-52312471 on Build GRCh38
rs77883523 Chr.15: 52312745-52312745 on Build GRCh38
rs537849315 Chr.15: 52312575-52312575 on Build GRCh38
rs1018914293 Chr.15: 52312795-52312795 on Build GRCh38
rs559898488 Chr.15: 52312515-52312515 on Build GRCh38
rs969925394 Chr.15: 52312411-52312411 on Build GRCh38
rs1027283806 Chr.15: 52312355-52312355 on Build GRCh38
rs568821628 Chr.15: 52312514-52312514 on Build GRCh38
rs188904089 Chr.15: 52312716-52312716 on Build GRCh38
rs984489997 Chr.15: 52312555-52312555 on Build GRCh38
rs1020015169 Chr.15: 52312384-52312384 on Build GRCh38
rs976415287 Chr.15: 52312757-52312757 on Build GRCh38
rs1006224440 Chr.15: 52312365-52312365 on Build GRCh38
rs28475475 Chr.15: 52312348-52312348 on Build GRCh38
rs888344438 Chr.15: 52312547-52312548 on Build GRCh38
rs1007872790 Chr.15: 52312775-52312775 on Build GRCh38
rs888799448 Chr.15: 52312770-52312770 on Build GRCh38
rs914174178 Chr.15: 52312736-52312736 on Build GRCh38
rs199930770 Chr.15: 52312369-52312370 on Build GRCh38
rs141598172 Chr.15: 52312444-52312444 on Build GRCh38
rs1040731530 Chr.15: 52312486-52312486 on Build GRCh38
rs150908009 Chr.15: 52312482-52312482 on Build GRCh38
rs1002763794 Chr.15: 52312635-52312635 on Build GRCh38
rs531298992 Chr.15: 52312607-52312607 on Build GRCh38
rs568192310 Chr.15: 52312657-52312657 on Build GRCh38
rs920994323 Chr.15: 52312758-52312758 on Build GRCh38
rs778356464 Chr.15: 52312351-52312351 on Build GRCh38
rs918711404 Chr.15: 52312521-52312521 on Build GRCh38
rs185603874 Chr.15: 52312450-52312450 on Build GRCh38
rs925522815 Chr.15: 52312420-52312420 on Build GRCh38
rs760978372 Chr.15: 52312395-52312395 on Build GRCh38
rs913541485 Chr.15: 52312468-52312468 on Build GRCh38
rs62015995 Chr.15: 52312336-52312336 on Build GRCh38
rs763225155 Chr.15: 52312776-52312776 on Build GRCh38
rs895969652 Chr.15: 52312638-52312638 on Build GRCh38
rs139335691 Chr.15: 52312483-52312483 on Build GRCh38
rs1025470565 Chr.15: 52312610-52312610 on Build GRCh38
rs28650960 Chr.15: 52312426-52312426 on Build GRCh38
rs368154854 Chr.15: 52312502-52312502 on Build GRCh38
rs562767390 Chr.15: 52312465-52312465 on Build GRCh38
rs1013650374 Chr.15: 52312381-52312381 on Build GRCh38
rs760186827 Chr.15: 52312785-52312785 on Build GRCh38
rs1025058346 Chr.15: 52312407-52312407 on Build GRCh38
rs112452193 Chr.15: 52312649-52312649 on Build GRCh38
rs375986156 Chr.15: 52312601-52312601 on Build GRCh38
rs902853939 Chr.15: 52312421-52312421 on Build GRCh38
rs930084485 Chr.15: 52312554-52312554 on Build GRCh38
rs888404678 Chr.15: 52312362-52312362 on Build GRCh38

View Full Product Details