Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 96333200-96333726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR1469
Entrez Gene ID
100302258
Gene Name
microRNA 1469
Gene Aliases
MIRN1469, hsa-mir-1469
UniGene
Gene Chromosome Location
Chr.15: 96333261-96333307 on Build GRCh38
Transcripts
NR_031715.1 NM_001145155.1 NM_001145156.1 NM_001145157.1 NM_021005.3

Gene Symbol
NR2F2
Entrez Gene ID
7026
Gene Name
nuclear receptor subfamily 2 group F member 2
Gene Aliases
ARP1, CHTD4, COUPTFB, COUPTFII, NF-E3, NR2F1, SVP40, TFCOUP2
UniGene
Hs.347991
Gene Chromosome Location
Chr.15: 96325928-96340263 on Build GRCh38
Transcripts
NR_031715.1 NM_001145155.1 NM_001145156.1 NM_001145157.1 NM_021005.3

SNP Detail

SNP ID Chromosome Location
rs909325570 Chr.15: 96333578-96333583 on Build GRCh38
rs974151068 Chr.15: 96333574-96333574 on Build GRCh38
rs961253741 Chr.15: 96333487-96333488 on Build GRCh38
rs1011754465 Chr.15: 96333442-96333442 on Build GRCh38
rs757925902 Chr.15: 96333476-96333476 on Build GRCh38
rs892872226 Chr.15: 96333479-96333479 on Build GRCh38
rs997146779 Chr.15: 96333694-96333694 on Build GRCh38
rs1049030751 Chr.15: 96333334-96333334 on Build GRCh38
rs969674891 Chr.15: 96333468-96333468 on Build GRCh38
rs1019945589 Chr.15: 96333508-96333508 on Build GRCh38
rs891508148 Chr.15: 96333425-96333425 on Build GRCh38
rs907462043 Chr.15: 96333457-96333457 on Build GRCh38
rs539024196 Chr.15: 96333653-96333653 on Build GRCh38
rs560226174 Chr.15: 96333282-96333282 on Build GRCh38
rs1030640684 Chr.15: 96333526-96333526 on Build GRCh38
rs911258881 Chr.15: 96333376-96333376 on Build GRCh38
rs1039094750 Chr.15: 96333609-96333609 on Build GRCh38
rs536581019 Chr.15: 96333579-96333579 on Build GRCh38
rs919435058 Chr.15: 96333557-96333557 on Build GRCh38
rs951352359 Chr.15: 96333578-96333578 on Build GRCh38
rs902882520 Chr.15: 96333513-96333513 on Build GRCh38
rs930209697 Chr.15: 96333424-96333424 on Build GRCh38
rs918626558 Chr.15: 96333409-96333409 on Build GRCh38
rs551175771 Chr.15: 96333505-96333505 on Build GRCh38
rs997197819 Chr.15: 96333525-96333525 on Build GRCh38
rs953325798 Chr.15: 96333529-96333529 on Build GRCh38
rs567596788 Chr.15: 96333563-96333563 on Build GRCh38
rs953305006 Chr.15: 96333363-96333363 on Build GRCh38
rs966788852 Chr.15: 96333528-96333538 on Build GRCh38
rs7173047 Chr.15: 96333382-96333382 on Build GRCh38
rs879787593 Chr.15: 96333368-96333368 on Build GRCh38
rs779366569 Chr.15: 96333278-96333278 on Build GRCh38
rs950809547 Chr.15: 96333561-96333562 on Build GRCh38
rs760113813 Chr.15: 96333340-96333340 on Build GRCh38
rs1016132198 Chr.15: 96333561-96333561 on Build GRCh38
rs1045871713 Chr.15: 96333227-96333227 on Build GRCh38
rs986336196 Chr.15: 96333553-96333553 on Build GRCh38
rs546578420 Chr.15: 96333589-96333589 on Build GRCh38
rs1034484134 Chr.15: 96333268-96333268 on Build GRCh38
rs976021873 Chr.15: 96333324-96333324 on Build GRCh38
rs527241233 Chr.15: 96333599-96333599 on Build GRCh38
rs992767986 Chr.15: 96333263-96333263 on Build GRCh38
rs984856055 Chr.15: 96333567-96333567 on Build GRCh38
rs981006826 Chr.15: 96333233-96333233 on Build GRCh38
rs960891041 Chr.15: 96333243-96333243 on Build GRCh38
rs1056366769 Chr.15: 96333652-96333652 on Build GRCh38
rs772835639 Chr.15: 96333307-96333307 on Build GRCh38
rs928839119 Chr.15: 96333597-96333597 on Build GRCh38
rs907386464 Chr.15: 96333241-96333241 on Build GRCh38
rs530978018 Chr.15: 96333417-96333417 on Build GRCh38
rs1055969188 Chr.15: 96333477-96333477 on Build GRCh38
rs997357234 Chr.15: 96333380-96333380 on Build GRCh38
rs947262784 Chr.15: 96333662-96333662 on Build GRCh38
rs562612328 Chr.15: 96333405-96333405 on Build GRCh38
rs574809934 Chr.15: 96333226-96333226 on Build GRCh38
rs190142071 Chr.15: 96333636-96333636 on Build GRCh38
rs754952993 Chr.15: 96333691-96333691 on Build GRCh38
rs879158049 Chr.15: 96333455-96333456 on Build GRCh38
rs892902444 Chr.15: 96333275-96333275 on Build GRCh38
rs937480565 Chr.15: 96333462-96333462 on Build GRCh38
rs1030157691 Chr.15: 96333352-96333352 on Build GRCh38
rs918572581 Chr.15: 96333577-96333577 on Build GRCh38
rs532236132 Chr.15: 96333369-96333369 on Build GRCh38
rs769559601 Chr.15: 96333263-96333264 on Build GRCh38
rs967287885 Chr.15: 96333323-96333323 on Build GRCh38
rs575711519 Chr.15: 96333671-96333671 on Build GRCh38
rs558743835 Chr.15: 96333664-96333664 on Build GRCh38
rs1011626691 Chr.15: 96333290-96333290 on Build GRCh38
rs923172908 Chr.15: 96333618-96333618 on Build GRCh38
rs540212013 Chr.15: 96333267-96333267 on Build GRCh38
rs981424622 Chr.15: 96333592-96333592 on Build GRCh38
rs1045990055 Chr.15: 96333456-96333456 on Build GRCh38

View Full Product Details