Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 372526-373027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM8A
Entrez Gene ID
58986
Gene Name
transmembrane protein 8A
Gene Aliases
M83, TMEM6, TMEM8
UniGene
Hs.288940
Gene Chromosome Location
Chr.16: 370776-381950 on Build GRCh38
Transcripts
NM_021259.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6306625 c.1981C>A p.P661T Chr.16: 372649-372649 on Build GRCh38
4960539 c.1979G>A p.G660E Chr.16: 372651-372651 on Build GRCh38
6144598 c.1967G>C p.W656S Chr.16: 372663-372663 on Build GRCh38
6804381 c.1936A>G p.M646V Chr.16: 372694-372694 on Build GRCh38
5575037 c.1999G>A p.V667M Chr.16: 372631-372631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007909660 Chr.16: 372966-372966 on Build GRCh38
rs756034774 Chr.16: 372630-372630 on Build GRCh38
rs925786983 Chr.16: 372797-372797 on Build GRCh38
rs780011039 Chr.16: 372631-372631 on Build GRCh38
rs767554044 Chr.16: 372718-372718 on Build GRCh38
rs768663498 Chr.16: 372617-372620 on Build GRCh38
rs561952784 Chr.16: 372752-372752 on Build GRCh38
rs757385172 Chr.16: 372747-372747 on Build GRCh38
rs767603254 Chr.16: 372626-372626 on Build GRCh38
rs1020450110 Chr.16: 372854-372873 on Build GRCh38
rs984786277 Chr.16: 372831-372831 on Build GRCh38
rs555031875 Chr.16: 372940-372940 on Build GRCh38
rs1021097379 Chr.16: 372736-372736 on Build GRCh38
rs376880071 Chr.16: 372732-372732 on Build GRCh38
rs753876139 Chr.16: 372845-372845 on Build GRCh38
rs896761898 Chr.16: 372959-372959 on Build GRCh38
rs567550157 Chr.16: 372667-372667 on Build GRCh38
rs375688501 Chr.16: 372698-372698 on Build GRCh38
rs746218290 Chr.16: 372694-372694 on Build GRCh38
rs926632813 Chr.16: 372856-372856 on Build GRCh38
rs953963877 Chr.16: 372778-372778 on Build GRCh38
rs934548783 Chr.16: 372652-372652 on Build GRCh38
rs180880774 Chr.16: 372969-372969 on Build GRCh38
rs972945255 Chr.16: 372992-372992 on Build GRCh38
rs1057089471 Chr.16: 372886-372886 on Build GRCh38
rs781350836 Chr.16: 372646-372647 on Build GRCh38
rs777915848 Chr.16: 372556-372556 on Build GRCh38
rs771391978 Chr.16: 372690-372690 on Build GRCh38
rs765677596 Chr.16: 373002-373002 on Build GRCh38
rs900487243 Chr.16: 372884-372884 on Build GRCh38
rs971496980 Chr.16: 372866-372875 on Build GRCh38
rs780806286 Chr.16: 372807-372807 on Build GRCh38
rs972610422 Chr.16: 372818-372818 on Build GRCh38
rs569048601 Chr.16: 372817-372817 on Build GRCh38
rs896750636 Chr.16: 372900-372900 on Build GRCh38
rs769431191 Chr.16: 372593-372593 on Build GRCh38
rs535670917 Chr.16: 372968-372968 on Build GRCh38
rs1049099741 Chr.16: 372899-372899 on Build GRCh38
rs1042567514 Chr.16: 372931-372931 on Build GRCh38
rs758523627 Chr.16: 372741-372741 on Build GRCh38
rs750242291 Chr.16: 372627-372627 on Build GRCh38
rs747431064 Chr.16: 372564-372564 on Build GRCh38
rs768191221 Chr.16: 372758-372758 on Build GRCh38
rs760638247 Chr.16: 372772-372772 on Build GRCh38
rs889481290 Chr.16: 372957-372957 on Build GRCh38
rs771991182 Chr.16: 372554-372554 on Build GRCh38
rs528615367 Chr.16: 372693-372693 on Build GRCh38
rs969680614 Chr.16: 372986-372986 on Build GRCh38
rs370609325 Chr.16: 372759-372759 on Build GRCh38
rs553921364 Chr.16: 372728-372728 on Build GRCh38
rs758675986 Chr.16: 372654-372654 on Build GRCh38
rs533658964 Chr.16: 372901-372901 on Build GRCh38
rs945494740 Chr.16: 372911-372911 on Build GRCh38
rs755481140 Chr.16: 372939-372939 on Build GRCh38
rs781260511 Chr.16: 372750-372750 on Build GRCh38
rs938230266 Chr.16: 372875-372875 on Build GRCh38
rs200542593 Chr.16: 372676-372676 on Build GRCh38
rs751104113 Chr.16: 372612-372612 on Build GRCh38
rs1005089022 Chr.16: 372916-372916 on Build GRCh38
rs115759137 Chr.16: 372601-372601 on Build GRCh38
rs374211218 Chr.16: 372755-372755 on Build GRCh38
rs934473876 Chr.16: 372808-372808 on Build GRCh38
rs777662862 Chr.16: 372675-372675 on Build GRCh38
rs748249711 Chr.16: 372549-372549 on Build GRCh38
rs546881073 Chr.16: 372830-372830 on Build GRCh38
rs144995005 Chr.16: 372697-372697 on Build GRCh38
rs575007316 Chr.16: 372945-372945 on Build GRCh38
rs978565181 Chr.16: 372621-372621 on Build GRCh38
rs999950751 Chr.16: 373003-373003 on Build GRCh38
rs961320760 Chr.16: 372978-372978 on Build GRCh38
rs890447552 Chr.16: 372666-372666 on Build GRCh38
rs771644618 Chr.16: 372762-372762 on Build GRCh38
rs372553836 Chr.16: 372731-372731 on Build GRCh38
rs969816377 Chr.16: 372791-372791 on Build GRCh38
rs147450150 Chr.16: 372793-372793 on Build GRCh38
rs773704912 Chr.16: 372715-372715 on Build GRCh38
rs190882418 Chr.16: 372658-372658 on Build GRCh38
rs900565987 Chr.16: 372685-372685 on Build GRCh38
rs772883753 Chr.16: 372767-372767 on Build GRCh38
rs566104555 Chr.16: 372896-372896 on Build GRCh38
rs370003191 Chr.16: 372709-372709 on Build GRCh38
rs371568475 Chr.16: 372744-372744 on Build GRCh38
rs761820371 Chr.16: 372624-372624 on Build GRCh38
rs199600677 Chr.16: 372704-372704 on Build GRCh38
rs759934087 Chr.16: 372578-372578 on Build GRCh38
rs745766808 Chr.16: 372589-372589 on Build GRCh38
rs200913077 Chr.16: 372600-372600 on Build GRCh38
rs1026854650 Chr.16: 372995-372995 on Build GRCh38
rs780144046 Chr.16: 372801-372801 on Build GRCh38
rs1022652805 Chr.16: 372973-372973 on Build GRCh38
rs780261427 Chr.16: 372753-372753 on Build GRCh38
rs775061407 Chr.16: 372594-372594 on Build GRCh38
rs961475769 Chr.16: 372816-372816 on Build GRCh38
rs528398828 Chr.16: 372826-372826 on Build GRCh38
rs747821245 Chr.16: 372761-372761 on Build GRCh38
rs200878430 Chr.16: 372571-372571 on Build GRCh38
rs1053073778 Chr.16: 372656-372656 on Build GRCh38
rs79047946 Chr.16: 372788-372788 on Build GRCh38
rs752798057 Chr.16: 372649-372649 on Build GRCh38

View Full Product Details