Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 567391-567886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC107987233
Entrez Gene ID
107987233
Gene Name
uncharacterized LOC107987233
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.16: 566056-568024 on Build GRCh38
Transcripts
XM_017023921.1 NM_001301159.1 NM_176677.2

Gene Symbol
NHLRC4
Entrez Gene ID
283948
Gene Name
NHL repeat containing 4
Gene Aliases
-
UniGene
Hs.664267
Gene Chromosome Location
Chr.16: 566995-569495 on Build GRCh38
Transcripts
XM_017023921.1 NM_001301159.1 NM_176677.2

SNP Detail

SNP ID Chromosome Location
rs1047297823 Chr.16: 567645-567645 on Build GRCh38
rs190879002 Chr.16: 567782-567782 on Build GRCh38
rs183394376 Chr.16: 567687-567687 on Build GRCh38
rs140645250 Chr.16: 567720-567720 on Build GRCh38
rs908586737 Chr.16: 567604-567604 on Build GRCh38
rs957304587 Chr.16: 567733-567733 on Build GRCh38
rs1054317120 Chr.16: 567597-567597 on Build GRCh38
rs75546591 Chr.16: 567435-567435 on Build GRCh38
rs868172931 Chr.16: 567620-567620 on Build GRCh38
rs952513654 Chr.16: 567602-567602 on Build GRCh38
rs1016677814 Chr.16: 567467-567467 on Build GRCh38
rs915903145 Chr.16: 567621-567621 on Build GRCh38
rs761708059 Chr.16: 567753-567753 on Build GRCh38
rs763049646 Chr.16: 567619-567619 on Build GRCh38
rs539014832 Chr.16: 567751-567751 on Build GRCh38
rs527948916 Chr.16: 567603-567603 on Build GRCh38
rs554715938 Chr.16: 567421-567421 on Build GRCh38
rs531971285 Chr.16: 567463-567463 on Build GRCh38
rs560390178 Chr.16: 567530-567530 on Build GRCh38
rs1032041822 Chr.16: 567811-567811 on Build GRCh38
rs765768656 Chr.16: 567739-567739 on Build GRCh38
rs1012728371 Chr.16: 567790-567790 on Build GRCh38
rs1019806435 Chr.16: 567727-567727 on Build GRCh38
rs576182104 Chr.16: 567789-567789 on Build GRCh38
rs2312776 Chr.16: 567683-567683 on Build GRCh38
rs1047466056 Chr.16: 567780-567780 on Build GRCh38
rs1026251437 Chr.16: 567428-567428 on Build GRCh38
rs1017067751 Chr.16: 567686-567686 on Build GRCh38
rs942753246 Chr.16: 567610-567610 on Build GRCh38
rs907077298 Chr.16: 567807-567807 on Build GRCh38
rs892225587 Chr.16: 567460-567460 on Build GRCh38
rs549424953 Chr.16: 567611-567611 on Build GRCh38
rs998867714 Chr.16: 567631-567631 on Build GRCh38
rs916886607 Chr.16: 567492-567492 on Build GRCh38
rs773501038 Chr.16: 567512-567512 on Build GRCh38
rs892186337 Chr.16: 567700-567700 on Build GRCh38
rs562067330 Chr.16: 567799-567799 on Build GRCh38
rs567803744 Chr.16: 567646-567646 on Build GRCh38
rs2312778 Chr.16: 567814-567814 on Build GRCh38
rs977459413 Chr.16: 567716-567716 on Build GRCh38
rs948756509 Chr.16: 567625-567625 on Build GRCh38
rs944908373 Chr.16: 567836-567836 on Build GRCh38
rs998781786 Chr.16: 567810-567810 on Build GRCh38
rs1013183682 Chr.16: 567413-567413 on Build GRCh38
rs952608480 Chr.16: 567769-567769 on Build GRCh38
rs750102774 Chr.16: 567798-567798 on Build GRCh38
rs957047873 Chr.16: 567824-567824 on Build GRCh38
rs1019289048 Chr.16: 567470-567470 on Build GRCh38
rs971319459 Chr.16: 567504-567504 on Build GRCh38
rs1040789244 Chr.16: 567865-567865 on Build GRCh38
rs958400486 Chr.16: 567468-567468 on Build GRCh38
rs536506174 Chr.16: 567779-567779 on Build GRCh38
rs1039867627 Chr.16: 567763-567763 on Build GRCh38
rs755730202 Chr.16: 567825-567825 on Build GRCh38
rs771701122 Chr.16: 567487-567487 on Build GRCh38
rs775474514 Chr.16: 567580-567580 on Build GRCh38
rs1039692054 Chr.16: 567612-567612 on Build GRCh38
rs982589996 Chr.16: 567775-567775 on Build GRCh38
rs115165076 Chr.16: 567757-567757 on Build GRCh38
rs546058057 Chr.16: 567531-567531 on Build GRCh38
rs186667504 Chr.16: 567770-567770 on Build GRCh38
rs79179803 Chr.16: 567469-567469 on Build GRCh38
rs74850920 Chr.16: 567436-567436 on Build GRCh38
rs1005635783 Chr.16: 567668-567668 on Build GRCh38
rs971354578 Chr.16: 567715-567715 on Build GRCh38
rs1038447713 Chr.16: 567786-567786 on Build GRCh38
rs1012781971 Chr.16: 567690-567690 on Build GRCh38
rs1043734816 Chr.16: 567415-567415 on Build GRCh38
rs564877551 Chr.16: 567461-567461 on Build GRCh38
rs764314248 Chr.16: 567783-567783 on Build GRCh38
rs550242007 Chr.16: 567708-567708 on Build GRCh38

View Full Product Details