Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 25164512-25165011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCMT1
Entrez Gene ID
51451
Gene Name
leucine carboxyl methyltransferase 1
Gene Aliases
CGI-68, LCMT, PPMT1
UniGene
Hs.337730
Gene Chromosome Location
Chr.16: 25111709-25178231 on Build GRCh38
Transcripts
NM_001032391.1 NM_016309.2 XM_005255354.3 XM_011545862.2 XM_011545864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5628426 c.796G>T p.E266* Chr.16: 25164713-25164713 on Build GRCh38
5628425 c.685G>T p.E229* Chr.16: 25164713-25164713 on Build GRCh38
3370373 c.628G>A p.A210T Chr.16: 25164656-25164656 on Build GRCh38
3280982 c.608A>G p.Y203C Chr.16: 25164636-25164636 on Build GRCh38
5208136 c.786C>G p.F262L Chr.16: 25164703-25164703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925747455 Chr.16: 25164742-25164742 on Build GRCh38
rs554375832 Chr.16: 25164954-25164954 on Build GRCh38
rs891152886 Chr.16: 25164955-25164955 on Build GRCh38
rs761046157 Chr.16: 25164645-25164645 on Build GRCh38
rs1024190789 Chr.16: 25164837-25164837 on Build GRCh38
rs898633541 Chr.16: 25164877-25164877 on Build GRCh38
rs1046010772 Chr.16: 25164790-25164790 on Build GRCh38
rs7342773 Chr.16: 25164947-25164947 on Build GRCh38
rs774907377 Chr.16: 25164591-25164591 on Build GRCh38
rs955529636 Chr.16: 25164656-25164656 on Build GRCh38
rs760993217 Chr.16: 25164762-25164762 on Build GRCh38
rs550609979 Chr.16: 25164685-25164685 on Build GRCh38
rs1055493782 Chr.16: 25164779-25164779 on Build GRCh38
rs373386079 Chr.16: 25164740-25164740 on Build GRCh38
rs898579887 Chr.16: 25164587-25164587 on Build GRCh38
rs758232893 Chr.16: 25164687-25164687 on Build GRCh38
rs773616958 Chr.16: 25164558-25164558 on Build GRCh38
rs546497114 Chr.16: 25164537-25164537 on Build GRCh38
rs763429375 Chr.16: 25164725-25164725 on Build GRCh38
rs778791989 Chr.16: 25164758-25164758 on Build GRCh38
rs552413993 Chr.16: 25164802-25164802 on Build GRCh38
rs377530979 Chr.16: 25164655-25164655 on Build GRCh38
rs199720154 Chr.16: 25164761-25164761 on Build GRCh38
rs559442959 Chr.16: 25164861-25164861 on Build GRCh38
rs199735451 Chr.16: 25164683-25164683 on Build GRCh38
rs538583081 Chr.16: 25164776-25164776 on Build GRCh38
rs1013852486 Chr.16: 25164795-25164795 on Build GRCh38
rs775788589 Chr.16: 25164605-25164605 on Build GRCh38
rs147289367 Chr.16: 25164712-25164712 on Build GRCh38
rs937564963 Chr.16: 25164824-25164824 on Build GRCh38
rs559727592 Chr.16: 25164538-25164538 on Build GRCh38
rs373044350 Chr.16: 25164601-25164601 on Build GRCh38
rs375789087 Chr.16: 25164749-25164749 on Build GRCh38
rs994216915 Chr.16: 25164610-25164610 on Build GRCh38
rs989951253 Chr.16: 25164980-25164980 on Build GRCh38
rs958185676 Chr.16: 25164970-25164970 on Build GRCh38
rs879148263 Chr.16: 25164908-25164908 on Build GRCh38
rs756750144 Chr.16: 25164681-25164681 on Build GRCh38
rs568692032 Chr.16: 25164669-25164669 on Build GRCh38
rs989537848 Chr.16: 25164624-25164624 on Build GRCh38
rs994269095 Chr.16: 25164894-25164894 on Build GRCh38
rs753527058 Chr.16: 25164678-25164678 on Build GRCh38
rs780064410 Chr.16: 25164691-25164691 on Build GRCh38
rs200758814 Chr.16: 25164581-25164581 on Build GRCh38
rs1008635411 Chr.16: 25164966-25164966 on Build GRCh38
rs1008835821 Chr.16: 25164667-25164667 on Build GRCh38
rs749349848 Chr.16: 25164564-25164564 on Build GRCh38
rs768072476 Chr.16: 25164603-25164603 on Build GRCh38
rs548412960 Chr.16: 25164639-25164639 on Build GRCh38
rs565761032 Chr.16: 25164906-25164906 on Build GRCh38
rs912117879 Chr.16: 25164725-25164725 on Build GRCh38
rs752358852 Chr.16: 25164579-25164579 on Build GRCh38
rs1033259423 Chr.16: 25164958-25164958 on Build GRCh38
rs927530087 Chr.16: 25164823-25164823 on Build GRCh38
rs921922130 Chr.16: 25164650-25164650 on Build GRCh38
rs745990576 Chr.16: 25164759-25164759 on Build GRCh38
rs778138977 Chr.16: 25164713-25164713 on Build GRCh38
rs769073236 Chr.16: 25164768-25164768 on Build GRCh38
rs746779348 Chr.16: 25164692-25164692 on Build GRCh38
rs750872217 Chr.16: 25164577-25164578 on Build GRCh38

View Full Product Details