Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31111777-31112268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCKDK
Entrez Gene ID
10295
Gene Name
branched chain ketoacid dehydrogenase kinase
Gene Aliases
BCKDKD, BDK
UniGene
Hs.513520
Gene Chromosome Location
Chr.16: 31108294-31117651 on Build GRCh38
Transcripts
NM_001122957.2 NM_001271926.1 NM_005881.3 XM_017022859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4522838 c.1182G>A p.T394T Chr.16: 31112208-31112208 on Build GRCh38
6002324 c.1130C>T p.A377V Chr.16: 31112156-31112156 on Build GRCh38
4514054 c.960C>T p.I320I Chr.16: 31111893-31111893 on Build GRCh38
6249098 c.1186G>T p.V396F Chr.16: 31112212-31112212 on Build GRCh38
4523542 c.1219G>T p.E407* Chr.16: 31112245-31112245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778430202 Chr.16: 31112006-31112006 on Build GRCh38
rs748188451 Chr.16: 31112060-31112060 on Build GRCh38
rs201881945 Chr.16: 31111828-31111828 on Build GRCh38
rs755584594 Chr.16: 31112005-31112005 on Build GRCh38
rs889660794 Chr.16: 31111812-31111812 on Build GRCh38
rs775185560 Chr.16: 31112194-31112194 on Build GRCh38
rs759341581 Chr.16: 31112099-31112099 on Build GRCh38
rs758812308 Chr.16: 31112027-31112027 on Build GRCh38
rs999998050 Chr.16: 31111893-31111893 on Build GRCh38
rs746177286 Chr.16: 31111937-31111937 on Build GRCh38
rs898935750 Chr.16: 31111817-31111817 on Build GRCh38
rs780604144 Chr.16: 31112034-31112034 on Build GRCh38
rs372199017 Chr.16: 31111954-31111954 on Build GRCh38
rs766922526 Chr.16: 31112222-31112222 on Build GRCh38
rs763653927 Chr.16: 31112207-31112207 on Build GRCh38
rs398124213 Chr.16: 31112175-31112189 on Build GRCh38
rs747227092 Chr.16: 31112037-31112037 on Build GRCh38
rs745623913 Chr.16: 31112179-31112179 on Build GRCh38
rs918369917 Chr.16: 31111997-31111997 on Build GRCh38
rs142542453 Chr.16: 31111999-31111999 on Build GRCh38
rs199838222 Chr.16: 31111943-31111943 on Build GRCh38
rs543643875 Chr.16: 31112191-31112191 on Build GRCh38
rs767003668 Chr.16: 31112001-31112001 on Build GRCh38
rs111551851 Chr.16: 31112142-31112142 on Build GRCh38
rs758137216 Chr.16: 31111858-31111858 on Build GRCh38
rs1035318937 Chr.16: 31112185-31112185 on Build GRCh38
rs776835457 Chr.16: 31111971-31111971 on Build GRCh38
rs531966645 Chr.16: 31112212-31112212 on Build GRCh38
rs780549207 Chr.16: 31112141-31112141 on Build GRCh38
rs935378287 Chr.16: 31112067-31112067 on Build GRCh38
rs546815456 Chr.16: 31111964-31111964 on Build GRCh38
rs756353382 Chr.16: 31112242-31112242 on Build GRCh38
rs769709880 Chr.16: 31111976-31111976 on Build GRCh38
rs778775154 Chr.16: 31112247-31112247 on Build GRCh38
rs749299758 Chr.16: 31112243-31112243 on Build GRCh38
rs374677650 Chr.16: 31111859-31111859 on Build GRCh38
rs569914362 Chr.16: 31111809-31111809 on Build GRCh38
rs761640047 Chr.16: 31111975-31111975 on Build GRCh38
rs754464146 Chr.16: 31111889-31111889 on Build GRCh38
rs767598142 Chr.16: 31111950-31111951 on Build GRCh38
rs777785417 Chr.16: 31112153-31112153 on Build GRCh38
rs764935168 Chr.16: 31111839-31111839 on Build GRCh38
rs141335706 Chr.16: 31112211-31112211 on Build GRCh38
rs959968340 Chr.16: 31112190-31112190 on Build GRCh38
rs748371469 Chr.16: 31112147-31112147 on Build GRCh38
rs753006924 Chr.16: 31112158-31112159 on Build GRCh38
rs760182220 Chr.16: 31112114-31112114 on Build GRCh38
rs779301595 Chr.16: 31111913-31111913 on Build GRCh38
rs201445468 Chr.16: 31111894-31111894 on Build GRCh38
rs759040301 Chr.16: 31112137-31112137 on Build GRCh38
rs577985544 Chr.16: 31112009-31112009 on Build GRCh38
rs773183825 Chr.16: 31112066-31112066 on Build GRCh38
rs749756059 Chr.16: 31111911-31111911 on Build GRCh38
rs769790766 Chr.16: 31112061-31112061 on Build GRCh38
rs1009193055 Chr.16: 31111915-31111915 on Build GRCh38
rs750287305 Chr.16: 31111851-31111851 on Build GRCh38
rs750916023 Chr.16: 31112026-31112026 on Build GRCh38
rs751238597 Chr.16: 31111869-31111869 on Build GRCh38
rs771457546 Chr.16: 31112092-31112092 on Build GRCh38
rs55939386 Chr.16: 31112238-31112238 on Build GRCh38
rs886238805 Chr.16: 31112232-31112232 on Build GRCh38
rs770711315 Chr.16: 31112087-31112087 on Build GRCh38
rs761419965 Chr.16: 31112221-31112221 on Build GRCh38
rs937650508 Chr.16: 31112167-31112167 on Build GRCh38
rs529218478 Chr.16: 31112231-31112231 on Build GRCh38
rs376691315 Chr.16: 31111978-31111978 on Build GRCh38
rs761847817 Chr.16: 31111835-31111835 on Build GRCh38
rs774276125 Chr.16: 31112166-31112166 on Build GRCh38
rs764289146 Chr.16: 31111826-31111826 on Build GRCh38
rs922590624 Chr.16: 31112054-31112054 on Build GRCh38
rs886306366 Chr.16: 31111880-31111880 on Build GRCh38
rs752324023 Chr.16: 31112103-31112103 on Build GRCh38
rs959757913 Chr.16: 31111990-31111990 on Build GRCh38
rs139044149 Chr.16: 31112131-31112131 on Build GRCh38
rs199683653 Chr.16: 31112025-31112025 on Build GRCh38
rs942651732 Chr.16: 31111822-31111822 on Build GRCh38
rs760283230 Chr.16: 31112202-31112202 on Build GRCh38
rs752139664 Chr.16: 31112227-31112227 on Build GRCh38
rs1055253754 Chr.16: 31111810-31111810 on Build GRCh38
rs747282203 Chr.16: 31111960-31111960 on Build GRCh38
rs377552512 Chr.16: 31112235-31112235 on Build GRCh38
rs531591891 Chr.16: 31112004-31112004 on Build GRCh38
rs769054677 Chr.16: 31112048-31112048 on Build GRCh38
rs148848087 Chr.16: 31111980-31111980 on Build GRCh38
rs201431301 Chr.16: 31111844-31111844 on Build GRCh38
rs756189742 Chr.16: 31112149-31112149 on Build GRCh38
rs151090898 Chr.16: 31112157-31112157 on Build GRCh38
rs781279805 Chr.16: 31112058-31112058 on Build GRCh38
rs749211768 Chr.16: 31112154-31112154 on Build GRCh38
rs781432488 Chr.16: 31112146-31112146 on Build GRCh38
rs11540063 Chr.16: 31111966-31111966 on Build GRCh38
rs560919863 Chr.16: 31111972-31111972 on Build GRCh38
rs377423130 Chr.16: 31111912-31111912 on Build GRCh38
rs534671893 Chr.16: 31111992-31111992 on Build GRCh38
rs200213677 Chr.16: 31111991-31111991 on Build GRCh38
rs772595672 Chr.16: 31111947-31111947 on Build GRCh38
rs145180240 Chr.16: 31112244-31112244 on Build GRCh38
rs201614853 Chr.16: 31112130-31112130 on Build GRCh38
rs901201449 Chr.16: 31111865-31111865 on Build GRCh38
rs762920594 Chr.16: 31112070-31112070 on Build GRCh38
rs776686374 Chr.16: 31111834-31111834 on Build GRCh38
rs767131632 Chr.16: 31112101-31112101 on Build GRCh38

View Full Product Details