Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44145062-44145560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf53
Entrez Gene ID
78995
Gene Name
chromosome 17 open reading frame 53
Gene Aliases
-
UniGene
Hs.437059
Gene Chromosome Location
Chr.17: 44141902-44162477 on Build GRCh38
Transcripts
NM_001171251.2 NM_001321310.1 NM_001321311.1 NM_024032.4 XM_011525187.1 XM_011525188.1 XM_011525189.1 XM_011525190.1 XM_011525191.2 XM_011525192.1 XM_011525193.1 XM_011525194.1 XM_011525195.1 XM_017025031.1 XM_017025032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
472871 c.17A>G p.Q6R Chr.17: 44145216-44145216 on Build GRCh38
6614666 c.4-7delT p.? Chr.17: 44145196-44145196 on Build GRCh38
460320 c.53A>C p.E18A Chr.17: 44145252-44145252 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758597492 Chr.17: 44145265-44145265 on Build GRCh38
rs550022902 Chr.17: 44145110-44145110 on Build GRCh38
rs553730961 Chr.17: 44145205-44145205 on Build GRCh38
rs574394445 Chr.17: 44145463-44145463 on Build GRCh38
rs764219272 Chr.17: 44145204-44145204 on Build GRCh38
rs757807182 Chr.17: 44145281-44145281 on Build GRCh38
rs950467262 Chr.17: 44145502-44145502 on Build GRCh38
rs916884272 Chr.17: 44145529-44145529 on Build GRCh38
rs762651837 Chr.17: 44145202-44145202 on Build GRCh38
rs770764734 Chr.17: 44145159-44145159 on Build GRCh38
rs983563261 Chr.17: 44145092-44145093 on Build GRCh38
rs536741686 Chr.17: 44145269-44145269 on Build GRCh38
rs541859904 Chr.17: 44145531-44145531 on Build GRCh38
rs776797320 Chr.17: 44145186-44145187 on Build GRCh38
rs776306997 Chr.17: 44145278-44145278 on Build GRCh38
rs559378791 Chr.17: 44145439-44145439 on Build GRCh38
rs200564421 Chr.17: 44145195-44145195 on Build GRCh38
rs199580277 Chr.17: 44145254-44145254 on Build GRCh38
rs763152490 Chr.17: 44145123-44145123 on Build GRCh38
rs950820739 Chr.17: 44145088-44145089 on Build GRCh38
rs756476781 Chr.17: 44145287-44145287 on Build GRCh38
rs979998204 Chr.17: 44145169-44145169 on Build GRCh38
rs768598388 Chr.17: 44145184-44145185 on Build GRCh38
rs558863087 Chr.17: 44145385-44145385 on Build GRCh38
rs762596657 Chr.17: 44145305-44145305 on Build GRCh38
rs935918281 Chr.17: 44145186-44145186 on Build GRCh38
rs867401444 Chr.17: 44145448-44145448 on Build GRCh38
rs185548039 Chr.17: 44145190-44145190 on Build GRCh38
rs766791699 Chr.17: 44145247-44145247 on Build GRCh38
rs188371540 Chr.17: 44145413-44145413 on Build GRCh38
rs756375893 Chr.17: 44145469-44145469 on Build GRCh38
rs917230109 Chr.17: 44145119-44145119 on Build GRCh38
rs983541659 Chr.17: 44145527-44145527 on Build GRCh38
rs778633249 Chr.17: 44145479-44145479 on Build GRCh38
rs201278031 Chr.17: 44145270-44145270 on Build GRCh38
rs748268833 Chr.17: 44145256-44145256 on Build GRCh38
rs1027745675 Chr.17: 44145423-44145423 on Build GRCh38
rs1009117291 Chr.17: 44145354-44145354 on Build GRCh38
rs778926360 Chr.17: 44145255-44145255 on Build GRCh38
rs770697806 Chr.17: 44145274-44145274 on Build GRCh38
rs767835644 Chr.17: 44145286-44145286 on Build GRCh38
rs780358506 Chr.17: 44145321-44145321 on Build GRCh38
rs531813172 Chr.17: 44145102-44145102 on Build GRCh38
rs994620036 Chr.17: 44145417-44145417 on Build GRCh38
rs571624710 Chr.17: 44145187-44145187 on Build GRCh38
rs115298176 Chr.17: 44145409-44145409 on Build GRCh38
rs576141709 Chr.17: 44145306-44145306 on Build GRCh38
rs1039355820 Chr.17: 44145381-44145381 on Build GRCh38
rs775846312 Chr.17: 44145466-44145466 on Build GRCh38
rs773084780 Chr.17: 44145187-44145187 on Build GRCh38
rs756557771 Chr.17: 44145245-44145245 on Build GRCh38
rs537256877 Chr.17: 44145347-44145347 on Build GRCh38
rs769880014 Chr.17: 44145284-44145284 on Build GRCh38
rs942408406 Chr.17: 44145343-44145343 on Build GRCh38

View Full Product Details