Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59737363-59737764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VMP1
Entrez Gene ID
81671
Gene Name
vacuole membrane protein 1
Gene Aliases
EPG3, TANGO5, TMEM49
UniGene
Hs.444569
Gene Chromosome Location
Chr.17: 59707502-59840593 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5724280 c.223C>T p.R75C Chr.17: 59737463-59737463 on Build GRCh38
1686750 c.213-1G>A p.? Chr.17: 59737452-59737452 on Build GRCh38
1384921 c.215T>C p.L72S Chr.17: 59737455-59737455 on Build GRCh38
4420118 c.223C>G p.R75G Chr.17: 59737463-59737463 on Build GRCh38
6201830 c.276G>A p.T92T Chr.17: 59737516-59737516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992165230 Chr.17: 59737738-59737738 on Build GRCh38
rs542286366 Chr.17: 59737546-59737546 on Build GRCh38
rs892424347 Chr.17: 59737631-59737631 on Build GRCh38
rs748315945 Chr.17: 59737419-59737419 on Build GRCh38
rs747483842 Chr.17: 59737478-59737478 on Build GRCh38
rs1036023507 Chr.17: 59737390-59737390 on Build GRCh38
rs368710608 Chr.17: 59737554-59737554 on Build GRCh38
rs754836462 Chr.17: 59737507-59737507 on Build GRCh38
rs776758308 Chr.17: 59737608-59737608 on Build GRCh38
rs764829047 Chr.17: 59737594-59737594 on Build GRCh38
rs759988312 Chr.17: 59737439-59737439 on Build GRCh38
rs781173942 Chr.17: 59737431-59737432 on Build GRCh38
rs898803383 Chr.17: 59737402-59737402 on Build GRCh38
rs763043785 Chr.17: 59737430-59737430 on Build GRCh38
rs779981605 Chr.17: 59737493-59737493 on Build GRCh38
rs767028719 Chr.17: 59737431-59737431 on Build GRCh38
rs776898365 Chr.17: 59737413-59737413 on Build GRCh38
rs746935995 Chr.17: 59737499-59737499 on Build GRCh38
rs755026145 Chr.17: 59737432-59737432 on Build GRCh38
rs1033013788 Chr.17: 59737512-59737512 on Build GRCh38
rs367803899 Chr.17: 59737460-59737460 on Build GRCh38
rs773146447 Chr.17: 59737428-59737428 on Build GRCh38
rs769960034 Chr.17: 59737420-59737420 on Build GRCh38
rs749386849 Chr.17: 59737553-59737553 on Build GRCh38
rs1009402952 Chr.17: 59737655-59737655 on Build GRCh38
rs755917088 Chr.17: 59737514-59737515 on Build GRCh38
rs373947335 Chr.17: 59737441-59737441 on Build GRCh38
rs576981057 Chr.17: 59737721-59737721 on Build GRCh38
rs768031075 Chr.17: 59737568-59737568 on Build GRCh38
rs372269498 Chr.17: 59737562-59737562 on Build GRCh38
rs777663985 Chr.17: 59737593-59737594 on Build GRCh38
rs768685950 Chr.17: 59737481-59737481 on Build GRCh38
rs371717717 Chr.17: 59737470-59737470 on Build GRCh38
rs1022092100 Chr.17: 59737714-59737714 on Build GRCh38
rs200132207 Chr.17: 59737579-59737579 on Build GRCh38
rs764849959 Chr.17: 59737471-59737471 on Build GRCh38
rs769934426 Chr.17: 59737540-59737540 on Build GRCh38
rs780819533 Chr.17: 59737508-59737508 on Build GRCh38
rs541346384 Chr.17: 59737692-59737692 on Build GRCh38
rs752642708 Chr.17: 59737437-59737440 on Build GRCh38
rs917619679 Chr.17: 59737477-59737477 on Build GRCh38
rs756827363 Chr.17: 59737463-59737463 on Build GRCh38
rs575292321 Chr.17: 59737433-59737433 on Build GRCh38
rs560922727 Chr.17: 59737573-59737573 on Build GRCh38
rs374605118 Chr.17: 59737527-59737527 on Build GRCh38
rs1034866642 Chr.17: 59737739-59737739 on Build GRCh38

View Full Product Details