Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49291878-49292387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF652
Entrez Gene ID
22834
Gene Name
zinc finger protein 652
Gene Aliases
-
UniGene
Hs.463375
Gene Chromosome Location
Chr.17: 49289206-49362473 on Build GRCh38
Transcripts
NM_001145365.1 NM_014897.2 NR_135579.1 XM_005257166.2

SNP Detail

SNP ID Chromosome Location
rs959460688 Chr.17: 49292073-49292073 on Build GRCh38
rs923887400 Chr.17: 49292198-49292198 on Build GRCh38
rs555026849 Chr.17: 49292318-49292318 on Build GRCh38
rs941514548 Chr.17: 49292359-49292359 on Build GRCh38
rs143776240 Chr.17: 49291951-49291951 on Build GRCh38
rs905220983 Chr.17: 49292100-49292100 on Build GRCh38
rs990849597 Chr.17: 49292115-49292115 on Build GRCh38
rs902986991 Chr.17: 49292176-49292176 on Build GRCh38
rs1038591563 Chr.17: 49292062-49292062 on Build GRCh38
rs777311019 Chr.17: 49291921-49291922 on Build GRCh38
rs1000696987 Chr.17: 49291926-49291926 on Build GRCh38
rs937995966 Chr.17: 49292104-49292104 on Build GRCh38
rs200069600 Chr.17: 49292225-49292226 on Build GRCh38
rs963733055 Chr.17: 49292221-49292221 on Build GRCh38
rs866336386 Chr.17: 49292013-49292013 on Build GRCh38
rs995951016 Chr.17: 49292343-49292343 on Build GRCh38
rs1032908917 Chr.17: 49292215-49292215 on Build GRCh38
rs757123340 Chr.17: 49292159-49292159 on Build GRCh38
rs72835478 Chr.17: 49292119-49292119 on Build GRCh38
rs920062699 Chr.17: 49292063-49292063 on Build GRCh38
rs1029455201 Chr.17: 49292346-49292346 on Build GRCh38
rs4794037 Chr.17: 49291940-49291940 on Build GRCh38
rs1053640067 Chr.17: 49292274-49292274 on Build GRCh38
rs763834726 Chr.17: 49291971-49291971 on Build GRCh38
rs954344738 Chr.17: 49291980-49291980 on Build GRCh38
rs908672065 Chr.17: 49292026-49292026 on Build GRCh38
rs1000539471 Chr.17: 49292214-49292215 on Build GRCh38
rs544870949 Chr.17: 49292351-49292351 on Build GRCh38
rs558486220 Chr.17: 49292353-49292353 on Build GRCh38
rs1014967385 Chr.17: 49292352-49292352 on Build GRCh38
rs187873548 Chr.17: 49292349-49292349 on Build GRCh38
rs987092013 Chr.17: 49291985-49291985 on Build GRCh38
rs961897909 Chr.17: 49292029-49292029 on Build GRCh38
rs146342527 Chr.17: 49291975-49291975 on Build GRCh38
rs975065833 Chr.17: 49292323-49292323 on Build GRCh38
rs936562904 Chr.17: 49292241-49292241 on Build GRCh38
rs900320838 Chr.17: 49292308-49292308 on Build GRCh38
rs983350739 Chr.17: 49292011-49292011 on Build GRCh38
rs941617837 Chr.17: 49292038-49292038 on Build GRCh38
rs974269402 Chr.17: 49292035-49292035 on Build GRCh38
rs553075442 Chr.17: 49292132-49292132 on Build GRCh38
rs80150712 Chr.17: 49292225-49292225 on Build GRCh38
rs535453184 Chr.17: 49292253-49292253 on Build GRCh38
rs550370703 Chr.17: 49291962-49291962 on Build GRCh38
rs114991573 Chr.17: 49292172-49292172 on Build GRCh38
rs1015842189 Chr.17: 49292007-49292007 on Build GRCh38
rs1051509493 Chr.17: 49292345-49292345 on Build GRCh38

View Full Product Details