Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50118072-50118572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAMD14
Entrez Gene ID
201191
Gene Name
sterile alpha motif domain containing 14
Gene Aliases
-
UniGene
Hs.567769
Gene Chromosome Location
Chr.17: 50111309-50129882 on Build GRCh38
Transcripts
NM_001257359.1 NM_174920.3 XM_017024322.1 XM_017024323.1 XM_017024324.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2698875 c.155G>T p.R52L Chr.17: 50118216-50118216 on Build GRCh38
5386647 c.128G>A p.R43K Chr.17: 50118243-50118243 on Build GRCh38
1135959 c.192G>A p.S64S Chr.17: 50118179-50118179 on Build GRCh38
3819915 c.191C>G p.S64W Chr.17: 50118180-50118180 on Build GRCh38
6147338 c.163G>A p.A55T Chr.17: 50118208-50118208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558349040 Chr.17: 50118337-50118337 on Build GRCh38
rs956547570 Chr.17: 50118498-50118498 on Build GRCh38
rs752937234 Chr.17: 50118338-50118338 on Build GRCh38
rs574738018 Chr.17: 50118451-50118451 on Build GRCh38
rs1023994347 Chr.17: 50118242-50118242 on Build GRCh38
rs75166937 Chr.17: 50118485-50118485 on Build GRCh38
rs763099525 Chr.17: 50118328-50118328 on Build GRCh38
rs1002001604 Chr.17: 50118484-50118484 on Build GRCh38
rs367654801 Chr.17: 50118197-50118197 on Build GRCh38
rs960994535 Chr.17: 50118237-50118237 on Build GRCh38
rs368368138 Chr.17: 50118169-50118169 on Build GRCh38
rs113701481 Chr.17: 50118545-50118545 on Build GRCh38
rs776235505 Chr.17: 50118270-50118270 on Build GRCh38
rs149737093 Chr.17: 50118266-50118266 on Build GRCh38
rs745369307 Chr.17: 50118236-50118236 on Build GRCh38
rs150420736 Chr.17: 50118344-50118344 on Build GRCh38
rs751097859 Chr.17: 50118356-50118356 on Build GRCh38
rs933656721 Chr.17: 50118384-50118384 on Build GRCh38
rs751877592 Chr.17: 50118198-50118198 on Build GRCh38
rs773355089 Chr.17: 50118190-50118190 on Build GRCh38
rs911471098 Chr.17: 50118363-50118363 on Build GRCh38
rs747878270 Chr.17: 50118364-50118364 on Build GRCh38
rs753127511 Chr.17: 50118217-50118217 on Build GRCh38
rs769610479 Chr.17: 50118292-50118292 on Build GRCh38
rs756519445 Chr.17: 50118227-50118227 on Build GRCh38
rs146044799 Chr.17: 50118546-50118546 on Build GRCh38
rs779650861 Chr.17: 50118247-50118247 on Build GRCh38
rs946946773 Chr.17: 50118339-50118339 on Build GRCh38
rs372332569 Chr.17: 50118352-50118352 on Build GRCh38
rs775182718 Chr.17: 50118174-50118174 on Build GRCh38
rs765238669 Chr.17: 50118180-50118180 on Build GRCh38
rs369989633 Chr.17: 50118351-50118351 on Build GRCh38
rs750659916 Chr.17: 50118364-50118393 on Build GRCh38
rs531127700 Chr.17: 50118533-50118533 on Build GRCh38
rs767732998 Chr.17: 50118214-50118214 on Build GRCh38
rs201749689 Chr.17: 50118374-50118374 on Build GRCh38
rs1023520497 Chr.17: 50118461-50118461 on Build GRCh38
rs1046697460 Chr.17: 50118448-50118448 on Build GRCh38
rs756559654 Chr.17: 50118341-50118341 on Build GRCh38
rs910759469 Chr.17: 50118549-50118549 on Build GRCh38
rs34204016 Chr.17: 50118388-50118388 on Build GRCh38
rs768377643 Chr.17: 50118177-50118177 on Build GRCh38
rs145637965 Chr.17: 50118302-50118302 on Build GRCh38
rs905026740 Chr.17: 50118482-50118482 on Build GRCh38
rs766604639 Chr.17: 50118331-50118331 on Build GRCh38
rs914153921 Chr.17: 50118279-50118279 on Build GRCh38
rs1035068685 Chr.17: 50118200-50118200 on Build GRCh38
rs754384281 Chr.17: 50118346-50118346 on Build GRCh38
rs739500 Chr.17: 50118454-50118454 on Build GRCh38
rs971056375 Chr.17: 50118481-50118481 on Build GRCh38
rs528590646 Chr.17: 50118532-50118532 on Build GRCh38
rs538760933 Chr.17: 50118179-50118179 on Build GRCh38
rs780960762 Chr.17: 50118362-50118362 on Build GRCh38
rs868369292 Chr.17: 50118322-50118322 on Build GRCh38
rs377680616 Chr.17: 50118369-50118369 on Build GRCh38
rs531650995 Chr.17: 50118455-50118458 on Build GRCh38
rs267604941 Chr.17: 50118168-50118168 on Build GRCh38
rs993864916 Chr.17: 50118429-50118429 on Build GRCh38
rs149556958 Chr.17: 50118529-50118529 on Build GRCh38
rs778410627 Chr.17: 50118228-50118228 on Build GRCh38
rs739499 Chr.17: 50118550-50118550 on Build GRCh38
rs773336853 Chr.17: 50118307-50118307 on Build GRCh38
rs1048156126 Chr.17: 50118434-50118434 on Build GRCh38
rs968672235 Chr.17: 50118547-50118547 on Build GRCh38
rs138169376 Chr.17: 50118359-50118359 on Build GRCh38
rs745513259 Chr.17: 50118456-50118456 on Build GRCh38
rs745395759 Chr.17: 50118150-50118150 on Build GRCh38
rs774602078 Chr.17: 50118336-50118336 on Build GRCh38
rs747983994 Chr.17: 50118278-50118278 on Build GRCh38
rs576341829 Chr.17: 50118496-50118496 on Build GRCh38
rs565202607 Chr.17: 50118530-50118530 on Build GRCh38
rs778236271 Chr.17: 50118136-50118136 on Build GRCh38
rs755337590 Chr.17: 50118199-50118199 on Build GRCh38
rs200210474 Chr.17: 50118246-50118246 on Build GRCh38
rs746627130 Chr.17: 50118263-50118263 on Build GRCh38
rs374109001 Chr.17: 50118195-50118195 on Build GRCh38
rs897356170 Chr.17: 50118401-50118401 on Build GRCh38

View Full Product Details