Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78356694-78357193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS3
Entrez Gene ID
9021
Gene Name
suppressor of cytokine signaling 3
Gene Aliases
ATOD4, CIS3, Cish3, SOCS-3, SSI-3, SSI3
UniGene
Hs.527973
Gene Chromosome Location
Chr.17: 78356777-78360079 on Build GRCh38
Transcripts
NM_003955.4

SNP Detail

SNP ID Chromosome Location
rs981124625 Chr.17: 78356735-78356735 on Build GRCh38
rs994474649 Chr.17: 78357054-78357054 on Build GRCh38
rs576391965 Chr.17: 78357023-78357023 on Build GRCh38
rs904782598 Chr.17: 78357158-78357158 on Build GRCh38
rs752619845 Chr.17: 78357157-78357157 on Build GRCh38
rs969935947 Chr.17: 78357036-78357036 on Build GRCh38
rs930341743 Chr.17: 78356715-78356715 on Build GRCh38
rs946056066 Chr.17: 78356937-78356937 on Build GRCh38
rs956150248 Chr.17: 78356771-78356771 on Build GRCh38
rs928151475 Chr.17: 78356742-78356742 on Build GRCh38
rs887244408 Chr.17: 78356731-78356737 on Build GRCh38
rs1024178483 Chr.17: 78356779-78356779 on Build GRCh38
rs565481649 Chr.17: 78356743-78356743 on Build GRCh38
rs980046535 Chr.17: 78356786-78356787 on Build GRCh38
rs557730211 Chr.17: 78356816-78356820 on Build GRCh38
rs897482663 Chr.17: 78357029-78357029 on Build GRCh38
rs891875300 Chr.17: 78357027-78357028 on Build GRCh38
rs970345699 Chr.17: 78356786-78356786 on Build GRCh38
rs555026188 Chr.17: 78357016-78357017 on Build GRCh38
rs558988561 Chr.17: 78357098-78357098 on Build GRCh38
rs975857301 Chr.17: 78357112-78357112 on Build GRCh38
rs907574216 Chr.17: 78357100-78357100 on Build GRCh38
rs78818634 Chr.17: 78357114-78357115 on Build GRCh38
rs938757580 Chr.17: 78356917-78356917 on Build GRCh38
rs980695874 Chr.17: 78356844-78356844 on Build GRCh38
rs561141527 Chr.17: 78356739-78356739 on Build GRCh38
rs904454249 Chr.17: 78356953-78356953 on Build GRCh38
rs1050693058 Chr.17: 78356960-78356960 on Build GRCh38
rs113024182 Chr.17: 78357013-78357013 on Build GRCh38
rs890327295 Chr.17: 78356971-78356971 on Build GRCh38
rs770095322 Chr.17: 78356837-78356837 on Build GRCh38
rs768656820 Chr.17: 78357113-78357113 on Build GRCh38
rs1031706139 Chr.17: 78356751-78356752 on Build GRCh38
rs1041240415 Chr.17: 78356736-78356736 on Build GRCh38
rs536972541 Chr.17: 78356974-78356974 on Build GRCh38
rs375207007 Chr.17: 78356914-78356914 on Build GRCh38
rs993070064 Chr.17: 78356772-78356772 on Build GRCh38
rs530233894 Chr.17: 78357060-78357061 on Build GRCh38
rs796507045 Chr.17: 78357029-78357029 on Build GRCh38
rs774218172 Chr.17: 78357166-78357166 on Build GRCh38
rs936293643 Chr.17: 78357165-78357165 on Build GRCh38
rs534412733 Chr.17: 78356754-78356754 on Build GRCh38
rs1001678143 Chr.17: 78357163-78357163 on Build GRCh38
rs147887475 Chr.17: 78356731-78356731 on Build GRCh38
rs901419125 Chr.17: 78356738-78356738 on Build GRCh38
rs961724804 Chr.17: 78356725-78356725 on Build GRCh38
rs141500264 Chr.17: 78356838-78356838 on Build GRCh38
rs1028705497 Chr.17: 78356743-78356743 on Build GRCh38
rs1000211656 Chr.17: 78356749-78356749 on Build GRCh38
rs1042968274 Chr.17: 78356941-78356941 on Build GRCh38
rs550408744 Chr.17: 78357105-78357105 on Build GRCh38
rs1047530889 Chr.17: 78356730-78356731 on Build GRCh38
rs556531028 Chr.17: 78356976-78356976 on Build GRCh38
rs1050026887 Chr.17: 78357009-78357012 on Build GRCh38
rs539072429 Chr.17: 78357024-78357024 on Build GRCh38
rs1027677632 Chr.17: 78356820-78356820 on Build GRCh38
rs992948499 Chr.17: 78356926-78356926 on Build GRCh38
rs1024519355 Chr.17: 78357034-78357034 on Build GRCh38
rs1050373126 Chr.17: 78357114-78357119 on Build GRCh38
rs913234222 Chr.17: 78356929-78356929 on Build GRCh38
rs376577899 Chr.17: 78357024-78357024 on Build GRCh38
rs866908336 Chr.17: 78356744-78356744 on Build GRCh38
rs931793104 Chr.17: 78356958-78356958 on Build GRCh38
rs567745033 Chr.17: 78356915-78356915 on Build GRCh38
rs951964738 Chr.17: 78357072-78357072 on Build GRCh38
rs980398153 Chr.17: 78357056-78357056 on Build GRCh38

View Full Product Details