Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1041696-1042228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA7
Entrez Gene ID
10347
Gene Name
ATP binding cassette subfamily A member 7
Gene Aliases
ABCA-SSN, ABCX, AD9
UniGene
Hs.134514
Gene Chromosome Location
Chr.19: 1040103-1065572 on Build GRCh38
Transcripts
NM_019112.3 XM_006722616.1 XM_006722617.2 XM_011527628.2 XM_011527629.1 XM_011527630.1 XM_011527631.1 XM_011527632.1 XM_011527633.2 XM_011527634.1 XM_011527635.1 XM_017026142.1 XM_017026143.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3388538 c.325G>T p.A109S Chr.19: 1042086-1042086 on Build GRCh38
6346606 c.376G>T p.G126W Chr.19: 1042137-1042137 on Build GRCh38
7353508 c.358A>T p.R120W Chr.19: 1042119-1042119 on Build GRCh38
5013337 c.275G>C p.G92A Chr.19: 1041945-1041945 on Build GRCh38
4427443 c.414G>A p.T138T Chr.19: 1042175-1042175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758503836 Chr.19: 1041924-1041924 on Build GRCh38
rs374346281 Chr.19: 1042060-1042060 on Build GRCh38
rs375082305 Chr.19: 1041866-1041866 on Build GRCh38
rs199776416 Chr.19: 1041844-1041844 on Build GRCh38
rs368255900 Chr.19: 1042075-1042075 on Build GRCh38
rs747692633 Chr.19: 1042043-1042043 on Build GRCh38
rs764668443 Chr.19: 1041814-1041814 on Build GRCh38
rs775154986 Chr.19: 1041918-1041918 on Build GRCh38
rs529378024 Chr.19: 1042071-1042071 on Build GRCh38
rs750496889 Chr.19: 1042181-1042181 on Build GRCh38
rs757154663 Chr.19: 1041956-1041956 on Build GRCh38
rs770647704 Chr.19: 1041859-1041859 on Build GRCh38
rs746400634 Chr.19: 1042112-1042112 on Build GRCh38
rs766043989 Chr.19: 1041808-1041808 on Build GRCh38
rs951266275 Chr.19: 1042021-1042021 on Build GRCh38
rs776280257 Chr.19: 1041861-1041861 on Build GRCh38
rs760077861 Chr.19: 1042056-1042056 on Build GRCh38
rs200059476 Chr.19: 1042037-1042037 on Build GRCh38
rs960827398 Chr.19: 1041921-1041921 on Build GRCh38
rs775890967 Chr.19: 1042065-1042065 on Build GRCh38
rs745685434 Chr.19: 1041963-1041963 on Build GRCh38
rs948732082 Chr.19: 1041780-1041780 on Build GRCh38
rs767018908 Chr.19: 1041995-1041995 on Build GRCh38
rs541206453 Chr.19: 1041955-1041955 on Build GRCh38
rs759984255 Chr.19: 1041999-1041999 on Build GRCh38
rs557727395 Chr.19: 1041812-1041812 on Build GRCh38
rs758423495 Chr.19: 1041947-1041947 on Build GRCh38
rs761317340 Chr.19: 1041991-1041991 on Build GRCh38
rs775808026 Chr.19: 1042148-1042148 on Build GRCh38
rs148580487 Chr.19: 1041872-1041872 on Build GRCh38
rs3764644 Chr.19: 1041853-1041853 on Build GRCh38
rs767649579 Chr.19: 1042175-1042175 on Build GRCh38
rs547528278 Chr.19: 1042163-1042163 on Build GRCh38
rs772151243 Chr.19: 1041880-1041880 on Build GRCh38
rs748203620 Chr.19: 1041875-1041875 on Build GRCh38
rs755349521 Chr.19: 1041944-1041944 on Build GRCh38
rs767354067 Chr.19: 1041969-1041969 on Build GRCh38
rs533584461 Chr.19: 1042153-1042153 on Build GRCh38
rs756548806 Chr.19: 1041868-1041868 on Build GRCh38
rs1048417153 Chr.19: 1041802-1041802 on Build GRCh38
rs200578438 Chr.19: 1041964-1041964 on Build GRCh38
rs762431840 Chr.19: 1041864-1041864 on Build GRCh38
rs764118325 Chr.19: 1042012-1042012 on Build GRCh38
rs144546979 Chr.19: 1041910-1041910 on Build GRCh38
rs780505999 Chr.19: 1041869-1041869 on Build GRCh38
rs547618420 Chr.19: 1041763-1041763 on Build GRCh38
rs753651975 Chr.19: 1042193-1042193 on Build GRCh38
rs778713340 Chr.19: 1042086-1042086 on Build GRCh38
rs763590231 Chr.19: 1041930-1041930 on Build GRCh38
rs746192296 Chr.19: 1041823-1041823 on Build GRCh38
rs966435425 Chr.19: 1041877-1041877 on Build GRCh38
rs754689810 Chr.19: 1042085-1042085 on Build GRCh38
rs752468393 Chr.19: 1042196-1042196 on Build GRCh38
rs370037870 Chr.19: 1041809-1041809 on Build GRCh38
rs200149451 Chr.19: 1041968-1041968 on Build GRCh38
rs551756997 Chr.19: 1042188-1042188 on Build GRCh38
rs753690015 Chr.19: 1042082-1042082 on Build GRCh38
rs939257344 Chr.19: 1041758-1041758 on Build GRCh38
rs781014241 Chr.19: 1041962-1041962 on Build GRCh38
rs765580385 Chr.19: 1042001-1042001 on Build GRCh38
rs768917670 Chr.19: 1041829-1041829 on Build GRCh38
rs773153252 Chr.19: 1041854-1041854 on Build GRCh38
rs771645047 Chr.19: 1042049-1042049 on Build GRCh38
rs368794748 Chr.19: 1041998-1041998 on Build GRCh38
rs370464878 Chr.19: 1041936-1041936 on Build GRCh38
rs760419954 Chr.19: 1041858-1041858 on Build GRCh38
rs918490906 Chr.19: 1041767-1041767 on Build GRCh38
rs768107424 Chr.19: 1041983-1041983 on Build GRCh38
rs767527015 Chr.19: 1042073-1042073 on Build GRCh38
rs777420995 Chr.19: 1042105-1042105 on Build GRCh38
rs942424899 Chr.19: 1041851-1041851 on Build GRCh38
rs781650490 Chr.19: 1041821-1041821 on Build GRCh38
rs927813382 Chr.19: 1041724-1041724 on Build GRCh38
rs746178935 Chr.19: 1042187-1042188 on Build GRCh38
rs892649102 Chr.19: 1042110-1042110 on Build GRCh38
rs563604342 Chr.19: 1041976-1041976 on Build GRCh38
rs748096156 Chr.19: 1041902-1041904 on Build GRCh38
rs752117203 Chr.19: 1041863-1041863 on Build GRCh38
rs909915393 Chr.19: 1041980-1041980 on Build GRCh38
rs766893839 Chr.19: 1041938-1041938 on Build GRCh38
rs776869916 Chr.19: 1042188-1042188 on Build GRCh38
rs781134970 Chr.19: 1042025-1042025 on Build GRCh38
rs377401443 Chr.19: 1041906-1041906 on Build GRCh38
rs1023492673 Chr.19: 1042119-1042119 on Build GRCh38
rs999238254 Chr.19: 1042088-1042088 on Build GRCh38
rs779882754 Chr.19: 1042030-1042030 on Build GRCh38
rs756495449 Chr.19: 1041826-1041826 on Build GRCh38
rs370701255 Chr.19: 1041862-1041862 on Build GRCh38
rs754236507 Chr.19: 1041943-1041943 on Build GRCh38
rs755872121 Chr.19: 1042029-1042029 on Build GRCh38
rs1012491588 Chr.19: 1042111-1042111 on Build GRCh38
rs199601811 Chr.19: 1041827-1041827 on Build GRCh38
rs762398042 Chr.19: 1041929-1041929 on Build GRCh38
rs772854996 Chr.19: 1041787-1041787 on Build GRCh38
rs998408745 Chr.19: 1041856-1041856 on Build GRCh38
rs746893001 Chr.19: 1041903-1041903 on Build GRCh38
rs778842151 Chr.19: 1042194-1042194 on Build GRCh38
rs771769452 Chr.19: 1042100-1042100 on Build GRCh38
rs567043891 Chr.19: 1041716-1041716 on Build GRCh38
rs768333996 Chr.19: 1042035-1042035 on Build GRCh38
rs779101555 Chr.19: 1041874-1041874 on Build GRCh38
rs771093319 Chr.19: 1042175-1042175 on Build GRCh38
rs981273963 Chr.19: 1041722-1041723 on Build GRCh38
rs760429218 Chr.19: 1041795-1041795 on Build GRCh38
rs767927767 Chr.19: 1041865-1041865 on Build GRCh38
rs770309608 Chr.19: 1042116-1042116 on Build GRCh38
rs1042231840 Chr.19: 1041886-1041886 on Build GRCh38
rs777874744 Chr.19: 1041941-1041941 on Build GRCh38
rs752922595 Chr.19: 1042006-1042006 on Build GRCh38
rs777294102 Chr.19: 1042050-1042050 on Build GRCh38
rs777577175 Chr.19: 1041891-1041891 on Build GRCh38
rs200999317 Chr.19: 1042010-1042010 on Build GRCh38
rs757699680 Chr.19: 1041819-1041819 on Build GRCh38
rs773970422 Chr.19: 1041987-1041987 on Build GRCh38
rs767415887 Chr.19: 1041785-1041785 on Build GRCh38
rs776405544 Chr.19: 1041907-1041907 on Build GRCh38
rs952004729 Chr.19: 1041957-1041957 on Build GRCh38
rs774569523 Chr.19: 1042154-1042154 on Build GRCh38
rs946915097 Chr.19: 1042155-1042155 on Build GRCh38
rs760624532 Chr.19: 1042183-1042183 on Build GRCh38
rs566014616 Chr.19: 1041776-1041776 on Build GRCh38
rs754183203 Chr.19: 1041870-1041870 on Build GRCh38
rs759225155 Chr.19: 1041909-1041909 on Build GRCh38
rs1044020461 Chr.19: 1042174-1042174 on Build GRCh38
rs146597357 Chr.19: 1041923-1041923 on Build GRCh38
rs867809034 Chr.19: 1041882-1041882 on Build GRCh38
rs374854257 Chr.19: 1042087-1042087 on Build GRCh38
rs201665195 Chr.19: 1041972-1041972 on Build GRCh38
rs201963646 Chr.19: 1042072-1042072 on Build GRCh38
rs371660184 Chr.19: 1042076-1042076 on Build GRCh38
rs763130547 Chr.19: 1042069-1042069 on Build GRCh38
rs751125068 Chr.19: 1041764-1041764 on Build GRCh38
rs565975503 Chr.19: 1042089-1042089 on Build GRCh38
rs573812498 Chr.19: 1041946-1041946 on Build GRCh38
rs532833882 Chr.19: 1042048-1042049 on Build GRCh38
rs372717654 Chr.19: 1042033-1042033 on Build GRCh38
rs866355955 Chr.19: 1042090-1042090 on Build GRCh38
rs140142327 Chr.19: 1041931-1041931 on Build GRCh38
rs951950929 Chr.19: 1042143-1042143 on Build GRCh38
rs1035192467 Chr.19: 1042104-1042104 on Build GRCh38

View Full Product Details