Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3196031-3196533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCLN
Entrez Gene ID
56926
Gene Name
nicalin
Gene Aliases
NET59
UniGene
Hs.657032
Gene Chromosome Location
Chr.19: 3185877-3209575 on Build GRCh38
Transcripts
NM_001321463.1 NM_020170.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7185708 c.547G>T p.G183C Chr.19: 3196209-3196209 on Build GRCh38
3822642 c.547G>A p.G183S Chr.19: 3196209-3196209 on Build GRCh38
3388838 c.534G>A p.T178T Chr.19: 3196196-3196196 on Build GRCh38
6465662 c.546C>T p.N182N Chr.19: 3196208-3196208 on Build GRCh38
4879687 c.608G>C p.S203T Chr.19: 3196270-3196270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374520752 Chr.19: 3196302-3196302 on Build GRCh38
rs376875761 Chr.19: 3196429-3196429 on Build GRCh38
rs745739355 Chr.19: 3196207-3196207 on Build GRCh38
rs777553397 Chr.19: 3196191-3196191 on Build GRCh38
rs931957021 Chr.19: 3196389-3196389 on Build GRCh38
rs940534049 Chr.19: 3196404-3196404 on Build GRCh38
rs1011176772 Chr.19: 3196447-3196447 on Build GRCh38
rs928033342 Chr.19: 3196475-3196475 on Build GRCh38
rs944339869 Chr.19: 3196125-3196125 on Build GRCh38
rs766782078 Chr.19: 3196289-3196289 on Build GRCh38
rs771930189 Chr.19: 3196209-3196209 on Build GRCh38
rs940696547 Chr.19: 3196187-3196187 on Build GRCh38
rs768936465 Chr.19: 3196260-3196260 on Build GRCh38
rs866017397 Chr.19: 3196197-3196197 on Build GRCh38
rs916768171 Chr.19: 3196320-3196320 on Build GRCh38
rs975748467 Chr.19: 3196349-3196349 on Build GRCh38
rs977755186 Chr.19: 3196296-3196296 on Build GRCh38
rs756327446 Chr.19: 3196061-3196061 on Build GRCh38
rs779305762 Chr.19: 3196261-3196261 on Build GRCh38
rs931251735 Chr.19: 3196348-3196348 on Build GRCh38
rs311615 Chr.19: 3196124-3196124 on Build GRCh38
rs751925198 Chr.19: 3196293-3196293 on Build GRCh38
rs1007158378 Chr.19: 3196354-3196354 on Build GRCh38
rs978330152 Chr.19: 3196073-3196073 on Build GRCh38
rs1036133119 Chr.19: 3196416-3196418 on Build GRCh38
rs918470829 Chr.19: 3196345-3196345 on Build GRCh38
rs935797206 Chr.19: 3196106-3196106 on Build GRCh38
rs200886519 Chr.19: 3196272-3196272 on Build GRCh38
rs570426727 Chr.19: 3196313-3196313 on Build GRCh38
rs1023554111 Chr.19: 3196248-3196248 on Build GRCh38
rs377186463 Chr.19: 3196474-3196474 on Build GRCh38
rs577883580 Chr.19: 3196453-3196453 on Build GRCh38
rs1040756196 Chr.19: 3196369-3196369 on Build GRCh38
rs749352174 Chr.19: 3196171-3196171 on Build GRCh38
rs199988862 Chr.19: 3196299-3196299 on Build GRCh38
rs537570606 Chr.19: 3196350-3196350 on Build GRCh38
rs948170502 Chr.19: 3196108-3196108 on Build GRCh38
rs776320101 Chr.19: 3196489-3196489 on Build GRCh38
rs797019288 Chr.19: 3196173-3196173 on Build GRCh38
rs755122974 Chr.19: 3196134-3196135 on Build GRCh38
rs555887858 Chr.19: 3196393-3196393 on Build GRCh38
rs3826932 Chr.19: 3196441-3196441 on Build GRCh38
rs201578716 Chr.19: 3196310-3196310 on Build GRCh38
rs760507228 Chr.19: 3196230-3196230 on Build GRCh38
rs924218387 Chr.19: 3196074-3196074 on Build GRCh38
rs940666862 Chr.19: 3196343-3196343 on Build GRCh38
rs1030626920 Chr.19: 3196430-3196430 on Build GRCh38
rs570135289 Chr.19: 3196339-3196339 on Build GRCh38
rs778242409 Chr.19: 3196318-3196318 on Build GRCh38
rs1031204826 Chr.19: 3196304-3196304 on Build GRCh38
rs948112593 Chr.19: 3196332-3196332 on Build GRCh38
rs920604825 Chr.19: 3196509-3196509 on Build GRCh38
rs187629347 Chr.19: 3196286-3196286 on Build GRCh38
rs559607078 Chr.19: 3196080-3196080 on Build GRCh38
rs1036405525 Chr.19: 3196344-3196344 on Build GRCh38
rs990021566 Chr.19: 3196319-3196319 on Build GRCh38
rs772476844 Chr.19: 3196508-3196508 on Build GRCh38
rs931846892 Chr.19: 3196163-3196163 on Build GRCh38
rs577813108 Chr.19: 3196091-3196091 on Build GRCh38
rs1006652528 Chr.19: 3196280-3196280 on Build GRCh38
rs755772538 Chr.19: 3196177-3196177 on Build GRCh38
rs7256000 Chr.19: 3196417-3196417 on Build GRCh38
rs969571102 Chr.19: 3196456-3196456 on Build GRCh38
rs999144388 Chr.19: 3196420-3196420 on Build GRCh38
rs1040032763 Chr.19: 3196138-3196138 on Build GRCh38
rs1049433296 Chr.19: 3196395-3196395 on Build GRCh38
rs563302059 Chr.19: 3196193-3196193 on Build GRCh38
rs975827084 Chr.19: 3196507-3196507 on Build GRCh38
rs909076586 Chr.19: 3196403-3196403 on Build GRCh38
rs528069942 Chr.19: 3196295-3196295 on Build GRCh38
rs889643503 Chr.19: 3196353-3196353 on Build GRCh38
rs1024332656 Chr.19: 3196451-3196451 on Build GRCh38
rs775077801 Chr.19: 3196229-3196229 on Build GRCh38
rs897110852 Chr.19: 3196205-3196205 on Build GRCh38
rs889497107 Chr.19: 3196279-3196279 on Build GRCh38
rs748872728 Chr.19: 3196192-3196192 on Build GRCh38
rs1001146217 Chr.19: 3196466-3196466 on Build GRCh38
rs965793375 Chr.19: 3196503-3196503 on Build GRCh38
rs59113663 Chr.19: 3196171-3196171 on Build GRCh38
rs765252185 Chr.19: 3196277-3196277 on Build GRCh38
rs757826815 Chr.19: 3196208-3196208 on Build GRCh38
rs192640034 Chr.19: 3196333-3196333 on Build GRCh38
rs373106167 Chr.19: 3196265-3196265 on Build GRCh38
rs570882624 Chr.19: 3196195-3196195 on Build GRCh38
rs1019754721 Chr.19: 3196494-3196494 on Build GRCh38
rs900437465 Chr.19: 3196144-3196144 on Build GRCh38
rs574140305 Chr.19: 3196394-3196394 on Build GRCh38
rs989698199 Chr.19: 3196088-3196088 on Build GRCh38
rs745821355 Chr.19: 3196328-3196328 on Build GRCh38
rs1045077051 Chr.19: 3196442-3196442 on Build GRCh38
rs113297795 Chr.19: 3196287-3196287 on Build GRCh38
rs1046800080 Chr.19: 3196110-3196110 on Build GRCh38

View Full Product Details