Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11832833-11833333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF440
Entrez Gene ID
126070
Gene Name
zinc finger protein 440
Gene Aliases
-
UniGene
Hs.418192
Gene Chromosome Location
Chr.19: 11814241-11835617 on Build GRCh38
Transcripts
NM_152357.2 XM_005259731.3 XM_017026253.1 XM_017026254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6830245 c.1680C>T p.Y560Y Chr.19: 11832856-11832856 on Build GRCh38
4743889 c.1748C>T p.P583L Chr.19: 11832924-11832924 on Build GRCh38
1271406 c.1678T>G p.Y560D Chr.19: 11832854-11832854 on Build GRCh38
3528640 c.1735C>T p.P579S Chr.19: 11832911-11832911 on Build GRCh38
286865 c.1740G>T p.S580S Chr.19: 11832916-11832916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748711346 Chr.19: 11832948-11832948 on Build GRCh38
rs868023420 Chr.19: 11832890-11832890 on Build GRCh38
rs749404632 Chr.19: 11832973-11832973 on Build GRCh38
rs892055262 Chr.19: 11833280-11833280 on Build GRCh38
rs1053913510 Chr.19: 11833214-11833214 on Build GRCh38
rs909815985 Chr.19: 11833119-11833119 on Build GRCh38
rs400106 Chr.19: 11832882-11832882 on Build GRCh38
rs1046562657 Chr.19: 11833165-11833165 on Build GRCh38
rs569145503 Chr.19: 11832946-11832946 on Build GRCh38
rs199519108 Chr.19: 11833299-11833299 on Build GRCh38
rs1033162799 Chr.19: 11832967-11832967 on Build GRCh38
rs1022678764 Chr.19: 11832931-11832931 on Build GRCh38
rs1033649925 Chr.19: 11833272-11833272 on Build GRCh38
rs1036870259 Chr.19: 11833187-11833187 on Build GRCh38
rs921959667 Chr.19: 11833116-11833116 on Build GRCh38
rs535783306 Chr.19: 11832892-11832892 on Build GRCh38
rs767467656 Chr.19: 11832902-11832902 on Build GRCh38
rs533414503 Chr.19: 11833244-11833244 on Build GRCh38
rs381973 Chr.19: 11833255-11833255 on Build GRCh38
rs895567789 Chr.19: 11833202-11833202 on Build GRCh38
rs766364154 Chr.19: 11832894-11832894 on Build GRCh38
rs375461820 Chr.19: 11833083-11833083 on Build GRCh38
rs201851810 Chr.19: 11832992-11832992 on Build GRCh38
rs373809305 Chr.19: 11832887-11832887 on Build GRCh38
rs200800492 Chr.19: 11832947-11832947 on Build GRCh38
rs3745669 Chr.19: 11833037-11833037 on Build GRCh38
rs893778295 Chr.19: 11833235-11833235 on Build GRCh38
rs745403018 Chr.19: 11832934-11832934 on Build GRCh38
rs368705646 Chr.19: 11833010-11833010 on Build GRCh38
rs879060047 Chr.19: 11833098-11833098 on Build GRCh38
rs375049 Chr.19: 11833008-11833008 on Build GRCh38
rs56167415 Chr.19: 11832898-11832898 on Build GRCh38
rs753423614 Chr.19: 11832916-11832916 on Build GRCh38
rs866633329 Chr.19: 11832875-11832875 on Build GRCh38
rs143895808 Chr.19: 11832878-11832878 on Build GRCh38
rs773282065 Chr.19: 11832891-11832891 on Build GRCh38
rs761211969 Chr.19: 11832878-11832879 on Build GRCh38
rs956379806 Chr.19: 11832988-11832988 on Build GRCh38
rs778225720 Chr.19: 11832929-11832929 on Build GRCh38
rs146601063 Chr.19: 11833004-11833004 on Build GRCh38
rs902689778 Chr.19: 11833201-11833201 on Build GRCh38
rs769343003 Chr.19: 11832917-11832918 on Build GRCh38
rs199933676 Chr.19: 11832950-11832950 on Build GRCh38
rs941730543 Chr.19: 11833042-11833042 on Build GRCh38
rs770269280 Chr.19: 11832968-11832968 on Build GRCh38
rs1006919162 Chr.19: 11833240-11833240 on Build GRCh38
rs939803309 Chr.19: 11833149-11833149 on Build GRCh38
rs770320447 Chr.19: 11832889-11832889 on Build GRCh38
rs752451455 Chr.19: 11832908-11832908 on Build GRCh38
rs566642234 Chr.19: 11832859-11832859 on Build GRCh38
rs763396819 Chr.19: 11833208-11833208 on Build GRCh38
rs430965 Chr.19: 11833251-11833251 on Build GRCh38
rs763928293 Chr.19: 11832915-11832915 on Build GRCh38
rs1030114927 Chr.19: 11833303-11833303 on Build GRCh38
rs755715970 Chr.19: 11832860-11832860 on Build GRCh38
rs755875761 Chr.19: 11832914-11832914 on Build GRCh38
rs763792937 Chr.19: 11833011-11833011 on Build GRCh38
rs527740574 Chr.19: 11832872-11832872 on Build GRCh38
rs771198546 Chr.19: 11832974-11832974 on Build GRCh38
rs573543742 Chr.19: 11833029-11833029 on Build GRCh38
rs974711512 Chr.19: 11833066-11833066 on Build GRCh38
rs1044055201 Chr.19: 11833223-11833223 on Build GRCh38
rs756909349 Chr.19: 11833118-11833118 on Build GRCh38
rs749012237 Chr.19: 11832871-11832871 on Build GRCh38
rs777437358 Chr.19: 11832866-11832866 on Build GRCh38
rs769295663 Chr.19: 11832880-11832880 on Build GRCh38
rs755010747 Chr.19: 11832857-11832857 on Build GRCh38
rs555345914 Chr.19: 11832935-11832935 on Build GRCh38
rs773693139 Chr.19: 11832969-11832969 on Build GRCh38
rs141420596 Chr.19: 11833117-11833117 on Build GRCh38
rs527833553 Chr.19: 11833306-11833306 on Build GRCh38

View Full Product Details