Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44308226-44308722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERI3
Entrez Gene ID
79033
Gene Name
ERI1 exoribonuclease family member 3
Gene Aliases
PINT1, PRNPIP
UniGene
Hs.731413
Gene Chromosome Location
Chr.1: 44221070-44355298 on Build GRCh38
Transcripts
NM_001301698.1 NM_001301699.1 NM_001301700.1 NM_001301701.1 NM_024066.2 XM_005271186.2 XM_006710891.3 XM_017002301.1 XM_017002302.1 XM_017002303.1 XM_017002304.1 XM_017002305.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3700743 c.184-2A>G p.? Chr.1: 44308403-44308403 on Build GRCh38
6656137 c.200T>C p.M67T Chr.1: 44308385-44308385 on Build GRCh38
5381078 c.358G>A p.D120N Chr.1: 44308365-44308365 on Build GRCh38
5420965 c.328G>A p.D110N Chr.1: 44308395-44308395 on Build GRCh38
6656132 c.711C>T p.N237N Chr.1: 44308357-44308357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760149625 Chr.1: 44308427-44308427 on Build GRCh38
rs1033037111 Chr.1: 44308649-44308649 on Build GRCh38
rs567579435 Chr.1: 44308511-44308511 on Build GRCh38
rs369748957 Chr.1: 44308381-44308381 on Build GRCh38
rs975804195 Chr.1: 44308256-44308256 on Build GRCh38
rs953203557 Chr.1: 44308633-44308633 on Build GRCh38
rs371849189 Chr.1: 44308357-44308357 on Build GRCh38
rs775296091 Chr.1: 44308408-44308408 on Build GRCh38
rs761450076 Chr.1: 44308434-44308437 on Build GRCh38
rs761457284 Chr.1: 44308416-44308416 on Build GRCh38
rs1014786934 Chr.1: 44308612-44308612 on Build GRCh38
rs752924404 Chr.1: 44308363-44308363 on Build GRCh38
rs200228759 Chr.1: 44308372-44308372 on Build GRCh38
rs755526499 Chr.1: 44308442-44308442 on Build GRCh38
rs758745548 Chr.1: 44308623-44308623 on Build GRCh38
rs776470793 Chr.1: 44308297-44308297 on Build GRCh38
rs199612153 Chr.1: 44308414-44308414 on Build GRCh38
rs941533380 Chr.1: 44308257-44308257 on Build GRCh38
rs931325495 Chr.1: 44308465-44308465 on Build GRCh38
rs751866284 Chr.1: 44308356-44308356 on Build GRCh38
rs112036613 Chr.1: 44308379-44308379 on Build GRCh38
rs141832971 Chr.1: 44308382-44308382 on Build GRCh38
rs41301062 Chr.1: 44308611-44308611 on Build GRCh38
rs777861206 Chr.1: 44308569-44308569 on Build GRCh38
rs990740072 Chr.1: 44308577-44308577 on Build GRCh38
rs887685205 Chr.1: 44308539-44308539 on Build GRCh38
rs372445345 Chr.1: 44308442-44308442 on Build GRCh38
rs752048711 Chr.1: 44308425-44308425 on Build GRCh38
rs146333790 Chr.1: 44308281-44308281 on Build GRCh38
rs1028703439 Chr.1: 44308628-44308628 on Build GRCh38
rs377092061 Chr.1: 44308368-44308368 on Build GRCh38
rs763558651 Chr.1: 44308270-44308270 on Build GRCh38
rs766785716 Chr.1: 44308417-44308417 on Build GRCh38
rs960701149 Chr.1: 44308550-44308550 on Build GRCh38
rs918062128 Chr.1: 44308398-44308398 on Build GRCh38
rs769648101 Chr.1: 44308434-44308435 on Build GRCh38
rs755558422 Chr.1: 44308530-44308530 on Build GRCh38
rs375750681 Chr.1: 44308268-44308268 on Build GRCh38
rs538618890 Chr.1: 44308648-44308648 on Build GRCh38
rs569597286 Chr.1: 44308624-44308624 on Build GRCh38
rs771790313 Chr.1: 44308406-44308406 on Build GRCh38
rs556336721 Chr.1: 44308435-44308436 on Build GRCh38
rs551631828 Chr.1: 44308294-44308294 on Build GRCh38
rs753414076 Chr.1: 44308278-44308278 on Build GRCh38
rs1004745564 Chr.1: 44308568-44308568 on Build GRCh38
rs956995731 Chr.1: 44308652-44308652 on Build GRCh38
rs3853366 Chr.1: 44308615-44308615 on Build GRCh38
rs115939678 Chr.1: 44308667-44308667 on Build GRCh38
rs767941156 Chr.1: 44308439-44308439 on Build GRCh38
rs966549115 Chr.1: 44308523-44308523 on Build GRCh38
rs775977494 Chr.1: 44308415-44308415 on Build GRCh38
rs781691741 Chr.1: 44308358-44308358 on Build GRCh38
rs538440708 Chr.1: 44308435-44308435 on Build GRCh38
rs918091939 Chr.1: 44308691-44308691 on Build GRCh38
rs778966331 Chr.1: 44308395-44308395 on Build GRCh38
rs1045093105 Chr.1: 44308433-44308433 on Build GRCh38
rs949514434 Chr.1: 44308697-44308697 on Build GRCh38
rs41311203 Chr.1: 44308396-44308396 on Build GRCh38

View Full Product Details