Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44352758-44353253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERI3
Entrez Gene ID
79033
Gene Name
ERI1 exoribonuclease family member 3
Gene Aliases
PINT1, PRNPIP
UniGene
Hs.731413
Gene Chromosome Location
Chr.1: 44221070-44355298 on Build GRCh38
Transcripts
NM_001301698.1 NM_001301699.1 NM_001301700.1 NM_001301701.1 NM_024066.2 XM_005271186.2 XM_006710891.3 XM_017002301.1 XM_017002303.1 XM_017002304.1 XM_017002305.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5992864 c.162A>C p.E54D Chr.1: 44352899-44352899 on Build GRCh38
4824744 c.167C>T p.S56L Chr.1: 44352894-44352894 on Build GRCh38
910008 c.199G>A p.E67K Chr.1: 44352862-44352862 on Build GRCh38
910009 c.188T>C p.L63P Chr.1: 44352873-44352873 on Build GRCh38
7279241 c.164C>A p.P55H Chr.1: 44352897-44352897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781707948 Chr.1: 44353021-44353021 on Build GRCh38
rs931348879 Chr.1: 44353048-44353048 on Build GRCh38
rs750838094 Chr.1: 44352844-44352844 on Build GRCh38
rs538552533 Chr.1: 44352902-44352902 on Build GRCh38
rs80152700 Chr.1: 44352963-44352963 on Build GRCh38
rs536800762 Chr.1: 44353007-44353007 on Build GRCh38
rs751794673 Chr.1: 44352935-44352935 on Build GRCh38
rs549669153 Chr.1: 44352782-44352782 on Build GRCh38
rs930122937 Chr.1: 44353114-44353114 on Build GRCh38
rs561351481 Chr.1: 44352870-44352870 on Build GRCh38
rs753164815 Chr.1: 44352877-44352877 on Build GRCh38
rs778018171 Chr.1: 44352801-44352801 on Build GRCh38
rs997737327 Chr.1: 44353180-44353180 on Build GRCh38
rs941658403 Chr.1: 44353041-44353041 on Build GRCh38
rs901703485 Chr.1: 44353172-44353172 on Build GRCh38
rs749182772 Chr.1: 44352966-44352966 on Build GRCh38
rs747247749 Chr.1: 44352857-44352857 on Build GRCh38
rs200153192 Chr.1: 44352969-44352969 on Build GRCh38
rs777706147 Chr.1: 44352965-44352965 on Build GRCh38
rs115845110 Chr.1: 44353131-44353131 on Build GRCh38
rs574286587 Chr.1: 44353002-44353002 on Build GRCh38
rs376914053 Chr.1: 44352905-44352905 on Build GRCh38
rs780571393 Chr.1: 44352926-44352926 on Build GRCh38
rs900159966 Chr.1: 44353186-44353186 on Build GRCh38
rs1004820233 Chr.1: 44353144-44353144 on Build GRCh38
rs770640386 Chr.1: 44352896-44352896 on Build GRCh38
rs140027583 Chr.1: 44352913-44352913 on Build GRCh38
rs752251292 Chr.1: 44352912-44352912 on Build GRCh38
rs748054752 Chr.1: 44352884-44352884 on Build GRCh38
rs781401406 Chr.1: 44352956-44352956 on Build GRCh38
rs772875297 Chr.1: 44352893-44352893 on Build GRCh38
rs780384183 Chr.1: 44352853-44352853 on Build GRCh38
rs201922019 Chr.1: 44352894-44352894 on Build GRCh38
rs1048389671 Chr.1: 44353071-44353071 on Build GRCh38
rs576721832 Chr.1: 44353077-44353077 on Build GRCh38
rs916548111 Chr.1: 44353033-44353033 on Build GRCh38
rs990863141 Chr.1: 44353032-44353032 on Build GRCh38
rs1028784059 Chr.1: 44353203-44353203 on Build GRCh38
rs778160132 Chr.1: 44352993-44352995 on Build GRCh38
rs989801102 Chr.1: 44352911-44352911 on Build GRCh38
rs200579579 Chr.1: 44352838-44352838 on Build GRCh38
rs773922846 Chr.1: 44352900-44352900 on Build GRCh38
rs1045178379 Chr.1: 44353011-44353011 on Build GRCh38
rs754302539 Chr.1: 44352836-44352836 on Build GRCh38
rs918760711 Chr.1: 44353075-44353075 on Build GRCh38
rs769821211 Chr.1: 44352887-44352887 on Build GRCh38
rs953098531 Chr.1: 44353221-44353221 on Build GRCh38
rs368985174 Chr.1: 44353146-44353146 on Build GRCh38
rs767915766 Chr.1: 44352811-44352811 on Build GRCh38
rs1025983088 Chr.1: 44352874-44352874 on Build GRCh38
rs941593341 Chr.1: 44352784-44352784 on Build GRCh38
rs1028731943 Chr.1: 44352863-44352863 on Build GRCh38
rs781456588 Chr.1: 44352878-44352878 on Build GRCh38
rs193206896 Chr.1: 44352916-44352916 on Build GRCh38
rs966499114 Chr.1: 44352941-44352941 on Build GRCh38
rs953046034 Chr.1: 44352897-44352897 on Build GRCh38
rs779123247 Chr.1: 44352839-44352839 on Build GRCh38
rs766119420 Chr.1: 44352823-44352823 on Build GRCh38
rs1005173148 Chr.1: 44352841-44352841 on Build GRCh38
rs1022181290 Chr.1: 44353231-44353231 on Build GRCh38
rs367944765 Chr.1: 44352923-44352923 on Build GRCh38
rs371633227 Chr.1: 44352830-44352830 on Build GRCh38
rs751599752 Chr.1: 44352954-44352954 on Build GRCh38
rs751054368 Chr.1: 44352828-44352828 on Build GRCh38
rs185205028 Chr.1: 44353026-44353026 on Build GRCh38
rs1047484836 Chr.1: 44353129-44353129 on Build GRCh38
rs767016022 Chr.1: 44352834-44352834 on Build GRCh38
rs758546359 Chr.1: 44352850-44352850 on Build GRCh38

View Full Product Details