Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16574766-16575308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED26
Entrez Gene ID
9441
Gene Name
mediator complex subunit 26
Gene Aliases
CRSP7, CRSP70
UniGene
Hs.715040
Gene Chromosome Location
Chr.19: 16574907-16628204 on Build GRCh38
Transcripts
NM_004831.3

SNP Detail

SNP ID Chromosome Location
rs962582791 Chr.19: 16574789-16574789 on Build GRCh38
rs59332250 Chr.19: 16575165-16575165 on Build GRCh38
rs956632188 Chr.19: 16574862-16574862 on Build GRCh38
rs946906065 Chr.19: 16575019-16575019 on Build GRCh38
rs59017601 Chr.19: 16575168-16575168 on Build GRCh38
rs1047223011 Chr.19: 16574931-16574931 on Build GRCh38
rs1039805920 Chr.19: 16575042-16575042 on Build GRCh38
rs531294418 Chr.19: 16575104-16575104 on Build GRCh38
rs1000549961 Chr.19: 16575224-16575224 on Build GRCh38
rs546239518 Chr.19: 16575142-16575143 on Build GRCh38
rs756811341 Chr.19: 16574879-16574879 on Build GRCh38
rs969275442 Chr.19: 16575272-16575272 on Build GRCh38
rs7248857 Chr.19: 16574827-16574827 on Build GRCh38
rs59479853 Chr.19: 16575154-16575154 on Build GRCh38
rs931394212 Chr.19: 16575147-16575147 on Build GRCh38
rs59759095 Chr.19: 16575146-16575146 on Build GRCh38
rs971032923 Chr.19: 16574842-16574842 on Build GRCh38
rs3210375 Chr.19: 16574942-16574942 on Build GRCh38
rs991972529 Chr.19: 16575084-16575084 on Build GRCh38
rs917846117 Chr.19: 16574894-16574894 on Build GRCh38
rs903546950 Chr.19: 16575223-16575223 on Build GRCh38
rs79266930 Chr.19: 16575128-16575128 on Build GRCh38
rs78091837 Chr.19: 16574920-16574920 on Build GRCh38
rs988422293 Chr.19: 16574926-16574926 on Build GRCh38
rs1038294288 Chr.19: 16575144-16575144 on Build GRCh38
rs763013237 Chr.19: 16575181-16575181 on Build GRCh38
rs751576995 Chr.19: 16575094-16575094 on Build GRCh38
rs748257733 Chr.19: 16575216-16575226 on Build GRCh38
rs1026972461 Chr.19: 16575281-16575282 on Build GRCh38
rs546131177 Chr.19: 16575183-16575183 on Build GRCh38
rs1041287775 Chr.19: 16574962-16574962 on Build GRCh38
rs1041236619 Chr.19: 16575195-16575195 on Build GRCh38
rs915360250 Chr.19: 16574927-16574927 on Build GRCh38
rs139895679 Chr.19: 16575278-16575278 on Build GRCh38
rs969286803 Chr.19: 16575117-16575117 on Build GRCh38
rs928203724 Chr.19: 16574905-16574905 on Build GRCh38
rs573118350 Chr.19: 16575203-16575203 on Build GRCh38
rs570476342 Chr.19: 16574833-16574833 on Build GRCh38
rs932921839 Chr.19: 16575204-16575205 on Build GRCh38
rs1024799547 Chr.19: 16575267-16575267 on Build GRCh38
rs955739671 Chr.19: 16575235-16575235 on Build GRCh38
rs746221348 Chr.19: 16574922-16574922 on Build GRCh38
rs181522188 Chr.19: 16575078-16575078 on Build GRCh38
rs3210374 Chr.19: 16574943-16574943 on Build GRCh38
rs1045049907 Chr.19: 16574921-16574922 on Build GRCh38
rs944162565 Chr.19: 16575190-16575190 on Build GRCh38
rs58343988 Chr.19: 16574937-16574938 on Build GRCh38
rs3210376 Chr.19: 16574966-16574966 on Build GRCh38
rs1013340920 Chr.19: 16575249-16575249 on Build GRCh38
rs899951464 Chr.19: 16575049-16575049 on Build GRCh38
rs189355295 Chr.19: 16574871-16574871 on Build GRCh38
rs371580744 Chr.19: 16575186-16575186 on Build GRCh38
rs919962926 Chr.19: 16575135-16575135 on Build GRCh38
rs752505918 Chr.19: 16575151-16575151 on Build GRCh38
rs1029083192 Chr.19: 16575025-16575025 on Build GRCh38
rs576862930 Chr.19: 16575150-16575150 on Build GRCh38
rs79066018 Chr.19: 16574922-16574922 on Build GRCh38
rs1008468929 Chr.19: 16574935-16574935 on Build GRCh38
rs566731335 Chr.19: 16574932-16574932 on Build GRCh38
rs1001046533 Chr.19: 16575034-16575034 on Build GRCh38
rs992066012 Chr.19: 16574863-16574863 on Build GRCh38
rs915531965 Chr.19: 16575263-16575263 on Build GRCh38
rs1031270958 Chr.19: 16575221-16575221 on Build GRCh38
rs900103484 Chr.19: 16575002-16575002 on Build GRCh38
rs776867632 Chr.19: 16574823-16574823 on Build GRCh38
rs1022658825 Chr.19: 16575175-16575175 on Build GRCh38
rs536715301 Chr.19: 16575124-16575124 on Build GRCh38
rs185370018 Chr.19: 16575247-16575247 on Build GRCh38
rs919990325 Chr.19: 16574922-16574926 on Build GRCh38
rs935560404 Chr.19: 16574924-16574924 on Build GRCh38
rs903593863 Chr.19: 16575033-16575033 on Build GRCh38
rs184522912 Chr.19: 16574850-16574850 on Build GRCh38
rs58589693 Chr.19: 16575129-16575129 on Build GRCh38
rs991607593 Chr.19: 16574818-16574818 on Build GRCh38
rs968613941 Chr.19: 16575276-16575276 on Build GRCh38
rs199602826 Chr.19: 16574938-16574938 on Build GRCh38
rs1051119230 Chr.19: 16575066-16575066 on Build GRCh38
rs568132677 Chr.19: 16575095-16575095 on Build GRCh38
rs933965232 Chr.19: 16575064-16575064 on Build GRCh38
rs887132846 Chr.19: 16575067-16575067 on Build GRCh38
rs1024810939 Chr.19: 16575110-16575110 on Build GRCh38
rs1030822638 Chr.19: 16575051-16575051 on Build GRCh38
rs536038310 Chr.19: 16575003-16575003 on Build GRCh38

View Full Product Details