Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40035809-40036307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF780B
Entrez Gene ID
163131
Gene Name
zinc finger protein 780B
Gene Aliases
ZNF779
UniGene
Hs.101139
Gene Chromosome Location
Chr.19: 40028260-40059026 on Build GRCh38
Transcripts
NM_001005851.2 XM_005258590.4 XM_005258591.4 XM_005258592.3 XM_005258593.2 XM_005258595.3 XM_006723072.3 XM_011526593.2 XM_017026425.1 XM_017026426.1 XM_017026427.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3823099 c.770C>A p.S257Y Chr.19: 40036089-40036089 on Build GRCh38
6833070 c.972C>T p.C324C Chr.19: 40035887-40035887 on Build GRCh38
5855765 c.862C>T p.R288C Chr.19: 40035997-40035997 on Build GRCh38
4393592 c.854C>G p.A285G Chr.19: 40036005-40036005 on Build GRCh38
3712928 c.990G>A p.E330E Chr.19: 40035869-40035869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763918403 Chr.19: 40035863-40035863 on Build GRCh38
rs766182178 Chr.19: 40036080-40036080 on Build GRCh38
rs754948603 Chr.19: 40036116-40036116 on Build GRCh38
rs760045917 Chr.19: 40036031-40036031 on Build GRCh38
rs762350314 Chr.19: 40035832-40035832 on Build GRCh38
rs370154602 Chr.19: 40036158-40036158 on Build GRCh38
rs377464481 Chr.19: 40036027-40036027 on Build GRCh38
rs771298388 Chr.19: 40036028-40036028 on Build GRCh38
rs757546815 Chr.19: 40036279-40036279 on Build GRCh38
rs144556991 Chr.19: 40035996-40035996 on Build GRCh38
rs774944439 Chr.19: 40036029-40036029 on Build GRCh38
rs369807076 Chr.19: 40036130-40036130 on Build GRCh38
rs775844712 Chr.19: 40036188-40036188 on Build GRCh38
rs767390188 Chr.19: 40036254-40036254 on Build GRCh38
rs376696846 Chr.19: 40035872-40035872 on Build GRCh38
rs376606215 Chr.19: 40036249-40036249 on Build GRCh38
rs749040462 Chr.19: 40036231-40036231 on Build GRCh38
rs753991521 Chr.19: 40036275-40036275 on Build GRCh38
rs987463596 Chr.19: 40036101-40036101 on Build GRCh38
rs759277704 Chr.19: 40036252-40036252 on Build GRCh38
rs371876870 Chr.19: 40035938-40035938 on Build GRCh38
rs369747781 Chr.19: 40035987-40035987 on Build GRCh38
rs201941034 Chr.19: 40035915-40035915 on Build GRCh38
rs747613300 Chr.19: 40036220-40036220 on Build GRCh38
rs781216510 Chr.19: 40036001-40036001 on Build GRCh38
rs775416273 Chr.19: 40036267-40036267 on Build GRCh38
rs1009151445 Chr.19: 40035900-40035900 on Build GRCh38
rs778933960 Chr.19: 40035875-40035875 on Build GRCh38
rs764225003 Chr.19: 40036272-40036272 on Build GRCh38
rs774154621 Chr.19: 40036167-40036167 on Build GRCh38
rs376957040 Chr.19: 40035967-40035967 on Build GRCh38
rs761617991 Chr.19: 40036061-40036061 on Build GRCh38
rs762872931 Chr.19: 40036073-40036073 on Build GRCh38
rs767079418 Chr.19: 40035843-40035843 on Build GRCh38
rs964676339 Chr.19: 40035994-40035994 on Build GRCh38
rs755111058 Chr.19: 40035997-40035997 on Build GRCh38
rs780738928 Chr.19: 40036215-40036215 on Build GRCh38
rs770621711 Chr.19: 40036244-40036244 on Build GRCh38
rs576073360 Chr.19: 40036091-40036091 on Build GRCh38
rs920066051 Chr.19: 40036014-40036014 on Build GRCh38
rs767461225 Chr.19: 40036170-40036170 on Build GRCh38
rs752891232 Chr.19: 40036123-40036123 on Build GRCh38
rs768215511 Chr.19: 40036039-40036039 on Build GRCh38
rs752729759 Chr.19: 40036172-40036172 on Build GRCh38
rs368838666 Chr.19: 40035916-40035916 on Build GRCh38
rs760259604 Chr.19: 40035943-40035943 on Build GRCh38
rs200075214 Chr.19: 40035911-40035911 on Build GRCh38
rs375599840 Chr.19: 40036147-40036147 on Build GRCh38
rs543748486 Chr.19: 40036134-40036134 on Build GRCh38
rs781401868 Chr.19: 40036120-40036120 on Build GRCh38
rs752379669 Chr.19: 40035855-40035855 on Build GRCh38
rs954368338 Chr.19: 40035978-40035978 on Build GRCh38
rs904622646 Chr.19: 40036190-40036190 on Build GRCh38
rs763373084 Chr.19: 40036189-40036189 on Build GRCh38
rs139701679 Chr.19: 40036005-40036005 on Build GRCh38
rs754063212 Chr.19: 40036187-40036187 on Build GRCh38
rs560041637 Chr.19: 40035993-40035993 on Build GRCh38
rs548221715 Chr.19: 40035835-40035835 on Build GRCh38
rs770657619 Chr.19: 40035891-40035891 on Build GRCh38
rs866042253 Chr.19: 40036081-40036081 on Build GRCh38
rs776913240 Chr.19: 40035831-40035831 on Build GRCh38
rs765104338 Chr.19: 40036063-40036063 on Build GRCh38
rs537099679 Chr.19: 40036230-40036230 on Build GRCh38
rs776236486 Chr.19: 40035976-40035976 on Build GRCh38
rs201098392 Chr.19: 40035886-40035886 on Build GRCh38
rs868746715 Chr.19: 40036048-40036048 on Build GRCh38
rs753540683 Chr.19: 40035974-40035974 on Build GRCh38
rs951528732 Chr.19: 40036051-40036051 on Build GRCh38
rs763507014 Chr.19: 40036255-40036281 on Build GRCh38
rs949346395 Chr.19: 40036096-40036096 on Build GRCh38
rs373997141 Chr.19: 40036169-40036169 on Build GRCh38
rs565606223 Chr.19: 40036146-40036146 on Build GRCh38
rs755770118 Chr.19: 40035858-40035858 on Build GRCh38
rs765694125 Chr.19: 40035833-40035833 on Build GRCh38
rs750374848 Chr.19: 40035981-40035981 on Build GRCh38
rs745803430 Chr.19: 40035885-40035885 on Build GRCh38
rs765188418 Chr.19: 40035977-40035977 on Build GRCh38
rs1045500165 Chr.19: 40036111-40036111 on Build GRCh38
rs770128550 Chr.19: 40036006-40036006 on Build GRCh38
rs1001179564 Chr.19: 40035865-40035865 on Build GRCh38
rs774899722 Chr.19: 40035940-40035940 on Build GRCh38
rs747772580 Chr.19: 40036149-40036149 on Build GRCh38
rs747330094 Chr.19: 40035966-40035967 on Build GRCh38
rs199500205 Chr.19: 40035871-40035871 on Build GRCh38
rs777998172 Chr.19: 40036127-40036127 on Build GRCh38
rs771303773 Chr.19: 40036025-40036025 on Build GRCh38
rs537348490 Chr.19: 40035897-40035897 on Build GRCh38
rs758986121 Chr.19: 40036213-40036213 on Build GRCh38
rs756366182 Chr.19: 40036125-40036125 on Build GRCh38
rs1009230187 Chr.19: 40035931-40035931 on Build GRCh38
rs373479514 Chr.19: 40036136-40036136 on Build GRCh38
rs576119624 Chr.19: 40036201-40036201 on Build GRCh38
rs879112437 Chr.19: 40036256-40036256 on Build GRCh38
rs760680461 Chr.19: 40036269-40036269 on Build GRCh38
rs201804097 Chr.19: 40036007-40036007 on Build GRCh38
rs780056896 Chr.19: 40035890-40035890 on Build GRCh38
rs191968005 Chr.19: 40035909-40035909 on Build GRCh38
rs766203771 Chr.19: 40036165-40036165 on Build GRCh38
rs201778936 Chr.19: 40035837-40035837 on Build GRCh38
rs762703905 Chr.19: 40036127-40036127 on Build GRCh38
rs532572879 Chr.19: 40036222-40036222 on Build GRCh38
rs769532695 Chr.19: 40036157-40036157 on Build GRCh38
rs775099168 Chr.19: 40036085-40036086 on Build GRCh38
rs758460066 Chr.19: 40035983-40035983 on Build GRCh38
rs756204807 Chr.19: 40036183-40036183 on Build GRCh38
rs763507391 Chr.19: 40035933-40035933 on Build GRCh38
rs750284909 Chr.19: 40036066-40036066 on Build GRCh38
rs560949536 Chr.19: 40036053-40036053 on Build GRCh38
rs1051459424 Chr.19: 40036160-40036160 on Build GRCh38

View Full Product Details