Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43534937-43535433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF575
Entrez Gene ID
284346
Gene Name
zinc finger protein 575
Gene Aliases
-
UniGene
Hs.213534
Gene Chromosome Location
Chr.19: 43531805-43536138 on Build GRCh38
Transcripts
NM_174945.2 XM_005258783.3 XM_011526793.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5728522 c.419C>T p.T140I Chr.19: 43535368-43535368 on Build GRCh38
1394285 c.450C>T p.P150P Chr.19: 43535399-43535399 on Build GRCh38
1394284 c.354G>A p.P118P Chr.19: 43535303-43535303 on Build GRCh38
5788806 c.160C>T p.R54C Chr.19: 43535109-43535109 on Build GRCh38
6033126 c.392G>A p.R131H Chr.19: 43535341-43535341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865820549 Chr.19: 43535379-43535379 on Build GRCh38
rs370571946 Chr.19: 43535151-43535151 on Build GRCh38
rs761803419 Chr.19: 43535403-43535403 on Build GRCh38
rs967013378 Chr.19: 43535101-43535101 on Build GRCh38
rs780782039 Chr.19: 43535293-43535294 on Build GRCh38
rs766787933 Chr.19: 43535022-43535022 on Build GRCh38
rs910855145 Chr.19: 43535146-43535146 on Build GRCh38
rs868655818 Chr.19: 43535123-43535123 on Build GRCh38
rs766156324 Chr.19: 43535205-43535205 on Build GRCh38
rs762044331 Chr.19: 43535299-43535299 on Build GRCh38
rs868491577 Chr.19: 43535289-43535289 on Build GRCh38
rs754164879 Chr.19: 43535188-43535188 on Build GRCh38
rs776713649 Chr.19: 43535393-43535393 on Build GRCh38
rs139734998 Chr.19: 43535329-43535329 on Build GRCh38
rs764311504 Chr.19: 43535285-43535285 on Build GRCh38
rs777378918 Chr.19: 43535238-43535238 on Build GRCh38
rs373118540 Chr.19: 43535320-43535320 on Build GRCh38
rs768507012 Chr.19: 43535130-43535130 on Build GRCh38
rs750239924 Chr.19: 43535116-43535116 on Build GRCh38
rs780677737 Chr.19: 43535353-43535353 on Build GRCh38
rs866525238 Chr.19: 43535244-43535244 on Build GRCh38
rs772615966 Chr.19: 43535342-43535342 on Build GRCh38
rs541167010 Chr.19: 43535074-43535074 on Build GRCh38
rs372606320 Chr.19: 43535134-43535134 on Build GRCh38
rs866892041 Chr.19: 43535304-43535304 on Build GRCh38
rs767597602 Chr.19: 43535081-43535081 on Build GRCh38
rs369065915 Chr.19: 43535122-43535122 on Build GRCh38
rs758096029 Chr.19: 43535190-43535190 on Build GRCh38
rs756871332 Chr.19: 43535335-43535335 on Build GRCh38
rs1029403067 Chr.19: 43534967-43534967 on Build GRCh38
rs776880800 Chr.19: 43535293-43535293 on Build GRCh38
rs149796259 Chr.19: 43535340-43535340 on Build GRCh38
rs769906507 Chr.19: 43535327-43535334 on Build GRCh38
rs147496313 Chr.19: 43535351-43535351 on Build GRCh38
rs778145408 Chr.19: 43535104-43535104 on Build GRCh38
rs910536461 Chr.19: 43535099-43535099 on Build GRCh38
rs373456986 Chr.19: 43535120-43535120 on Build GRCh38
rs547190692 Chr.19: 43535330-43535330 on Build GRCh38
rs765391464 Chr.19: 43535301-43535301 on Build GRCh38
rs1040486624 Chr.19: 43535217-43535217 on Build GRCh38
rs201054447 Chr.19: 43535277-43535277 on Build GRCh38
rs767927673 Chr.19: 43535163-43535163 on Build GRCh38
rs1050720257 Chr.19: 43535023-43535023 on Build GRCh38
rs769365877 Chr.19: 43535145-43535145 on Build GRCh38
rs747649150 Chr.19: 43535234-43535234 on Build GRCh38
rs867493067 Chr.19: 43535220-43535220 on Build GRCh38
rs775644996 Chr.19: 43535164-43535164 on Build GRCh38
rs746622666 Chr.19: 43535089-43535090 on Build GRCh38
rs141800849 Chr.19: 43535232-43535232 on Build GRCh38
rs756350661 Chr.19: 43535100-43535100 on Build GRCh38
rs773503728 Chr.19: 43535005-43535005 on Build GRCh38
rs1041945678 Chr.19: 43535334-43535334 on Build GRCh38
rs745366645 Chr.19: 43535295-43535319 on Build GRCh38
rs566145753 Chr.19: 43535278-43535278 on Build GRCh38
rs756079055 Chr.19: 43535321-43535321 on Build GRCh38
rs772912592 Chr.19: 43535147-43535147 on Build GRCh38
rs774723566 Chr.19: 43535259-43535259 on Build GRCh38
rs760905964 Chr.19: 43535172-43535172 on Build GRCh38
rs777219264 Chr.19: 43535324-43535324 on Build GRCh38
rs769457948 Chr.19: 43534981-43534981 on Build GRCh38
rs747667451 Chr.19: 43535140-43535140 on Build GRCh38
rs746227238 Chr.19: 43535260-43535260 on Build GRCh38
rs767425208 Chr.19: 43535306-43535306 on Build GRCh38
rs770360039 Chr.19: 43535250-43535251 on Build GRCh38
rs754887004 Chr.19: 43535211-43535211 on Build GRCh38
rs747476632 Chr.19: 43535358-43535358 on Build GRCh38
rs751333442 Chr.19: 43535206-43535206 on Build GRCh38
rs141825879 Chr.19: 43535247-43535247 on Build GRCh38
rs528658004 Chr.19: 43535280-43535280 on Build GRCh38
rs759506363 Chr.19: 43535077-43535077 on Build GRCh38
rs866347256 Chr.19: 43535187-43535187 on Build GRCh38
rs769307860 Chr.19: 43535383-43535383 on Build GRCh38
rs752678334 Chr.19: 43535309-43535309 on Build GRCh38
rs375927172 Chr.19: 43535148-43535148 on Build GRCh38
rs985114697 Chr.19: 43535058-43535058 on Build GRCh38
rs374781386 Chr.19: 43535098-43535098 on Build GRCh38
rs866041359 Chr.19: 43535295-43535295 on Build GRCh38
rs535870980 Chr.19: 43535375-43535375 on Build GRCh38
rs955061850 Chr.19: 43535007-43535007 on Build GRCh38
rs982694350 Chr.19: 43535195-43535195 on Build GRCh38
rs746230424 Chr.19: 43535341-43535341 on Build GRCh38
rs774613560 Chr.19: 43535161-43535161 on Build GRCh38
rs200449716 Chr.19: 43535412-43535412 on Build GRCh38
rs150616280 Chr.19: 43535248-43535248 on Build GRCh38
rs1050994738 Chr.19: 43535273-43535273 on Build GRCh38
rs763308002 Chr.19: 43535012-43535012 on Build GRCh38
rs751302181 Chr.19: 43535302-43535302 on Build GRCh38
rs907911175 Chr.19: 43535204-43535204 on Build GRCh38
rs199846217 Chr.19: 43535121-43535121 on Build GRCh38
rs138495162 Chr.19: 43535174-43535174 on Build GRCh38

View Full Product Details