Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48615790-48616282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL18
Entrez Gene ID
6141
Gene Name
ribosomal protein L18
Gene Aliases
L18
UniGene
Hs.515517
Gene Chromosome Location
Chr.19: 48615327-48619418 on Build GRCh38
Transcripts
NM_000979.3 NM_001270490.1 NR_073022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5390248 c.396G>A p.K132K Chr.19: 48616104-48616104 on Build GRCh38
3893010 c.369C>T p.F123F Chr.19: 48616131-48616131 on Build GRCh38
3536576 c.454T>A p.F152I Chr.19: 48615914-48615914 on Build GRCh38
6502484 c.420C>T p.S140S Chr.19: 48616080-48616080 on Build GRCh38
3536577 c.436C>T p.R146* Chr.19: 48615932-48615932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749282951 Chr.19: 48616058-48616058 on Build GRCh38
rs113893377 Chr.19: 48615824-48615824 on Build GRCh38
rs747523641 Chr.19: 48616071-48616071 on Build GRCh38
rs767560988 Chr.19: 48616194-48616194 on Build GRCh38
rs748777728 Chr.19: 48616262-48616262 on Build GRCh38
rs939229440 Chr.19: 48616143-48616143 on Build GRCh38
rs373070088 Chr.19: 48616059-48616059 on Build GRCh38
rs1804326 Chr.19: 48616165-48616165 on Build GRCh38
rs376311951 Chr.19: 48615980-48615980 on Build GRCh38
rs879165350 Chr.19: 48615825-48615825 on Build GRCh38
rs769334709 Chr.19: 48616169-48616169 on Build GRCh38
rs748611471 Chr.19: 48616126-48616126 on Build GRCh38
rs752010192 Chr.19: 48616238-48616238 on Build GRCh38
rs533455813 Chr.19: 48616211-48616211 on Build GRCh38
rs1048829394 Chr.19: 48616187-48616187 on Build GRCh38
rs879214694 Chr.19: 48616076-48616076 on Build GRCh38
rs930358862 Chr.19: 48615991-48615991 on Build GRCh38
rs748264652 Chr.19: 48615939-48615939 on Build GRCh38
rs774185847 Chr.19: 48616188-48616188 on Build GRCh38
rs756023543 Chr.19: 48616221-48616221 on Build GRCh38
rs772717055 Chr.19: 48616178-48616178 on Build GRCh38
rs878944212 Chr.19: 48616075-48616075 on Build GRCh38
rs756802709 Chr.19: 48615912-48615912 on Build GRCh38
rs13870 Chr.19: 48616148-48616148 on Build GRCh38
rs772732417 Chr.19: 48616092-48616092 on Build GRCh38
rs371342921 Chr.19: 48615911-48615911 on Build GRCh38
rs754935395 Chr.19: 48616114-48616114 on Build GRCh38
rs368131281 Chr.19: 48615931-48615931 on Build GRCh38
rs1010715216 Chr.19: 48616206-48616206 on Build GRCh38
rs879093904 Chr.19: 48616073-48616073 on Build GRCh38
rs766171358 Chr.19: 48616110-48616110 on Build GRCh38
rs769856888 Chr.19: 48615940-48615940 on Build GRCh38
rs370173428 Chr.19: 48616243-48616243 on Build GRCh38
rs755388488 Chr.19: 48616240-48616240 on Build GRCh38
rs944507754 Chr.19: 48616060-48616060 on Build GRCh38
rs763908563 Chr.19: 48615900-48615900 on Build GRCh38
rs776972248 Chr.19: 48616171-48616171 on Build GRCh38
rs200238434 Chr.19: 48616227-48616227 on Build GRCh38
rs1024686202 Chr.19: 48615823-48615823 on Build GRCh38
rs11554934 Chr.19: 48616161-48616161 on Build GRCh38
rs755559605 Chr.19: 48615841-48615841 on Build GRCh38
rs751481958 Chr.19: 48616044-48616044 on Build GRCh38
rs374370198 Chr.19: 48616091-48616091 on Build GRCh38
rs113531766 Chr.19: 48615949-48615949 on Build GRCh38
rs772749012 Chr.19: 48615968-48615968 on Build GRCh38
rs751285320 Chr.19: 48616111-48616111 on Build GRCh38
rs202212986 Chr.19: 48615956-48615956 on Build GRCh38
rs200395884 Chr.19: 48616086-48616086 on Build GRCh38
rs1023036681 Chr.19: 48616061-48616061 on Build GRCh38
rs368542541 Chr.19: 48615961-48615961 on Build GRCh38
rs368592951 Chr.19: 48615862-48615862 on Build GRCh38
rs777831730 Chr.19: 48616052-48616052 on Build GRCh38
rs780597789 Chr.19: 48616163-48616163 on Build GRCh38
rs761204720 Chr.19: 48616082-48616082 on Build GRCh38
rs780030838 Chr.19: 48616038-48616038 on Build GRCh38
rs201450233 Chr.19: 48615971-48615971 on Build GRCh38
rs780399859 Chr.19: 48616231-48616231 on Build GRCh38
rs200880036 Chr.19: 48615932-48615932 on Build GRCh38
rs370747953 Chr.19: 48616080-48616080 on Build GRCh38
rs373946681 Chr.19: 48615919-48615919 on Build GRCh38
rs374788995 Chr.19: 48615957-48615957 on Build GRCh38
rs760233721 Chr.19: 48615896-48615896 on Build GRCh38
rs767423084 Chr.19: 48616118-48616118 on Build GRCh38
rs752307128 Chr.19: 48615892-48615892 on Build GRCh38
rs763291387 Chr.19: 48615870-48615870 on Build GRCh38
rs770264798 Chr.19: 48616248-48616248 on Build GRCh38
rs771309189 Chr.19: 48615878-48615878 on Build GRCh38
rs769150722 Chr.19: 48615837-48615838 on Build GRCh38
rs559300689 Chr.19: 48616177-48616177 on Build GRCh38
rs767866392 Chr.19: 48615891-48615891 on Build GRCh38
rs778775546 Chr.19: 48615848-48615848 on Build GRCh38
rs889267092 Chr.19: 48616009-48616009 on Build GRCh38
rs771837915 Chr.19: 48615850-48615850 on Build GRCh38
rs753994101 Chr.19: 48616119-48616119 on Build GRCh38
rs199608349 Chr.19: 48616226-48616226 on Build GRCh38
rs774517554 Chr.19: 48615882-48615882 on Build GRCh38
rs551842835 Chr.19: 48616081-48616081 on Build GRCh38
rs372151601 Chr.19: 48615845-48615845 on Build GRCh38
rs371360534 Chr.19: 48616068-48616068 on Build GRCh38
rs549893596 Chr.19: 48616149-48616149 on Build GRCh38
rs377412422 Chr.19: 48616062-48616062 on Build GRCh38
rs535680354 Chr.19: 48616241-48616241 on Build GRCh38
rs752566977 Chr.19: 48616217-48616217 on Build GRCh38
rs140941620 Chr.19: 48616098-48616098 on Build GRCh38
rs369266322 Chr.19: 48615994-48615994 on Build GRCh38
rs776139313 Chr.19: 48615969-48615969 on Build GRCh38
rs758096012 Chr.19: 48615995-48615995 on Build GRCh38
rs747758739 Chr.19: 48616173-48616173 on Build GRCh38
rs911515343 Chr.19: 48616016-48616016 on Build GRCh38
rs749534867 Chr.19: 48615952-48615952 on Build GRCh38
rs369589141 Chr.19: 48615988-48615988 on Build GRCh38
rs112968556 Chr.19: 48615948-48615948 on Build GRCh38
rs554024004 Chr.19: 48616253-48616253 on Build GRCh38
rs746313886 Chr.19: 48616064-48616064 on Build GRCh38
rs760124704 Chr.19: 48615888-48615888 on Build GRCh38
rs373747398 Chr.19: 48615989-48615989 on Build GRCh38
rs759298767 Chr.19: 48616189-48616189 on Build GRCh38
rs186259312 Chr.19: 48615856-48615856 on Build GRCh38
rs754848715 Chr.19: 48616049-48616049 on Build GRCh38
rs11554939 Chr.19: 48616160-48616160 on Build GRCh38
rs138053229 Chr.19: 48616131-48616131 on Build GRCh38
rs11554940 Chr.19: 48615920-48615920 on Build GRCh38
rs200188369 Chr.19: 48615951-48615951 on Build GRCh38
rs878964621 Chr.19: 48616074-48616074 on Build GRCh38
rs1019199806 Chr.19: 48616015-48616015 on Build GRCh38
rs574852321 Chr.19: 48615964-48615964 on Build GRCh38
rs765238385 Chr.19: 48616225-48616225 on Build GRCh38
rs773389388 Chr.19: 48615864-48615864 on Build GRCh38

View Full Product Details