Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48718692-48719187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAMSTR
Entrez Gene ID
284358
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases
MASTR
UniGene
Hs.191815
Gene Chromosome Location
Chr.19: 48705718-48719898 on Build GRCh38
Transcripts
NM_001130915.1 XM_011526808.2 XM_017026640.1 XM_017026641.1 XM_017026643.1 XM_017026644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1612541 c.58+6A>G p.? Chr.19: 48718968-48718968 on Build GRCh38
1590203 c.6C>T p.T2T Chr.19: 48719026-48719026 on Build GRCh38
6372236 c.4A>G p.T2A Chr.19: 48719028-48719028 on Build GRCh38
4790051 c.30C>G p.S10S Chr.19: 48719002-48719002 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs537016582 Chr.19: 48719006-48719006 on Build GRCh38
rs894687615 Chr.19: 48718778-48718778 on Build GRCh38
rs917044280 Chr.19: 48719064-48719064 on Build GRCh38
rs768503596 Chr.19: 48718950-48718950 on Build GRCh38
rs1054718589 Chr.19: 48718776-48718776 on Build GRCh38
rs949730440 Chr.19: 48719072-48719072 on Build GRCh38
rs1012937353 Chr.19: 48718807-48718807 on Build GRCh38
rs748011381 Chr.19: 48718951-48718951 on Build GRCh38
rs56736082 Chr.19: 48718825-48718825 on Build GRCh38
rs112642599 Chr.19: 48718808-48718808 on Build GRCh38
rs868516650 Chr.19: 48718755-48718755 on Build GRCh38
rs767914692 Chr.19: 48719039-48719039 on Build GRCh38
rs866991977 Chr.19: 48718987-48718987 on Build GRCh38
rs907187441 Chr.19: 48718809-48718809 on Build GRCh38
rs1045972845 Chr.19: 48719129-48719129 on Build GRCh38
rs760918395 Chr.19: 48719049-48719049 on Build GRCh38
rs949016076 Chr.19: 48719103-48719103 on Build GRCh38
rs752969236 Chr.19: 48719042-48719042 on Build GRCh38
rs180832216 Chr.19: 48718866-48718866 on Build GRCh38
rs936255053 Chr.19: 48719148-48719148 on Build GRCh38
rs903456526 Chr.19: 48718895-48718895 on Build GRCh38
rs949780659 Chr.19: 48718875-48718875 on Build GRCh38
rs555431100 Chr.19: 48719007-48719007 on Build GRCh38
rs924826761 Chr.19: 48719102-48719102 on Build GRCh38
rs868411573 Chr.19: 48718901-48718901 on Build GRCh38
rs757822537 Chr.19: 48719066-48719066 on Build GRCh38
rs760818720 Chr.19: 48718749-48718749 on Build GRCh38
rs573764910 Chr.19: 48719021-48719021 on Build GRCh38
rs936165394 Chr.19: 48719073-48719073 on Build GRCh38
rs895479269 Chr.19: 48718830-48718830 on Build GRCh38
rs1042356555 Chr.19: 48718883-48718883 on Build GRCh38
rs143605694 Chr.19: 48719152-48719152 on Build GRCh38
rs761837490 Chr.19: 48718991-48718991 on Build GRCh38
rs756156447 Chr.19: 48719118-48719118 on Build GRCh38
rs955551336 Chr.19: 48719010-48719010 on Build GRCh38
rs900011818 Chr.19: 48719153-48719153 on Build GRCh38
rs776231532 Chr.19: 48718989-48718989 on Build GRCh38
rs986049015 Chr.19: 48719041-48719041 on Build GRCh38
rs1015237570 Chr.19: 48719025-48719025 on Build GRCh38
rs764726028 Chr.19: 48718955-48718958 on Build GRCh38
rs759668909 Chr.19: 48719026-48719026 on Build GRCh38
rs899895815 Chr.19: 48718954-48718954 on Build GRCh38
rs754111384 Chr.19: 48719062-48719062 on Build GRCh38
rs973183356 Chr.19: 48719045-48719045 on Build GRCh38
rs936113114 Chr.19: 48718775-48718775 on Build GRCh38
rs73942312 Chr.19: 48719088-48719088 on Build GRCh38
rs924320574 Chr.19: 48719068-48719068 on Build GRCh38
rs138411987 Chr.19: 48718741-48718741 on Build GRCh38
rs1000350657 Chr.19: 48718911-48718911 on Build GRCh38
rs1054604632 Chr.19: 48719081-48719081 on Build GRCh38
rs973234752 Chr.19: 48718739-48718739 on Build GRCh38
rs997053705 Chr.19: 48719163-48719163 on Build GRCh38
rs764536902 Chr.19: 48719058-48719058 on Build GRCh38
rs754440528 Chr.19: 48718795-48718795 on Build GRCh38
rs958413589 Chr.19: 48719056-48719056 on Build GRCh38

View Full Product Details