Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 14635678-14636211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM84A
Entrez Gene ID
151354
Gene Name
family with sequence similarity 84 member A
Gene Aliases
NSE1, PP11517
UniGene
Hs.260855
Gene Chromosome Location
Chr.2: 14632686-14640046 on Build GRCh38
Transcripts
NM_145175.2 NM_001291410.1

Gene Symbol
LOC653602
Entrez Gene ID
653602
Gene Name
uncharacterized LOC653602
Gene Aliases
-
UniGene
Hs.657296
Gene Chromosome Location
Chr.2: 14635091-14650819 on Build GRCh38
Transcripts
NM_145175.2 NM_001291410.1

SNP Detail

SNP ID Chromosome Location
rs1008749642 Chr.2: 14635937-14635937 on Build GRCh38
rs987695943 Chr.2: 14636048-14636049 on Build GRCh38
rs939552524 Chr.2: 14635782-14635782 on Build GRCh38
rs533699192 Chr.2: 14636093-14636093 on Build GRCh38
rs1043827238 Chr.2: 14635809-14635809 on Build GRCh38
rs899876594 Chr.2: 14635857-14635857 on Build GRCh38
rs939624551 Chr.2: 14636183-14636183 on Build GRCh38
rs1029590315 Chr.2: 14635933-14635933 on Build GRCh38
rs906504166 Chr.2: 14636150-14636150 on Build GRCh38
rs935599598 Chr.2: 14636072-14636072 on Build GRCh38
rs1024056205 Chr.2: 14635801-14635801 on Build GRCh38
rs1054345991 Chr.2: 14636089-14636089 on Build GRCh38
rs964130813 Chr.2: 14635941-14635941 on Build GRCh38
rs973610492 Chr.2: 14636028-14636028 on Build GRCh38
rs1011443829 Chr.2: 14635807-14635807 on Build GRCh38
rs980444204 Chr.2: 14636101-14636101 on Build GRCh38
rs1036631356 Chr.2: 14636107-14636107 on Build GRCh38
rs550942143 Chr.2: 14635727-14635727 on Build GRCh38
rs1003382218 Chr.2: 14635877-14635877 on Build GRCh38
rs762705778 Chr.2: 14635901-14635901 on Build GRCh38
rs776465281 Chr.2: 14635921-14635921 on Build GRCh38
rs149856983 Chr.2: 14635711-14635711 on Build GRCh38
rs1025518579 Chr.2: 14635890-14635890 on Build GRCh38
rs922227254 Chr.2: 14635995-14635995 on Build GRCh38
rs954656665 Chr.2: 14635936-14635936 on Build GRCh38
rs948129513 Chr.2: 14636122-14636122 on Build GRCh38
rs763945047 Chr.2: 14635763-14635763 on Build GRCh38
rs757638629 Chr.2: 14635745-14635745 on Build GRCh38
rs927599450 Chr.2: 14636115-14636115 on Build GRCh38
rs530394704 Chr.2: 14636084-14636084 on Build GRCh38
rs374066235 Chr.2: 14635702-14635702 on Build GRCh38
rs892637825 Chr.2: 14635795-14635795 on Build GRCh38
rs928218017 Chr.2: 14635757-14635757 on Build GRCh38
rs1045163547 Chr.2: 14636146-14636146 on Build GRCh38
rs779205507 Chr.2: 14635972-14635972 on Build GRCh38
rs749889231 Chr.2: 14636067-14636067 on Build GRCh38
rs764901843 Chr.2: 14636045-14636045 on Build GRCh38
rs867699028 Chr.2: 14635775-14635775 on Build GRCh38
rs1021041973 Chr.2: 14636102-14636116 on Build GRCh38
rs935502737 Chr.2: 14635713-14635713 on Build GRCh38
rs906579294 Chr.2: 14635851-14635851 on Build GRCh38
rs1052630532 Chr.2: 14635786-14635786 on Build GRCh38
rs961895883 Chr.2: 14636005-14636005 on Build GRCh38
rs569482335 Chr.2: 14635956-14635956 on Build GRCh38

View Full Product Details