Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 33519866-33520367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGRP3
Entrez Gene ID
25780
Gene Name
RAS guanyl releasing protein 3
Gene Aliases
GRP3
UniGene
Hs.143674
Gene Chromosome Location
Chr.2: 33436348-33564731 on Build GRCh38
Transcripts
NM_001139488.1 NM_015376.2 NM_170672.2 XM_005264246.3 XM_011532746.2 XM_011532747.2 XM_011532748.2 XM_017003759.1 XM_017003760.1 XM_017003761.1 XM_017003762.1 XM_017003763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1407884 c.178C>T p.R60* Chr.2: 33519956-33519956 on Build GRCh38
3581320 c.211C>T p.R71* Chr.2: 33519989-33519989 on Build GRCh38
5651903 c.210T>G p.F70L Chr.2: 33519988-33519988 on Build GRCh38
5180261 c.189T>C p.T63T Chr.2: 33519967-33519967 on Build GRCh38
2943118 c.174G>A p.M58I Chr.2: 33519952-33519952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052345951 Chr.2: 33519914-33519914 on Build GRCh38
rs2305574 Chr.2: 33520335-33520335 on Build GRCh38
rs755026837 Chr.2: 33519997-33519997 on Build GRCh38
rs745710928 Chr.2: 33520194-33520194 on Build GRCh38
rs1010205593 Chr.2: 33519943-33519943 on Build GRCh38
rs974747435 Chr.2: 33519992-33519992 on Build GRCh38
rs897474133 Chr.2: 33520118-33520118 on Build GRCh38
rs372849070 Chr.2: 33519975-33519975 on Build GRCh38
rs369737208 Chr.2: 33519900-33519900 on Build GRCh38
rs551613087 Chr.2: 33520136-33520136 on Build GRCh38
rs759127605 Chr.2: 33520063-33520063 on Build GRCh38
rs139310832 Chr.2: 33520224-33520224 on Build GRCh38
rs377114910 Chr.2: 33519981-33519981 on Build GRCh38
rs763885185 Chr.2: 33520271-33520271 on Build GRCh38
rs947842940 Chr.2: 33520076-33520076 on Build GRCh38
rs747302924 Chr.2: 33519989-33519989 on Build GRCh38
rs2305575 Chr.2: 33520269-33520269 on Build GRCh38
rs556215136 Chr.2: 33519929-33519929 on Build GRCh38
rs141425351 Chr.2: 33520081-33520081 on Build GRCh38
rs7597095 Chr.2: 33519891-33519891 on Build GRCh38
rs891117498 Chr.2: 33520162-33520162 on Build GRCh38
rs914700487 Chr.2: 33520086-33520086 on Build GRCh38
rs1026320166 Chr.2: 33520325-33520325 on Build GRCh38
rs938895467 Chr.2: 33520317-33520317 on Build GRCh38
rs149997838 Chr.2: 33520225-33520225 on Build GRCh38
rs2305577 Chr.2: 33520111-33520111 on Build GRCh38
rs759039096 Chr.2: 33519945-33519945 on Build GRCh38
rs560604374 Chr.2: 33520061-33520061 on Build GRCh38
rs1031622446 Chr.2: 33520203-33520203 on Build GRCh38
rs907424355 Chr.2: 33520109-33520109 on Build GRCh38
rs928802920 Chr.2: 33520305-33520305 on Build GRCh38
rs991746100 Chr.2: 33520059-33520059 on Build GRCh38
rs1018887299 Chr.2: 33519958-33519958 on Build GRCh38
rs750786013 Chr.2: 33519963-33519963 on Build GRCh38
rs993252482 Chr.2: 33519939-33519939 on Build GRCh38
rs375539955 Chr.2: 33520084-33520084 on Build GRCh38
rs769582835 Chr.2: 33519902-33519902 on Build GRCh38
rs773178507 Chr.2: 33520051-33520051 on Build GRCh38
rs918818174 Chr.2: 33520019-33520019 on Build GRCh38
rs368067717 Chr.2: 33520053-33520053 on Build GRCh38
rs139065864 Chr.2: 33520043-33520043 on Build GRCh38
rs375583502 Chr.2: 33520134-33520134 on Build GRCh38
rs369611073 Chr.2: 33519957-33519957 on Build GRCh38
rs976414193 Chr.2: 33520184-33520184 on Build GRCh38
rs113430585 Chr.2: 33519905-33519905 on Build GRCh38
rs1044573640 Chr.2: 33520101-33520101 on Build GRCh38
rs79038952 Chr.2: 33520341-33520341 on Build GRCh38
rs995655969 Chr.2: 33520300-33520300 on Build GRCh38
rs1052788484 Chr.2: 33520114-33520114 on Build GRCh38
rs892340819 Chr.2: 33520135-33520135 on Build GRCh38
rs770776589 Chr.2: 33520056-33520056 on Build GRCh38
rs1048772057 Chr.2: 33520342-33520343 on Build GRCh38
rs985985680 Chr.2: 33519977-33519977 on Build GRCh38
rs527260981 Chr.2: 33520167-33520167 on Build GRCh38
rs370289650 Chr.2: 33519934-33519934 on Build GRCh38
rs200653982 Chr.2: 33519924-33519924 on Build GRCh38
rs866243572 Chr.2: 33519983-33519983 on Build GRCh38
rs757730510 Chr.2: 33519956-33519956 on Build GRCh38
rs781152306 Chr.2: 33520009-33520009 on Build GRCh38
rs767774248 Chr.2: 33520165-33520169 on Build GRCh38
rs192662835 Chr.2: 33520099-33520099 on Build GRCh38
rs759481075 Chr.2: 33519928-33519929 on Build GRCh38
rs200007507 Chr.2: 33519952-33519952 on Build GRCh38
rs576391331 Chr.2: 33519951-33519951 on Build GRCh38
rs887554926 Chr.2: 33520138-33520138 on Build GRCh38
rs547198548 Chr.2: 33520192-33520192 on Build GRCh38
rs765989167 Chr.2: 33519925-33519925 on Build GRCh38
rs777885378 Chr.2: 33520064-33520064 on Build GRCh38
rs1051127733 Chr.2: 33520142-33520142 on Build GRCh38
rs2305576 Chr.2: 33520141-33520141 on Build GRCh38

View Full Product Details