Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85607536-85608047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf68
Entrez Gene ID
388969
Gene Name
chromosome 2 open reading frame 68
Gene Aliases
HCRCN81
UniGene
Hs.516159
Gene Chromosome Location
Chr.2: 85605253-85612073 on Build GRCh38
Transcripts
NM_001013649.3 NR_046347.1 XM_006711922.1

Gene Symbol
USP39
Entrez Gene ID
10713
Gene Name
ubiquitin specific peptidase 39
Gene Aliases
65K, CGI-21, HSPC332, SAD1, SNRNP65
UniGene
Hs.654745
Gene Chromosome Location
Chr.2: 85602842-85649284 on Build GRCh38
Transcripts
NM_001013649.3 NR_046347.1 XM_006711922.1

SNP Detail

SNP ID Chromosome Location
rs967404331 Chr.2: 85607592-85607592 on Build GRCh38
rs1055005137 Chr.2: 85608020-85608020 on Build GRCh38
rs144704026 Chr.2: 85607777-85607777 on Build GRCh38
rs183012712 Chr.2: 85607598-85607598 on Build GRCh38
rs976786013 Chr.2: 85607842-85607842 on Build GRCh38
rs371618042 Chr.2: 85607876-85607876 on Build GRCh38
rs1010657587 Chr.2: 85607795-85607795 on Build GRCh38
rs149338104 Chr.2: 85607770-85607770 on Build GRCh38
rs529206615 Chr.2: 85607936-85607936 on Build GRCh38
rs764892936 Chr.2: 85607859-85607859 on Build GRCh38
rs957906170 Chr.2: 85607576-85607576 on Build GRCh38
rs187832994 Chr.2: 85607874-85607874 on Build GRCh38
rs1046315541 Chr.2: 85607765-85607765 on Build GRCh38
rs577067534 Chr.2: 85607667-85607668 on Build GRCh38
rs966410216 Chr.2: 85607841-85607841 on Build GRCh38
rs763500959 Chr.2: 85607647-85607647 on Build GRCh38
rs973190257 Chr.2: 85607707-85607707 on Build GRCh38
rs913550577 Chr.2: 85607583-85607583 on Build GRCh38
rs941433941 Chr.2: 85607672-85607672 on Build GRCh38
rs1014639110 Chr.2: 85607754-85607754 on Build GRCh38
rs931285408 Chr.2: 85607752-85607752 on Build GRCh38
rs974416465 Chr.2: 85607578-85607578 on Build GRCh38
rs1014846218 Chr.2: 85607970-85607970 on Build GRCh38
rs761278444 Chr.2: 85607620-85607620 on Build GRCh38
rs543649860 Chr.2: 85607835-85607835 on Build GRCh38
rs1055067037 Chr.2: 85607759-85607759 on Build GRCh38
rs147830362 Chr.2: 85607582-85607582 on Build GRCh38
rs564469610 Chr.2: 85607975-85607975 on Build GRCh38
rs985620479 Chr.2: 85607869-85607869 on Build GRCh38
rs972832659 Chr.2: 85607883-85607883 on Build GRCh38
rs779834547 Chr.2: 85607941-85607941 on Build GRCh38
rs920031202 Chr.2: 85607745-85607745 on Build GRCh38
rs1028691234 Chr.2: 85607922-85607922 on Build GRCh38
rs995821838 Chr.2: 85607885-85607885 on Build GRCh38
rs553347689 Chr.2: 85607677-85607677 on Build GRCh38
rs918629289 Chr.2: 85607894-85607894 on Build GRCh38
rs72923031 Chr.2: 85607949-85607949 on Build GRCh38
rs772599839 Chr.2: 85607565-85607565 on Build GRCh38
rs1046543157 Chr.2: 85607782-85607782 on Build GRCh38
rs922810441 Chr.2: 85607630-85607630 on Build GRCh38
rs576344532 Chr.2: 85607815-85607815 on Build GRCh38
rs138869489 Chr.2: 85607758-85607758 on Build GRCh38
rs954265680 Chr.2: 85607863-85607863 on Build GRCh38
rs536105338 Chr.2: 85607676-85607676 on Build GRCh38
rs973274746 Chr.2: 85607985-85607985 on Build GRCh38
rs751481652 Chr.2: 85607976-85607976 on Build GRCh38
rs1021074313 Chr.2: 85607804-85607804 on Build GRCh38
rs1049859987 Chr.2: 85607655-85607655 on Build GRCh38
rs1046117991 Chr.2: 85607948-85607948 on Build GRCh38
rs1002052434 Chr.2: 85607775-85607775 on Build GRCh38
rs909981721 Chr.2: 85607667-85607667 on Build GRCh38
rs1033449237 Chr.2: 85607560-85607560 on Build GRCh38
rs756317431 Chr.2: 85607607-85607607 on Build GRCh38

View Full Product Details