Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176094361-176094897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD13
Entrez Gene ID
3239
Gene Name
homeobox D13
Gene Aliases
BDE, BDSD, HOX4I, SPD, SPD1
UniGene
Hs.152414
Gene Chromosome Location
Chr.2: 176087487-176095938 on Build GRCh38
Transcripts
NM_000523.3 XM_011511068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1401418 c.797G>A p.R266Q Chr.2: 176094519-176094519 on Build GRCh38
3570176 c.980G>A p.R327Q Chr.2: 176094678-176094678 on Build GRCh38
1009840 c.844G>T p.E282* Chr.2: 176094566-176094566 on Build GRCh38
6681804 c.912C>A p.D304E Chr.2: 176094610-176094610 on Build GRCh38
353052 c.782G>T p.G261V Chr.2: 176094480-176094480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933088116 Chr.2: 176094789-176094789 on Build GRCh38
rs28933082 Chr.2: 176094614-176094614 on Build GRCh38
rs138630870 Chr.2: 176094512-176094512 on Build GRCh38
rs149097061 Chr.2: 176094550-176094550 on Build GRCh38
rs751010417 Chr.2: 176094446-176094446 on Build GRCh38
rs199549554 Chr.2: 176094779-176094779 on Build GRCh38
rs373235672 Chr.2: 176094447-176094447 on Build GRCh38
rs778217352 Chr.2: 176094770-176094770 on Build GRCh38
rs781344527 Chr.2: 176094753-176094753 on Build GRCh38
rs749985629 Chr.2: 176094590-176094592 on Build GRCh38
rs1017490101 Chr.2: 176094827-176094827 on Build GRCh38
rs780305650 Chr.2: 176094493-176094493 on Build GRCh38
rs751801650 Chr.2: 176094731-176094731 on Build GRCh38
rs757927553 Chr.2: 176094639-176094651 on Build GRCh38
rs964748050 Chr.2: 176094830-176094830 on Build GRCh38
rs778129623 Chr.2: 176094628-176094628 on Build GRCh38
rs770226826 Chr.2: 176094621-176094621 on Build GRCh38
rs943267669 Chr.2: 176094623-176094623 on Build GRCh38
rs535266089 Chr.2: 176094852-176094852 on Build GRCh38
rs775653772 Chr.2: 176094387-176094388 on Build GRCh38
rs982975708 Chr.2: 176094422-176094422 on Build GRCh38
rs1005878815 Chr.2: 176094835-176094835 on Build GRCh38
rs752471203 Chr.2: 176094464-176094464 on Build GRCh38
rs879255265 Chr.2: 176094615-176094615 on Build GRCh38
rs866506676 Chr.2: 176094526-176094526 on Build GRCh38
rs104893635 Chr.2: 176094672-176094672 on Build GRCh38
rs567865840 Chr.2: 176094805-176094805 on Build GRCh38
rs767444448 Chr.2: 176094461-176094461 on Build GRCh38
rs376366804 Chr.2: 176094609-176094609 on Build GRCh38
rs185454060 Chr.2: 176094508-176094508 on Build GRCh38
rs200799183 Chr.2: 176094448-176094448 on Build GRCh38
rs1038624473 Chr.2: 176094501-176094501 on Build GRCh38
rs764838478 Chr.2: 176094478-176094478 on Build GRCh38
rs373737493 Chr.2: 176094488-176094488 on Build GRCh38
rs755071995 Chr.2: 176094605-176094605 on Build GRCh38
rs945367146 Chr.2: 176094520-176094520 on Build GRCh38
rs71004256 Chr.2: 176094416-176094417 on Build GRCh38
rs774807865 Chr.2: 176094677-176094677 on Build GRCh38
rs28928892 Chr.2: 176094645-176094645 on Build GRCh38
rs113439497 Chr.2: 176094392-176094393 on Build GRCh38
rs199982017 Chr.2: 176094511-176094511 on Build GRCh38
rs759051786 Chr.2: 176094547-176094547 on Build GRCh38
rs753726144 Chr.2: 176094708-176094708 on Build GRCh38
rs759038167 Chr.2: 176094661-176094661 on Build GRCh38
rs747071040 Chr.2: 176094502-176094502 on Build GRCh38
rs879010051 Chr.2: 176094386-176094389 on Build GRCh38
rs1030395491 Chr.2: 176094864-176094864 on Build GRCh38
rs779349115 Chr.2: 176094485-176094485 on Build GRCh38
rs759071152 Chr.2: 176094561-176094561 on Build GRCh38
rs773255556 Chr.2: 176094387-176094387 on Build GRCh38
rs200750564 Chr.2: 176094518-176094518 on Build GRCh38
rs760465283 Chr.2: 176094562-176094562 on Build GRCh38
rs150319624 Chr.2: 176094637-176094637 on Build GRCh38
rs769084692 Chr.2: 176094505-176094505 on Build GRCh38
rs950519196 Chr.2: 176094866-176094866 on Build GRCh38
rs766764389 Chr.2: 176094721-176094721 on Build GRCh38
rs772837876 Chr.2: 176094442-176094442 on Build GRCh38
rs543897513 Chr.2: 176094769-176094769 on Build GRCh38
rs755087311 Chr.2: 176094745-176094745 on Build GRCh38
rs570355606 Chr.2: 176094799-176094799 on Build GRCh38
rs28928891 Chr.2: 176094662-176094662 on Build GRCh38
rs767313108 Chr.2: 176094617-176094617 on Build GRCh38
rs753639374 Chr.2: 176094577-176094577 on Build GRCh38
rs751640729 Chr.2: 176094591-176094591 on Build GRCh38
rs770519205 Chr.2: 176094519-176094519 on Build GRCh38
rs369216922 Chr.2: 176094703-176094703 on Build GRCh38
rs372343718 Chr.2: 176094587-176094587 on Build GRCh38
rs113848671 Chr.2: 176094385-176094385 on Build GRCh38
rs1017223163 Chr.2: 176094845-176094845 on Build GRCh38
rs370288408 Chr.2: 176094475-176094475 on Build GRCh38
rs762792202 Chr.2: 176094435-176094435 on Build GRCh38
rs761520404 Chr.2: 176094452-176094453 on Build GRCh38
rs773877947 Chr.2: 176094543-176094543 on Build GRCh38
rs925188102 Chr.2: 176094593-176094593 on Build GRCh38
rs763998494 Chr.2: 176094569-176094569 on Build GRCh38
rs752667409 Chr.2: 176094758-176094758 on Build GRCh38
rs875989842 Chr.2: 176094671-176094671 on Build GRCh38
rs962901444 Chr.2: 176094418-176094418 on Build GRCh38
rs898766013 Chr.2: 176094857-176094857 on Build GRCh38
rs975743189 Chr.2: 176094855-176094855 on Build GRCh38
rs758638811 Chr.2: 176094491-176094491 on Build GRCh38
rs758726777 Chr.2: 176094579-176094579 on Build GRCh38
rs761679301 Chr.2: 176094712-176094712 on Build GRCh38
rs765284913 Chr.2: 176094386-176094387 on Build GRCh38
rs781708716 Chr.2: 176094507-176094507 on Build GRCh38
rs10649769 Chr.2: 176094385-176094386 on Build GRCh38
rs201504145 Chr.2: 176094436-176094436 on Build GRCh38
rs760687062 Chr.2: 176094679-176094679 on Build GRCh38
rs111593893 Chr.2: 176094749-176094749 on Build GRCh38
rs548947724 Chr.2: 176094426-176094426 on Build GRCh38
rs34471425 Chr.2: 176094417-176094418 on Build GRCh38
rs878854344 Chr.2: 176094532-176094532 on Build GRCh38

View Full Product Details