Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 177626819-177627365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE11A
Entrez Gene ID
50940
Gene Name
phosphodiesterase 11A
Gene Aliases
PPNAD2
UniGene
Hs.570273
Gene Chromosome Location
Chr.2: 177623249-178108339 on Build GRCh38
Transcripts
NM_001077196.1 NM_001077197.1 NM_016953.3

SNP Detail

SNP ID Chromosome Location
rs891240587 Chr.2: 177627082-177627083 on Build GRCh38
rs754548452 Chr.2: 177627103-177627103 on Build GRCh38
rs897329962 Chr.2: 177627332-177627332 on Build GRCh38
rs1043088064 Chr.2: 177627313-177627313 on Build GRCh38
rs998629761 Chr.2: 177627137-177627137 on Build GRCh38
rs751195588 Chr.2: 177626882-177626882 on Build GRCh38
rs950832420 Chr.2: 177627258-177627258 on Build GRCh38
rs369865310 Chr.2: 177626939-177626939 on Build GRCh38
rs1053251243 Chr.2: 177626850-177626850 on Build GRCh38
rs376383732 Chr.2: 177627288-177627288 on Build GRCh38
rs947072055 Chr.2: 177626860-177626860 on Build GRCh38
rs1026331997 Chr.2: 177627251-177627251 on Build GRCh38
rs990860171 Chr.2: 177627179-177627179 on Build GRCh38
rs1005936884 Chr.2: 177626861-177626861 on Build GRCh38
rs892914186 Chr.2: 177627285-177627285 on Build GRCh38
rs547265893 Chr.2: 177627104-177627104 on Build GRCh38
rs1013173432 Chr.2: 177627186-177627186 on Build GRCh38
rs963671043 Chr.2: 177627221-177627221 on Build GRCh38
rs188093837 Chr.2: 177627068-177627068 on Build GRCh38
rs896011188 Chr.2: 177627173-177627173 on Build GRCh38
rs902910730 Chr.2: 177626868-177626868 on Build GRCh38
rs11694013 Chr.2: 177627211-177627211 on Build GRCh38
rs180948506 Chr.2: 177627109-177627109 on Build GRCh38
rs552609160 Chr.2: 177626917-177626917 on Build GRCh38
rs1057309849 Chr.2: 177627159-177627159 on Build GRCh38
rs201367138 Chr.2: 177626896-177626896 on Build GRCh38
rs112052828 Chr.2: 177627301-177627301 on Build GRCh38
rs934452954 Chr.2: 177627214-177627214 on Build GRCh38
rs1008422814 Chr.2: 177627029-177627029 on Build GRCh38
rs545098608 Chr.2: 177626871-177626871 on Build GRCh38
rs147825907 Chr.2: 177627014-177627017 on Build GRCh38
rs965221246 Chr.2: 177627287-177627287 on Build GRCh38
rs71010806 Chr.2: 177626900-177626900 on Build GRCh38
rs371176337 Chr.2: 177627011-177627020 on Build GRCh38
rs13002711 Chr.2: 177627015-177627015 on Build GRCh38
rs573770060 Chr.2: 177627309-177627309 on Build GRCh38
rs190752755 Chr.2: 177627305-177627305 on Build GRCh38
rs75238103 Chr.2: 177626958-177626958 on Build GRCh38
rs920934389 Chr.2: 177626849-177626849 on Build GRCh38
rs912375840 Chr.2: 177627283-177627283 on Build GRCh38
rs185047838 Chr.2: 177627005-177627005 on Build GRCh38
rs1015968320 Chr.2: 177626897-177626897 on Build GRCh38
rs573174750 Chr.2: 177626844-177626844 on Build GRCh38
rs948641485 Chr.2: 177627197-177627197 on Build GRCh38
rs892808939 Chr.2: 177626857-177626857 on Build GRCh38
rs921144760 Chr.2: 177627290-177627290 on Build GRCh38
rs1011617071 Chr.2: 177626867-177626867 on Build GRCh38
rs181277510 Chr.2: 177627002-177627002 on Build GRCh38
rs889433262 Chr.2: 177627299-177627299 on Build GRCh38
rs987544157 Chr.2: 177627279-177627279 on Build GRCh38
rs1042735177 Chr.2: 177626865-177626865 on Build GRCh38
rs1019766618 Chr.2: 177627030-177627030 on Build GRCh38
rs185167360 Chr.2: 177627120-177627120 on Build GRCh38
rs768524227 Chr.2: 177627023-177627041 on Build GRCh38
rs565303705 Chr.2: 177626942-177626942 on Build GRCh38
rs981333187 Chr.2: 177627199-177627199 on Build GRCh38
rs200710199 Chr.2: 177626900-177626900 on Build GRCh38
rs924330784 Chr.2: 177627326-177627326 on Build GRCh38
rs13002713 Chr.2: 177627018-177627018 on Build GRCh38
rs550925125 Chr.2: 177627134-177627134 on Build GRCh38
rs1023257365 Chr.2: 177627192-177627192 on Build GRCh38
rs995206781 Chr.2: 177627227-177627227 on Build GRCh38

View Full Product Details