Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203390947-203391487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI2
Entrez Gene ID
10152
Gene Name
abl-interactor 2
Gene Aliases
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
UniGene
Hs.471156
Gene Chromosome Location
Chr.2: 203327874-203432173 on Build GRCh38
Transcripts
NM_001282925.1 NM_001282926.1 NM_001282927.1 NM_001282932.1 NM_005759.5 XM_005246217.2 XM_005246218.2 XM_005246221.2 XM_005246225.2 XM_005246226.2 XM_006712174.2 XM_006712177.2 XM_006712179.2 XM_006712180.2 XM_006712181.2 XM_006712182.2 XM_006712185.1 XM_006712186.2 XM_011510472.2 XM_011510473.2 XM_017003118.1 XM_017003119.1 XM_017003120.1 XM_017003121.1 XM_017003122.1 XM_017003123.1 XM_017003124.1 XM_017003125.1 XM_017003126.1 XM_017003127.1 XM_017003128.1 XM_017003129.1 XM_017003130.1 XM_017003131.1 XM_017003132.1 XM_017003133.1 XM_017003134.1 XM_017003135.1 XM_017003136.1 XM_017003137.1 XM_017003138.1 XM_017003139.1 XM_017003140.1 XM_017003141.1 XM_017003142.1 XM_017003143.1 XM_017003144.1 XM_017003145.1 XM_017003146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6856207 c.477C>T p.N159N Chr.2: 203391060-203391060 on Build GRCh38
4881584 c.578G>T p.G193V Chr.2: 203391143-203391143 on Build GRCh38
6254231 c.577G>T p.G193W Chr.2: 203391142-203391142 on Build GRCh38
5058579 c.515C>T p.P172L Chr.2: 203391080-203391080 on Build GRCh38
6856209 c.342C>T p.N114N Chr.2: 203391060-203391060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901507586 Chr.2: 203390978-203390978 on Build GRCh38
rs374374902 Chr.2: 203391146-203391146 on Build GRCh38
rs372878946 Chr.2: 203391264-203391264 on Build GRCh38
rs775631405 Chr.2: 203391032-203391032 on Build GRCh38
rs888837499 Chr.2: 203391336-203391336 on Build GRCh38
rs762848081 Chr.2: 203391104-203391104 on Build GRCh38
rs973595586 Chr.2: 203391026-203391026 on Build GRCh38
rs897566686 Chr.2: 203391369-203391369 on Build GRCh38
rs1021431041 Chr.2: 203391367-203391367 on Build GRCh38
rs569237316 Chr.2: 203391199-203391199 on Build GRCh38
rs530240719 Chr.2: 203391285-203391285 on Build GRCh38
rs901499235 Chr.2: 203391212-203391212 on Build GRCh38
rs997619589 Chr.2: 203391232-203391232 on Build GRCh38
rs199948488 Chr.2: 203391016-203391016 on Build GRCh38
rs997270987 Chr.2: 203390989-203390989 on Build GRCh38
rs756210107 Chr.2: 203391444-203391444 on Build GRCh38
rs1008500983 Chr.2: 203391281-203391281 on Build GRCh38
rs1050273590 Chr.2: 203391220-203391220 on Build GRCh38
rs147307505 Chr.2: 203391096-203391096 on Build GRCh38
rs13030411 Chr.2: 203391160-203391160 on Build GRCh38
rs753004161 Chr.2: 203391315-203391315 on Build GRCh38
rs560063469 Chr.2: 203390972-203390972 on Build GRCh38
rs531533750 Chr.2: 203391083-203391083 on Build GRCh38
rs890403892 Chr.2: 203391223-203391223 on Build GRCh38
rs1041319117 Chr.2: 203391207-203391207 on Build GRCh38
rs377657499 Chr.2: 203391172-203391172 on Build GRCh38
rs749830902 Chr.2: 203391038-203391038 on Build GRCh38
rs765198855 Chr.2: 203391147-203391148 on Build GRCh38
rs1050162256 Chr.2: 203391270-203391270 on Build GRCh38
rs769070391 Chr.2: 203391170-203391170 on Build GRCh38
rs748533381 Chr.2: 203391176-203391176 on Build GRCh38
rs200958880 Chr.2: 203391081-203391081 on Build GRCh38
rs372227424 Chr.2: 203391179-203391179 on Build GRCh38
rs182980568 Chr.2: 203391356-203391356 on Build GRCh38
rs756981033 Chr.2: 203391117-203391117 on Build GRCh38
rs769180413 Chr.2: 203391040-203391040 on Build GRCh38
rs916004956 Chr.2: 203391154-203391154 on Build GRCh38
rs953223587 Chr.2: 203390999-203390999 on Build GRCh38
rs749337376 Chr.2: 203391156-203391156 on Build GRCh38
rs549992078 Chr.2: 203391135-203391135 on Build GRCh38
rs770708704 Chr.2: 203391076-203391076 on Build GRCh38
rs773704866 Chr.2: 203391182-203391182 on Build GRCh38
rs1029192791 Chr.2: 203390990-203390990 on Build GRCh38
rs369937232 Chr.2: 203391112-203391112 on Build GRCh38
rs767822250 Chr.2: 203390971-203390971 on Build GRCh38
rs199992443 Chr.2: 203391061-203391061 on Build GRCh38
rs533933845 Chr.2: 203391362-203391362 on Build GRCh38
rs199497527 Chr.2: 203391072-203391072 on Build GRCh38
rs962127937 Chr.2: 203391023-203391023 on Build GRCh38
rs113705942 Chr.2: 203391314-203391314 on Build GRCh38
rs967245250 Chr.2: 203391410-203391410 on Build GRCh38
rs769464448 Chr.2: 203391194-203391194 on Build GRCh38
rs940236510 Chr.2: 203391203-203391203 on Build GRCh38
rs1015045866 Chr.2: 203391289-203391289 on Build GRCh38
rs991577243 Chr.2: 203391111-203391111 on Build GRCh38
rs372974083 Chr.2: 203391359-203391359 on Build GRCh38
rs781408647 Chr.2: 203391132-203391132 on Build GRCh38
rs769708409 Chr.2: 203391002-203391002 on Build GRCh38
rs745954384 Chr.2: 203391000-203391000 on Build GRCh38
rs1011846065 Chr.2: 203391014-203391014 on Build GRCh38
rs139291552 Chr.2: 203391080-203391080 on Build GRCh38
rs759150569 Chr.2: 203391185-203391185 on Build GRCh38
rs764043362 Chr.2: 203391110-203391110 on Build GRCh38
rs994962151 Chr.2: 203391375-203391375 on Build GRCh38
rs548416866 Chr.2: 203391301-203391301 on Build GRCh38
rs756359107 Chr.2: 203391138-203391138 on Build GRCh38
rs946823262 Chr.2: 203391152-203391152 on Build GRCh38

View Full Product Details