Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215943049-215943553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MREG
Entrez Gene ID
55686
Gene Name
melanoregulin
Gene Aliases
DSU, WDT2
UniGene
Hs.620391
Gene Chromosome Location
Chr.2: 215939097-216013623 on Build GRCh38
Transcripts
NM_018000.2 XM_011511468.2

SNP Detail

SNP ID Chromosome Location
rs561133504 Chr.2: 215943465-215943465 on Build GRCh38
rs891471861 Chr.2: 215943345-215943345 on Build GRCh38
rs973858019 Chr.2: 215943474-215943474 on Build GRCh38
rs184239338 Chr.2: 215943348-215943348 on Build GRCh38
rs761831379 Chr.2: 215943375-215943377 on Build GRCh38
rs545041911 Chr.2: 215943136-215943136 on Build GRCh38
rs1045407877 Chr.2: 215943128-215943128 on Build GRCh38
rs980257943 Chr.2: 215943391-215943391 on Build GRCh38
rs938948888 Chr.2: 215943114-215943114 on Build GRCh38
rs748751995 Chr.2: 215943442-215943442 on Build GRCh38
rs940759339 Chr.2: 215943169-215943169 on Build GRCh38
rs373377568 Chr.2: 215943447-215943447 on Build GRCh38
rs957203929 Chr.2: 215943357-215943357 on Build GRCh38
rs1049599699 Chr.2: 215943276-215943276 on Build GRCh38
rs548840818 Chr.2: 215943338-215943338 on Build GRCh38
rs1007556358 Chr.2: 215943274-215943274 on Build GRCh38
rs1018993999 Chr.2: 215943281-215943281 on Build GRCh38
rs771666049 Chr.2: 215943373-215943373 on Build GRCh38
rs948370766 Chr.2: 215943093-215943093 on Build GRCh38
rs969820571 Chr.2: 215943405-215943405 on Build GRCh38
rs140398441 Chr.2: 215943327-215943327 on Build GRCh38
rs906792326 Chr.2: 215943134-215943134 on Build GRCh38
rs550980031 Chr.2: 215943186-215943186 on Build GRCh38
rs1049128499 Chr.2: 215943252-215943252 on Build GRCh38
rs766329119 Chr.2: 215943441-215943442 on Build GRCh38
rs188697413 Chr.2: 215943469-215943469 on Build GRCh38
rs564899879 Chr.2: 215943155-215943155 on Build GRCh38
rs995434219 Chr.2: 215943219-215943219 on Build GRCh38
rs13553 Chr.2: 215943191-215943191 on Build GRCh38
rs915647492 Chr.2: 215943382-215943382 on Build GRCh38
rs1036595008 Chr.2: 215943470-215943470 on Build GRCh38
rs550009001 Chr.2: 215943446-215943446 on Build GRCh38
rs915678790 Chr.2: 215943081-215943081 on Build GRCh38
rs773883592 Chr.2: 215943458-215943458 on Build GRCh38
rs532728693 Chr.2: 215943185-215943185 on Build GRCh38
rs6435927 Chr.2: 215943429-215943429 on Build GRCh38
rs754135033 Chr.2: 215943463-215943463 on Build GRCh38
rs889234488 Chr.2: 215943255-215943255 on Build GRCh38

View Full Product Details