Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1306190-1306685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNPH
Entrez Gene ID
9751
Gene Name
syntaphilin
Gene Aliases
-
UniGene
Hs.323833
Gene Chromosome Location
Chr.20: 1266291-1309327 on Build GRCh38
Transcripts
NM_001318234.1 NM_014723.3 XM_005260889.3 XM_011529402.2 XM_011529403.2 XM_011529404.2

SNP Detail

SNP ID Chromosome Location
rs925295237 Chr.20: 1306325-1306325 on Build GRCh38
rs769213729 Chr.20: 1306588-1306588 on Build GRCh38
rs6033384 Chr.20: 1306489-1306489 on Build GRCh38
rs1032880596 Chr.20: 1306662-1306662 on Build GRCh38
rs987810536 Chr.20: 1306303-1306303 on Build GRCh38
rs1050235060 Chr.20: 1306449-1306449 on Build GRCh38
rs752496353 Chr.20: 1306287-1306287 on Build GRCh38
rs931470708 Chr.20: 1306375-1306375 on Build GRCh38
rs1000669932 Chr.20: 1306465-1306465 on Build GRCh38
rs547032987 Chr.20: 1306285-1306285 on Build GRCh38
rs912236461 Chr.20: 1306324-1306324 on Build GRCh38
rs765851209 Chr.20: 1306243-1306243 on Build GRCh38
rs887368922 Chr.20: 1306401-1306401 on Build GRCh38
rs753370819 Chr.20: 1306260-1306260 on Build GRCh38
rs908751248 Chr.20: 1306641-1306641 on Build GRCh38
rs3764715 Chr.20: 1306407-1306407 on Build GRCh38
rs745917424 Chr.20: 1306473-1306473 on Build GRCh38
rs975119862 Chr.20: 1306604-1306604 on Build GRCh38
rs910270029 Chr.20: 1306460-1306460 on Build GRCh38
rs1000951783 Chr.20: 1306644-1306644 on Build GRCh38
rs539332108 Chr.20: 1306415-1306415 on Build GRCh38
rs6033383 Chr.20: 1306342-1306342 on Build GRCh38
rs994415935 Chr.20: 1306216-1306216 on Build GRCh38
rs552124609 Chr.20: 1306608-1306608 on Build GRCh38
rs566666568 Chr.20: 1306491-1306491 on Build GRCh38
rs933121185 Chr.20: 1306423-1306423 on Build GRCh38
rs373808836 Chr.20: 1306592-1306592 on Build GRCh38
rs758132158 Chr.20: 1306339-1306339 on Build GRCh38
rs749505258 Chr.20: 1306495-1306495 on Build GRCh38
rs960827066 Chr.20: 1306257-1306257 on Build GRCh38
rs1048576604 Chr.20: 1306640-1306640 on Build GRCh38
rs1047311694 Chr.20: 1306545-1306545 on Build GRCh38
rs956957086 Chr.20: 1306327-1306327 on Build GRCh38
rs530972980 Chr.20: 1306500-1306500 on Build GRCh38
rs1025942611 Chr.20: 1306218-1306218 on Build GRCh38
rs1048652899 Chr.20: 1306389-1306389 on Build GRCh38
rs567743817 Chr.20: 1306660-1306661 on Build GRCh38
rs759134117 Chr.20: 1306268-1306268 on Build GRCh38
rs6041281 Chr.20: 1306425-1306425 on Build GRCh38
rs921399136 Chr.20: 1306355-1306355 on Build GRCh38
rs569453938 Chr.20: 1306472-1306472 on Build GRCh38
rs969172895 Chr.20: 1306463-1306463 on Build GRCh38
rs3764714 Chr.20: 1306461-1306461 on Build GRCh38
rs770743384 Chr.20: 1306328-1306328 on Build GRCh38
rs529928452 Chr.20: 1306432-1306432 on Build GRCh38
rs146810897 Chr.20: 1306273-1306273 on Build GRCh38
rs965424618 Chr.20: 1306548-1306548 on Build GRCh38
rs3764713 Chr.20: 1306607-1306607 on Build GRCh38
rs969203811 Chr.20: 1306270-1306270 on Build GRCh38
rs1013771848 Chr.20: 1306264-1306264 on Build GRCh38

View Full Product Details