Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31860962-31861463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP15
Entrez Gene ID
128853
Gene Name
dual specificity phosphatase 15
Gene Aliases
C20orf57, VHY
UniGene
Hs.434108
Gene Chromosome Location
Chr.20: 31845578-31870744 on Build GRCh38
Transcripts
NM_001012644.2 NM_001320478.1 NM_001320479.1 NM_080611.4 NM_177991.2 XM_017027654.1 XM_017027655.1 XM_017027656.1 XM_017027657.1 XM_017027658.1 XM_017027659.1 XM_017027660.1 XM_017027661.1 XM_017027662.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6359679 c.658C>A p.Q220K Chr.20: 31861453-31861453 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs957751801 Chr.20: 31861017-31861017 on Build GRCh38
rs566167449 Chr.20: 31861406-31861406 on Build GRCh38
rs957804295 Chr.20: 31861353-31861353 on Build GRCh38
rs779614720 Chr.20: 31861321-31861321 on Build GRCh38
rs779252037 Chr.20: 31861382-31861382 on Build GRCh38
rs6121248 Chr.20: 31861284-31861284 on Build GRCh38
rs545338861 Chr.20: 31861081-31861081 on Build GRCh38
rs756760766 Chr.20: 31861371-31861371 on Build GRCh38
rs945022704 Chr.20: 31861084-31861084 on Build GRCh38
rs913589864 Chr.20: 31861064-31861064 on Build GRCh38
rs1008085924 Chr.20: 31861008-31861008 on Build GRCh38
rs1026988772 Chr.20: 31861117-31861117 on Build GRCh38
rs752601342 Chr.20: 31861414-31861414 on Build GRCh38
rs1041346229 Chr.20: 31861040-31861040 on Build GRCh38
rs765501755 Chr.20: 31861411-31861411 on Build GRCh38
rs970905311 Chr.20: 31860999-31860999 on Build GRCh38
rs943185606 Chr.20: 31861119-31861119 on Build GRCh38
rs763978815 Chr.20: 31861419-31861419 on Build GRCh38
rs751494522 Chr.20: 31861421-31861421 on Build GRCh38
rs760705192 Chr.20: 31861394-31861394 on Build GRCh38
rs987868670 Chr.20: 31861330-31861330 on Build GRCh38
rs868584754 Chr.20: 31861083-31861083 on Build GRCh38
rs913388023 Chr.20: 31861097-31861097 on Build GRCh38
rs930451220 Chr.20: 31861432-31861432 on Build GRCh38
rs780784853 Chr.20: 31861439-31861439 on Build GRCh38
rs997228460 Chr.20: 31861068-31861068 on Build GRCh38
rs946167060 Chr.20: 31861115-31861115 on Build GRCh38
rs772625177 Chr.20: 31861388-31861388 on Build GRCh38
rs1015670340 Chr.20: 31861212-31861212 on Build GRCh38
rs369148629 Chr.20: 31861369-31861369 on Build GRCh38
rs879608489 Chr.20: 31861039-31861039 on Build GRCh38
rs113505727 Chr.20: 31861380-31861380 on Build GRCh38
rs771018692 Chr.20: 31861400-31861400 on Build GRCh38
rs776536595 Chr.20: 31861404-31861404 on Build GRCh38
rs967563723 Chr.20: 31861273-31861296 on Build GRCh38
rs758427395 Chr.20: 31861365-31861365 on Build GRCh38
rs1004245238 Chr.20: 31861147-31861147 on Build GRCh38
rs762838662 Chr.20: 31861418-31861418 on Build GRCh38
rs527776079 Chr.20: 31861215-31861215 on Build GRCh38
rs556390346 Chr.20: 31861019-31861019 on Build GRCh38
rs544099319 Chr.20: 31861013-31861013 on Build GRCh38
rs879165089 Chr.20: 31861009-31861009 on Build GRCh38
rs560331706 Chr.20: 31861082-31861082 on Build GRCh38
rs12569 Chr.20: 31861378-31861378 on Build GRCh38
rs200474617 Chr.20: 31861431-31861431 on Build GRCh38
rs1004613957 Chr.20: 31861300-31861300 on Build GRCh38
rs1040563533 Chr.20: 31861305-31861305 on Build GRCh38
rs577868639 Chr.20: 31861031-31861031 on Build GRCh38
rs924951101 Chr.20: 31861319-31861319 on Build GRCh38
rs373419129 Chr.20: 31861374-31861374 on Build GRCh38
rs899760014 Chr.20: 31861052-31861052 on Build GRCh38
rs1048824993 Chr.20: 31861438-31861438 on Build GRCh38
rs980807136 Chr.20: 31861278-31861278 on Build GRCh38
rs952764374 Chr.20: 31861133-31861133 on Build GRCh38
rs748454392 Chr.20: 31861385-31861385 on Build GRCh38
rs550120515 Chr.20: 31861323-31861323 on Build GRCh38
rs987438335 Chr.20: 31861102-31861102 on Build GRCh38
rs531911083 Chr.20: 31861311-31861311 on Build GRCh38
rs993041392 Chr.20: 31861234-31861234 on Build GRCh38
rs146846724 Chr.20: 31861240-31861240 on Build GRCh38
rs777843675 Chr.20: 31861368-31861368 on Build GRCh38
rs752476551 Chr.20: 31861361-31861361 on Build GRCh38
rs561443325 Chr.20: 31861270-31861270 on Build GRCh38

View Full Product Details