Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33239719-33240224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA1
Entrez Gene ID
51297
Gene Name
BPI fold containing family A member 1
Gene Aliases
LUNX, NASG, PLUNC, SPLUNC1, SPURT, bA49G10.5
UniGene
Hs.211092
Gene Chromosome Location
Chr.20: 33235996-33243309 on Build GRCh38
Transcripts
NM_001243193.1 NM_016583.3 NM_130852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5606355 c.327G>A p.K109K Chr.20: 33239809-33239809 on Build GRCh38
1713351 c.349G>A p.E117K Chr.20: 33239831-33239831 on Build GRCh38
1686804 c.321-2A>C p.? Chr.20: 33239801-33239801 on Build GRCh38
3740085 c.428C>T p.T143M Chr.20: 33239910-33239910 on Build GRCh38
5449410 c.364C>T p.Q122* Chr.20: 33239846-33239846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6120180 Chr.20: 33239840-33239840 on Build GRCh38
rs577746952 Chr.20: 33239933-33239933 on Build GRCh38
rs201372901 Chr.20: 33239896-33239896 on Build GRCh38
rs572530902 Chr.20: 33240009-33240009 on Build GRCh38
rs914267502 Chr.20: 33239978-33239978 on Build GRCh38
rs543740138 Chr.20: 33240054-33240054 on Build GRCh38
rs763357921 Chr.20: 33239767-33239767 on Build GRCh38
rs375711911 Chr.20: 33239839-33239839 on Build GRCh38
rs746872004 Chr.20: 33239907-33239907 on Build GRCh38
rs202095042 Chr.20: 33239776-33239776 on Build GRCh38
rs766661598 Chr.20: 33239871-33239871 on Build GRCh38
rs772751120 Chr.20: 33239927-33239927 on Build GRCh38
rs777549818 Chr.20: 33239825-33239825 on Build GRCh38
rs1008259136 Chr.20: 33239839-33239839 on Build GRCh38
rs1004458368 Chr.20: 33240061-33240061 on Build GRCh38
rs142118863 Chr.20: 33239858-33239858 on Build GRCh38
rs147665868 Chr.20: 33240035-33240035 on Build GRCh38
rs766679407 Chr.20: 33239929-33239929 on Build GRCh38
rs750605916 Chr.20: 33239800-33239800 on Build GRCh38
rs745341631 Chr.20: 33239843-33239843 on Build GRCh38
rs772064828 Chr.20: 33240140-33240140 on Build GRCh38
rs765297141 Chr.20: 33239936-33239936 on Build GRCh38
rs200285879 Chr.20: 33239925-33239925 on Build GRCh38
rs369979063 Chr.20: 33239949-33239949 on Build GRCh38
rs746837151 Chr.20: 33239903-33239910 on Build GRCh38
rs773414927 Chr.20: 33239928-33239928 on Build GRCh38
rs774412988 Chr.20: 33240072-33240072 on Build GRCh38
rs776271566 Chr.20: 33239763-33239763 on Build GRCh38
rs533550245 Chr.20: 33239887-33239887 on Build GRCh38
rs933956436 Chr.20: 33239986-33239986 on Build GRCh38
rs138917166 Chr.20: 33239889-33239889 on Build GRCh38
rs778785291 Chr.20: 33239806-33239806 on Build GRCh38
rs767985920 Chr.20: 33239864-33239864 on Build GRCh38
rs774941156 Chr.20: 33239850-33239850 on Build GRCh38
rs1049716541 Chr.20: 33240007-33240007 on Build GRCh38
rs567352932 Chr.20: 33240118-33240118 on Build GRCh38
rs777531573 Chr.20: 33239899-33239899 on Build GRCh38
rs62209724 Chr.20: 33239863-33239863 on Build GRCh38
rs770280558 Chr.20: 33239761-33239761 on Build GRCh38
rs372326541 Chr.20: 33239810-33239810 on Build GRCh38
rs758134197 Chr.20: 33239818-33239818 on Build GRCh38
rs996967534 Chr.20: 33240016-33240016 on Build GRCh38
rs751180637 Chr.20: 33239869-33239869 on Build GRCh38
rs371600051 Chr.20: 33239865-33239865 on Build GRCh38
rs183459848 Chr.20: 33240124-33240124 on Build GRCh38
rs776859002 Chr.20: 33239931-33239931 on Build GRCh38
rs770729858 Chr.20: 33240194-33240195 on Build GRCh38
rs779685835 Chr.20: 33240185-33240185 on Build GRCh38
rs922615327 Chr.20: 33239984-33239984 on Build GRCh38
rs866874899 Chr.20: 33240075-33240075 on Build GRCh38
rs115838258 Chr.20: 33239950-33239950 on Build GRCh38
rs375024916 Chr.20: 33239913-33239913 on Build GRCh38
rs772819924 Chr.20: 33239826-33239826 on Build GRCh38
rs2738314 Chr.20: 33239914-33239914 on Build GRCh38
rs866550342 Chr.20: 33239905-33239905 on Build GRCh38
rs543084602 Chr.20: 33240034-33240034 on Build GRCh38
rs903065131 Chr.20: 33239812-33239812 on Build GRCh38
rs754843094 Chr.20: 33239805-33239805 on Build GRCh38
rs191104003 Chr.20: 33239954-33239954 on Build GRCh38
rs551074214 Chr.20: 33239837-33239837 on Build GRCh38
rs200300856 Chr.20: 33239781-33239781 on Build GRCh38
rs906201582 Chr.20: 33240199-33240199 on Build GRCh38
rs888953040 Chr.20: 33239796-33239796 on Build GRCh38
rs756422518 Chr.20: 33239801-33239801 on Build GRCh38
rs377378402 Chr.20: 33240202-33240202 on Build GRCh38
rs763660109 Chr.20: 33240006-33240006 on Build GRCh38
rs775206851 Chr.20: 33239924-33239924 on Build GRCh38
rs762856637 Chr.20: 33239944-33239944 on Build GRCh38
rs765317724 Chr.20: 33239888-33239888 on Build GRCh38
rs150989115 Chr.20: 33239848-33239848 on Build GRCh38
rs373565061 Chr.20: 33239938-33239938 on Build GRCh38
rs763681505 Chr.20: 33239824-33239824 on Build GRCh38
rs924415963 Chr.20: 33239988-33239988 on Build GRCh38
rs764134147 Chr.20: 33239946-33239946 on Build GRCh38
rs180752131 Chr.20: 33239911-33239911 on Build GRCh38
rs768310988 Chr.20: 33240196-33240196 on Build GRCh38
rs532895600 Chr.20: 33239803-33239803 on Build GRCh38
rs757780467 Chr.20: 33240033-33240033 on Build GRCh38
rs146179198 Chr.20: 33239874-33239874 on Build GRCh38
rs62209725 Chr.20: 33240064-33240064 on Build GRCh38
rs758397951 Chr.20: 33239892-33239892 on Build GRCh38
rs114381669 Chr.20: 33240039-33240039 on Build GRCh38
rs748844198 Chr.20: 33240193-33240193 on Build GRCh38
rs748198526 Chr.20: 33240121-33240121 on Build GRCh38
rs6120181 Chr.20: 33239853-33239853 on Build GRCh38
rs370348230 Chr.20: 33239783-33239783 on Build GRCh38
rs186232151 Chr.20: 33239920-33239920 on Build GRCh38
rs142723109 Chr.20: 33239910-33239910 on Build GRCh38
rs753996528 Chr.20: 33239804-33239804 on Build GRCh38
rs752141661 Chr.20: 33239779-33239779 on Build GRCh38
rs112287711 Chr.20: 33239976-33239976 on Build GRCh38
rs978556228 Chr.20: 33239983-33239983 on Build GRCh38

View Full Product Details